Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 78935710-78936217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRAK1BP1
Entrez Gene ID
134728
Gene Name
interleukin 1 receptor associated kinase 1 binding protein 1
Gene Aliases
AIP70, SIMPL
UniGene
Hs.656212
Gene Chromosome Location
Chr.6: 78867486-78979411 on Build GRCh38
Transcripts
NM_017934.5

Gene Symbol
PHIP
Entrez Gene ID
55023
Gene Name
pleckstrin homology domain interacting protein
Gene Aliases
BRWD2, DCAF14, WDR11, ndrp
UniGene
Hs.511817
Gene Chromosome Location
Chr.6: 78934419-79078294 on Build GRCh38
Transcripts
NM_017934.5

SNP Detail

SNP ID Chromosome Location
rs763846179 Chr.6: 78935834-78935834 on Build GRCh38
rs867989443 Chr.6: 78936108-78936108 on Build GRCh38
rs916239810 Chr.6: 78935828-78935828 on Build GRCh38
rs559624004 Chr.6: 78936114-78936115 on Build GRCh38
rs754227929 Chr.6: 78935945-78935945 on Build GRCh38
rs893772229 Chr.6: 78935783-78935783 on Build GRCh38
rs935168305 Chr.6: 78936002-78936002 on Build GRCh38
rs999046834 Chr.6: 78935914-78935914 on Build GRCh38
rs990694113 Chr.6: 78936133-78936133 on Build GRCh38
rs1047171134 Chr.6: 78936161-78936161 on Build GRCh38
rs575179530 Chr.6: 78936058-78936058 on Build GRCh38
rs1054745370 Chr.6: 78935784-78935784 on Build GRCh38
rs948050546 Chr.6: 78935791-78935791 on Build GRCh38
rs927919924 Chr.6: 78935742-78935742 on Build GRCh38
rs887318408 Chr.6: 78935867-78935867 on Build GRCh38
rs939438240 Chr.6: 78936100-78936100 on Build GRCh38
rs974833771 Chr.6: 78935956-78935956 on Build GRCh38
rs147482190 Chr.6: 78936060-78936060 on Build GRCh38
rs1054145982 Chr.6: 78935767-78935767 on Build GRCh38
rs187416759 Chr.6: 78935935-78935935 on Build GRCh38
rs902225885 Chr.6: 78935883-78935883 on Build GRCh38
rs761533852 Chr.6: 78935969-78935969 on Build GRCh38
rs1025752376 Chr.6: 78935852-78935852 on Build GRCh38
rs923760964 Chr.6: 78935999-78935999 on Build GRCh38
rs1005796004 Chr.6: 78935889-78935889 on Build GRCh38
rs776537251 Chr.6: 78935953-78935953 on Build GRCh38
rs961734842 Chr.6: 78936079-78936081 on Build GRCh38
rs144369887 Chr.6: 78936109-78936109 on Build GRCh38
rs538835079 Chr.6: 78936012-78936012 on Build GRCh38
rs948050002 Chr.6: 78936038-78936038 on Build GRCh38
rs139983593 Chr.6: 78936104-78936104 on Build GRCh38
rs906638015 Chr.6: 78935831-78935831 on Build GRCh38
rs765457263 Chr.6: 78935988-78935988 on Build GRCh38
rs1044313 Chr.6: 78935903-78935903 on Build GRCh38
rs998831685 Chr.6: 78935994-78935994 on Build GRCh38
rs1028718869 Chr.6: 78935968-78935968 on Build GRCh38
rs760663933 Chr.6: 78935824-78935824 on Build GRCh38
rs757727780 Chr.6: 78936087-78936091 on Build GRCh38
rs558369284 Chr.6: 78936028-78936028 on Build GRCh38

View Full Product Details