Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 83512366-83512879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS35
Entrez Gene ID
167681
Gene Name
protease, serine 35
Gene Aliases
C6orf158, dJ223E3.1
UniGene
Hs.98381
Gene Chromosome Location
Chr.6: 83512475-83525704 on Build GRCh38
Transcripts
NM_001170423.1 NM_153362.2

SNP Detail

SNP ID Chromosome Location
rs919773603 Chr.6: 83512560-83512561 on Build GRCh38
rs931120941 Chr.6: 83512823-83512823 on Build GRCh38
rs796121354 Chr.6: 83512431-83512431 on Build GRCh38
rs945004282 Chr.6: 83512513-83512513 on Build GRCh38
rs886500108 Chr.6: 83512790-83512790 on Build GRCh38
rs987072770 Chr.6: 83512683-83512683 on Build GRCh38
rs752576817 Chr.6: 83512568-83512568 on Build GRCh38
rs1025218929 Chr.6: 83512449-83512449 on Build GRCh38
rs543588568 Chr.6: 83512780-83512780 on Build GRCh38
rs912450739 Chr.6: 83512533-83512533 on Build GRCh38
rs1171108 Chr.6: 83512554-83512554 on Build GRCh38
rs958330475 Chr.6: 83512483-83512483 on Build GRCh38
rs755201921 Chr.6: 83512800-83512800 on Build GRCh38
rs1032883588 Chr.6: 83512650-83512650 on Build GRCh38
rs920779091 Chr.6: 83512728-83512728 on Build GRCh38
rs183291108 Chr.6: 83512529-83512529 on Build GRCh38
rs980578311 Chr.6: 83512388-83512388 on Build GRCh38
rs1039200132 Chr.6: 83512531-83512531 on Build GRCh38
rs995267545 Chr.6: 83512386-83512386 on Build GRCh38
rs541621769 Chr.6: 83512629-83512629 on Build GRCh38
rs555924925 Chr.6: 83512791-83512791 on Build GRCh38
rs188672668 Chr.6: 83512713-83512713 on Build GRCh38
rs556994236 Chr.6: 83512539-83512539 on Build GRCh38
rs957903992 Chr.6: 83512858-83512858 on Build GRCh38
rs1045791400 Chr.6: 83512829-83512829 on Build GRCh38
rs967012711 Chr.6: 83512452-83512452 on Build GRCh38
rs574469276 Chr.6: 83512818-83512818 on Build GRCh38
rs773885102 Chr.6: 83512548-83512548 on Build GRCh38
rs565635092 Chr.6: 83512413-83512413 on Build GRCh38
rs926431983 Chr.6: 83512390-83512390 on Build GRCh38
rs556139285 Chr.6: 83512612-83512612 on Build GRCh38
rs954699048 Chr.6: 83512530-83512530 on Build GRCh38
rs1033975342 Chr.6: 83512854-83512854 on Build GRCh38
rs139921826 Chr.6: 83512516-83512516 on Build GRCh38
rs913606407 Chr.6: 83512505-83512505 on Build GRCh38
rs1046085416 Chr.6: 83512594-83512594 on Build GRCh38
rs533033506 Chr.6: 83512577-83512577 on Build GRCh38
rs958228040 Chr.6: 83512656-83512656 on Build GRCh38
rs371203206 Chr.6: 83512540-83512540 on Build GRCh38
rs902737395 Chr.6: 83512609-83512609 on Build GRCh38
rs899323825 Chr.6: 83512534-83512534 on Build GRCh38
rs894926426 Chr.6: 83512583-83512583 on Build GRCh38
rs1054988947 Chr.6: 83512832-83512832 on Build GRCh38
rs1054757228 Chr.6: 83512409-83512409 on Build GRCh38
rs911537691 Chr.6: 83512688-83512688 on Build GRCh38
rs936461686 Chr.6: 83512396-83512396 on Build GRCh38
rs1024915317 Chr.6: 83512572-83512572 on Build GRCh38
rs1039693093 Chr.6: 83512721-83512721 on Build GRCh38
rs760670942 Chr.6: 83512589-83512589 on Build GRCh38
rs966359793 Chr.6: 83512676-83512676 on Build GRCh38
rs1032526637 Chr.6: 83512461-83512461 on Build GRCh38
rs894811830 Chr.6: 83512428-83512428 on Build GRCh38
rs958107835 Chr.6: 83512616-83512616 on Build GRCh38
rs961555195 Chr.6: 83512798-83512798 on Build GRCh38
rs1033551322 Chr.6: 83512592-83512592 on Build GRCh38
rs551249428 Chr.6: 83512486-83512486 on Build GRCh38
rs564486577 Chr.6: 83512631-83512631 on Build GRCh38
rs1047821372 Chr.6: 83512767-83512767 on Build GRCh38
rs930774543 Chr.6: 83512756-83512756 on Build GRCh38
rs112550597 Chr.6: 83512853-83512853 on Build GRCh38
rs1013292969 Chr.6: 83512436-83512436 on Build GRCh38
rs905853447 Chr.6: 83512834-83512834 on Build GRCh38
rs572730422 Chr.6: 83512624-83512624 on Build GRCh38
rs481687 Chr.6: 83512458-83512458 on Build GRCh38
rs1051972999 Chr.6: 83512515-83512515 on Build GRCh38
rs974799948 Chr.6: 83512494-83512494 on Build GRCh38
rs36079295 Chr.6: 83512670-83512670 on Build GRCh38
rs886452189 Chr.6: 83512565-83512565 on Build GRCh38
rs894324422 Chr.6: 83512439-83512439 on Build GRCh38
rs989626597 Chr.6: 83512410-83512410 on Build GRCh38
rs866099126 Chr.6: 83512604-83512604 on Build GRCh38
rs767449743 Chr.6: 83512567-83512567 on Build GRCh38

View Full Product Details