Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 83525367-83525869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS35
Entrez Gene ID
167681
Gene Name
protease, serine 35
Gene Aliases
C6orf158, dJ223E3.1
UniGene
Hs.98381
Gene Chromosome Location
Chr.6: 83512475-83525704 on Build GRCh38
Transcripts
NM_001170423.1 NM_153362.2

SNP Detail

SNP ID Chromosome Location
rs866693343 Chr.6: 83525819-83525819 on Build GRCh38
rs556822334 Chr.6: 83525835-83525835 on Build GRCh38
rs895355545 Chr.6: 83525674-83525674 on Build GRCh38
rs911909386 Chr.6: 83525789-83525789 on Build GRCh38
rs931455283 Chr.6: 83525591-83525591 on Build GRCh38
rs554514148 Chr.6: 83525456-83525456 on Build GRCh38
rs767373752 Chr.6: 83525528-83525528 on Build GRCh38
rs893060834 Chr.6: 83525828-83525828 on Build GRCh38
rs973379882 Chr.6: 83525557-83525557 on Build GRCh38
rs987522226 Chr.6: 83525785-83525785 on Build GRCh38
rs989701348 Chr.6: 83525592-83525592 on Build GRCh38
rs1033958435 Chr.6: 83525543-83525543 on Build GRCh38
rs1003878432 Chr.6: 83525490-83525490 on Build GRCh38
rs1021540045 Chr.6: 83525632-83525632 on Build GRCh38
rs117011725 Chr.6: 83525555-83525555 on Build GRCh38
rs1011376216 Chr.6: 83525834-83525834 on Build GRCh38
rs970336203 Chr.6: 83525500-83525500 on Build GRCh38
rs541372459 Chr.6: 83525810-83525810 on Build GRCh38
rs577577727 Chr.6: 83525497-83525497 on Build GRCh38
rs920209696 Chr.6: 83525576-83525579 on Build GRCh38
rs1055229574 Chr.6: 83525611-83525611 on Build GRCh38
rs949460984 Chr.6: 83525700-83525700 on Build GRCh38
rs574000478 Chr.6: 83525487-83525487 on Build GRCh38
rs958517579 Chr.6: 83525584-83525584 on Build GRCh38
rs770641115 Chr.6: 83525788-83525788 on Build GRCh38
rs77746135 Chr.6: 83525410-83525410 on Build GRCh38
rs966789511 Chr.6: 83525682-83525682 on Build GRCh38
rs753590991 Chr.6: 83525602-83525602 on Build GRCh38
rs975131828 Chr.6: 83525814-83525814 on Build GRCh38
rs995376078 Chr.6: 83525394-83525394 on Build GRCh38
rs886900983 Chr.6: 83525450-83525450 on Build GRCh38

View Full Product Details