Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89327640-89328185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2J1
Entrez Gene ID
51465
Gene Name
ubiquitin conjugating enzyme E2 J1
Gene Aliases
CGI-76, HSPC153, HSPC205, HSU93243, NCUBE-1, NCUBE1, UBC6, UBC6E, Ubc6p
UniGene
Hs.163776
Gene Chromosome Location
Chr.6: 89326625-89352917 on Build GRCh38
Transcripts
NM_016021.2 XM_011535887.1 XM_011535888.2

SNP Detail

SNP ID Chromosome Location
rs115131734 Chr.6: 89327800-89327800 on Build GRCh38
rs984829813 Chr.6: 89328118-89328118 on Build GRCh38
rs1036868533 Chr.6: 89327886-89327886 on Build GRCh38
rs966417288 Chr.6: 89327783-89327783 on Build GRCh38
rs532403046 Chr.6: 89327797-89327797 on Build GRCh38
rs982894973 Chr.6: 89327791-89327791 on Build GRCh38
rs543554959 Chr.6: 89327720-89327720 on Build GRCh38
rs1029541560 Chr.6: 89327709-89327709 on Build GRCh38
rs960884429 Chr.6: 89328047-89328047 on Build GRCh38
rs537124323 Chr.6: 89328032-89328032 on Build GRCh38
rs752180323 Chr.6: 89327685-89327687 on Build GRCh38
rs1039118712 Chr.6: 89327950-89327950 on Build GRCh38
rs999199235 Chr.6: 89328005-89328005 on Build GRCh38
rs962128578 Chr.6: 89328161-89328161 on Build GRCh38
rs951292521 Chr.6: 89327773-89327773 on Build GRCh38
rs766006179 Chr.6: 89327912-89327912 on Build GRCh38
rs114144399 Chr.6: 89327865-89327865 on Build GRCh38
rs564914052 Chr.6: 89327754-89327754 on Build GRCh38
rs148172099 Chr.6: 89328104-89328104 on Build GRCh38
rs565856114 Chr.6: 89327804-89327804 on Build GRCh38
rs926344024 Chr.6: 89327819-89327819 on Build GRCh38
rs1022023098 Chr.6: 89327781-89327781 on Build GRCh38
rs1029448341 Chr.6: 89327985-89327985 on Build GRCh38
rs1021907221 Chr.6: 89327957-89327958 on Build GRCh38
rs901128960 Chr.6: 89327696-89327696 on Build GRCh38
rs1011089074 Chr.6: 89327744-89327744 on Build GRCh38
rs1004121592 Chr.6: 89327992-89327992 on Build GRCh38
rs992348644 Chr.6: 89328094-89328094 on Build GRCh38
rs1017235531 Chr.6: 89328013-89328013 on Build GRCh38
rs980807140 Chr.6: 89328080-89328080 on Build GRCh38
rs998073441 Chr.6: 89327703-89327703 on Build GRCh38
rs992296113 Chr.6: 89327855-89327855 on Build GRCh38
rs933913678 Chr.6: 89327922-89327922 on Build GRCh38
rs998008200 Chr.6: 89327956-89327956 on Build GRCh38
rs887049188 Chr.6: 89327986-89327986 on Build GRCh38
rs903864459 Chr.6: 89327917-89327917 on Build GRCh38
rs187177422 Chr.6: 89328031-89328031 on Build GRCh38
rs960438847 Chr.6: 89328085-89328085 on Build GRCh38
rs939505055 Chr.6: 89327823-89327823 on Build GRCh38
rs551372913 Chr.6: 89328084-89328084 on Build GRCh38
rs148982987 Chr.6: 89327821-89327821 on Build GRCh38
rs143798378 Chr.6: 89327939-89327939 on Build GRCh38
rs901900869 Chr.6: 89327952-89327952 on Build GRCh38
rs760307417 Chr.6: 89327892-89327892 on Build GRCh38
rs962426009 Chr.6: 89328050-89328050 on Build GRCh38
rs9048 Chr.6: 89327862-89327862 on Build GRCh38
rs571351473 Chr.6: 89328153-89328153 on Build GRCh38
rs1023919500 Chr.6: 89328102-89328102 on Build GRCh38
rs914194677 Chr.6: 89327856-89327856 on Build GRCh38

View Full Product Details