Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 95606329-95606852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MANEA
Entrez Gene ID
79694
Gene Name
mannosidase endo-alpha
Gene Aliases
ENDO, hEndo
UniGene
Hs.734632
Gene Chromosome Location
Chr.6: 95577497-95609452 on Build GRCh38
Transcripts
NM_024641.3 XM_005267147.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
232751 c.1348A>G p.R450G Chr.6: 95606364-95606364 on Build GRCh38
4999848 c.1319G>A p.R440H Chr.6: 95606335-95606335 on Build GRCh38
1082688 c.1318C>T p.R440C Chr.6: 95606334-95606334 on Build GRCh38
6706723 c.1320C>T p.R440R Chr.6: 95606336-95606336 on Build GRCh38
5034692 c.1361C>T p.A454V Chr.6: 95606377-95606377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965720011 Chr.6: 95606463-95606463 on Build GRCh38
rs75510775 Chr.6: 95606386-95606386 on Build GRCh38
rs1021468891 Chr.6: 95606776-95606776 on Build GRCh38
rs527258577 Chr.6: 95606538-95606538 on Build GRCh38
rs748564585 Chr.6: 95606410-95606410 on Build GRCh38
rs777837314 Chr.6: 95606584-95606584 on Build GRCh38
rs771205447 Chr.6: 95606442-95606442 on Build GRCh38
rs11153709 Chr.6: 95606693-95606693 on Build GRCh38
rs763705331 Chr.6: 95606455-95606455 on Build GRCh38
rs181498694 Chr.6: 95606614-95606614 on Build GRCh38
rs1004655251 Chr.6: 95606461-95606461 on Build GRCh38
rs991251138 Chr.6: 95606600-95606600 on Build GRCh38
rs966355788 Chr.6: 95606669-95606669 on Build GRCh38
rs139275538 Chr.6: 95606353-95606353 on Build GRCh38
rs893890902 Chr.6: 95606765-95606765 on Build GRCh38
rs548612249 Chr.6: 95606745-95606745 on Build GRCh38
rs780620663 Chr.6: 95606445-95606445 on Build GRCh38
rs996902383 Chr.6: 95606494-95606494 on Build GRCh38
rs1133503 Chr.6: 95606712-95606712 on Build GRCh38
rs551660634 Chr.6: 95606649-95606650 on Build GRCh38
rs1055513716 Chr.6: 95606390-95606390 on Build GRCh38
rs757917387 Chr.6: 95606384-95606384 on Build GRCh38
rs117982267 Chr.6: 95606378-95606378 on Build GRCh38
rs887240363 Chr.6: 95606647-95606647 on Build GRCh38
rs117238191 Chr.6: 95606774-95606774 on Build GRCh38
rs9401034 Chr.6: 95606664-95606664 on Build GRCh38
rs760746636 Chr.6: 95606449-95606449 on Build GRCh38
rs9374688 Chr.6: 95606533-95606533 on Build GRCh38
rs967315260 Chr.6: 95606554-95606554 on Build GRCh38
rs778056777 Chr.6: 95606431-95606431 on Build GRCh38
rs774544685 Chr.6: 95606443-95606443 on Build GRCh38
rs762624280 Chr.6: 95606414-95606420 on Build GRCh38
rs977774021 Chr.6: 95606681-95606681 on Build GRCh38
rs1016288875 Chr.6: 95606596-95606596 on Build GRCh38
rs768737935 Chr.6: 95606456-95606456 on Build GRCh38
rs770010197 Chr.6: 95606421-95606421 on Build GRCh38
rs781457807 Chr.6: 95606409-95606409 on Build GRCh38
rs1055335138 Chr.6: 95606525-95606526 on Build GRCh38
rs764902952 Chr.6: 95606529-95606529 on Build GRCh38
rs755446383 Chr.6: 95606397-95606397 on Build GRCh38
rs80216717 Chr.6: 95606825-95606825 on Build GRCh38
rs749513152 Chr.6: 95606432-95606432 on Build GRCh38
rs764973822 Chr.6: 95606382-95606382 on Build GRCh38
rs188735649 Chr.6: 95606532-95606532 on Build GRCh38
rs749049377 Chr.6: 95606738-95606738 on Build GRCh38
rs146501555 Chr.6: 95606385-95606385 on Build GRCh38
rs1048547565 Chr.6: 95606644-95606644 on Build GRCh38
rs989924588 Chr.6: 95606535-95606535 on Build GRCh38
rs560191537 Chr.6: 95606362-95606362 on Build GRCh38
rs1041667183 Chr.6: 95606670-95606670 on Build GRCh38
rs531584796 Chr.6: 95606526-95606526 on Build GRCh38
rs1048707023 Chr.6: 95606450-95606450 on Build GRCh38
rs1022002588 Chr.6: 95606543-95606543 on Build GRCh38
rs567363135 Chr.6: 95606580-95606580 on Build GRCh38
rs543483790 Chr.6: 95606462-95606462 on Build GRCh38
rs139923844 Chr.6: 95606568-95606568 on Build GRCh38
rs931508099 Chr.6: 95606629-95606629 on Build GRCh38
rs555950996 Chr.6: 95606655-95606655 on Build GRCh38
rs560543699 Chr.6: 95606457-95606457 on Build GRCh38

View Full Product Details