Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 96526184-96526707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UFL1
Entrez Gene ID
23376
Gene Name
UFM1 specific ligase 1
Gene Aliases
KIAA0776, Maxer, NLBP, RCAD
UniGene
Hs.149367
Gene Chromosome Location
Chr.6: 96521826-96555276 on Build GRCh38
Transcripts
NM_015323.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7244263 c.369G>T p.L123F Chr.6: 96526339-96526339 on Build GRCh38
5079744 c.465+4delT p.? Chr.6: 96526439-96526439 on Build GRCh38
5701761 c.449_450GG>TT p.G150V Chr.6: 96526419-96526420 on Build GRCh38
3876234 c.429T>C p.C143C Chr.6: 96526399-96526399 on Build GRCh38
1471966 c.463C>G p.Q155E Chr.6: 96526433-96526433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915862999 Chr.6: 96526608-96526608 on Build GRCh38
rs774612428 Chr.6: 96526297-96526297 on Build GRCh38
rs112477727 Chr.6: 96526209-96526209 on Build GRCh38
rs1016374622 Chr.6: 96526533-96526533 on Build GRCh38
rs758570081 Chr.6: 96526438-96526439 on Build GRCh38
rs368118548 Chr.6: 96526335-96526335 on Build GRCh38
rs375574942 Chr.6: 96526275-96526275 on Build GRCh38
rs369510193 Chr.6: 96526449-96526449 on Build GRCh38
rs199790595 Chr.6: 96526451-96526451 on Build GRCh38
rs1004864550 Chr.6: 96526557-96526557 on Build GRCh38
rs141559002 Chr.6: 96526241-96526241 on Build GRCh38
rs181630674 Chr.6: 96526511-96526511 on Build GRCh38
rs989878279 Chr.6: 96526431-96526431 on Build GRCh38
rs768608472 Chr.6: 96526433-96526433 on Build GRCh38
rs759134279 Chr.6: 96526570-96526570 on Build GRCh38
rs779277736 Chr.6: 96526407-96526409 on Build GRCh38
rs150559148 Chr.6: 96526326-96526326 on Build GRCh38
rs778097329 Chr.6: 96526287-96526287 on Build GRCh38
rs139586367 Chr.6: 96526408-96526408 on Build GRCh38
rs543563242 Chr.6: 96526580-96526580 on Build GRCh38
rs755706321 Chr.6: 96526470-96526470 on Build GRCh38
rs976325264 Chr.6: 96526385-96526385 on Build GRCh38
rs1049136556 Chr.6: 96526517-96526517 on Build GRCh38
rs773270637 Chr.6: 96526436-96526436 on Build GRCh38
rs775664571 Chr.6: 96526323-96526323 on Build GRCh38
rs758177640 Chr.6: 96526610-96526610 on Build GRCh38
rs751295138 Chr.6: 96526351-96526351 on Build GRCh38
rs772191237 Chr.6: 96526316-96526316 on Build GRCh38
rs376534666 Chr.6: 96526438-96526438 on Build GRCh38
rs778357058 Chr.6: 96526390-96526390 on Build GRCh38
rs938550511 Chr.6: 96526508-96526508 on Build GRCh38
rs771349140 Chr.6: 96526399-96526399 on Build GRCh38
rs752233719 Chr.6: 96526463-96526463 on Build GRCh38
rs754812782 Chr.6: 96526288-96526288 on Build GRCh38
rs987538446 Chr.6: 96526618-96526618 on Build GRCh38
rs374738871 Chr.6: 96526299-96526299 on Build GRCh38
rs146188768 Chr.6: 96526334-96526334 on Build GRCh38
rs757784684 Chr.6: 96526487-96526487 on Build GRCh38
rs749743876 Chr.6: 96526397-96526397 on Build GRCh38
rs886521741 Chr.6: 96526516-96526516 on Build GRCh38
rs1055148339 Chr.6: 96526510-96526511 on Build GRCh38
rs766943351 Chr.6: 96526537-96526537 on Build GRCh38
rs753728518 Chr.6: 96526278-96526285 on Build GRCh38
rs372729271 Chr.6: 96526240-96526240 on Build GRCh38
rs967518872 Chr.6: 96526668-96526668 on Build GRCh38
rs773789156 Chr.6: 96526582-96526582 on Build GRCh38
rs754651588 Chr.6: 96526361-96526361 on Build GRCh38
rs752907757 Chr.6: 96526439-96526439 on Build GRCh38
rs749664618 Chr.6: 96526292-96526292 on Build GRCh38
rs748303113 Chr.6: 96526239-96526239 on Build GRCh38
rs754394358 Chr.6: 96526486-96526486 on Build GRCh38
rs1020065206 Chr.6: 96526667-96526667 on Build GRCh38
rs372624410 Chr.6: 96526579-96526579 on Build GRCh38
rs937222553 Chr.6: 96526510-96526510 on Build GRCh38
rs748651240 Chr.6: 96526270-96526270 on Build GRCh38
rs367987328 Chr.6: 96526480-96526480 on Build GRCh38
rs938329748 Chr.6: 96526603-96526603 on Build GRCh38
rs966033414 Chr.6: 96526569-96526569 on Build GRCh38
rs779229392 Chr.6: 96526404-96526404 on Build GRCh38
rs767203299 Chr.6: 96526461-96526461 on Build GRCh38
rs886963097 Chr.6: 96526683-96526683 on Build GRCh38
rs977211120 Chr.6: 96526596-96526596 on Build GRCh38
rs777867853 Chr.6: 96526452-96526453 on Build GRCh38
rs1048725053 Chr.6: 96526613-96526613 on Build GRCh38
rs1055716563 Chr.6: 96526607-96526607 on Build GRCh38
rs139058346 Chr.6: 96526572-96526572 on Build GRCh38
rs576567834 Chr.6: 96526383-96526383 on Build GRCh38
rs751419690 Chr.6: 96526455-96526455 on Build GRCh38
rs28372909 Chr.6: 96526379-96526379 on Build GRCh38
rs887885803 Chr.6: 96526549-96526549 on Build GRCh38
rs9320194 Chr.6: 96526510-96526510 on Build GRCh38
rs759360534 Chr.6: 96526458-96526458 on Build GRCh38
rs771224323 Chr.6: 96526296-96526296 on Build GRCh38
rs775509242 Chr.6: 96526411-96526411 on Build GRCh38
rs928250940 Chr.6: 96526601-96526601 on Build GRCh38
rs925888443 Chr.6: 96526504-96526505 on Build GRCh38
rs776956293 Chr.6: 96526560-96526560 on Build GRCh38
rs1015317742 Chr.6: 96526558-96526558 on Build GRCh38
rs376029557 Chr.6: 96526242-96526242 on Build GRCh38
rs1046438310 Chr.6: 96526512-96526512 on Build GRCh38
rs561207317 Chr.6: 96526514-96526514 on Build GRCh38
rs765224813 Chr.6: 96526328-96526328 on Build GRCh38
rs953545552 Chr.6: 96526575-96526575 on Build GRCh38
rs199639976 Chr.6: 96526482-96526482 on Build GRCh38
rs886575212 Chr.6: 96526393-96526393 on Build GRCh38
rs963591477 Chr.6: 96526414-96526414 on Build GRCh38
rs760826756 Chr.6: 96526423-96526423 on Build GRCh38
rs780655887 Chr.6: 96526368-96526368 on Build GRCh38

View Full Product Details