Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 97143191-97143697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMS22L
Entrez Gene ID
253714
Gene Name
MMS22 like, DNA repair protein
Gene Aliases
C6orf167, dJ39B17.2
UniGene
Hs.444292
Gene Chromosome Location
Chr.6: 97142161-97283437 on Build GRCh38
Transcripts
NM_198468.2 XM_006715430.1

SNP Detail

SNP ID Chromosome Location
rs62413910 Chr.6: 97143366-97143366 on Build GRCh38
rs181164455 Chr.6: 97143382-97143382 on Build GRCh38
rs890081877 Chr.6: 97143464-97143464 on Build GRCh38
rs1031533091 Chr.6: 97143558-97143558 on Build GRCh38
rs374878872 Chr.6: 97143557-97143557 on Build GRCh38
rs1007109773 Chr.6: 97143473-97143473 on Build GRCh38
rs577693905 Chr.6: 97143577-97143577 on Build GRCh38
rs9384858 Chr.6: 97143631-97143631 on Build GRCh38
rs947968097 Chr.6: 97143327-97143327 on Build GRCh38
rs555699484 Chr.6: 97143295-97143295 on Build GRCh38
rs770382683 Chr.6: 97143471-97143471 on Build GRCh38
rs957511148 Chr.6: 97143358-97143358 on Build GRCh38
rs931130019 Chr.6: 97143582-97143585 on Build GRCh38
rs560134637 Chr.6: 97143590-97143590 on Build GRCh38
rs931181982 Chr.6: 97143259-97143259 on Build GRCh38
rs1037234649 Chr.6: 97143234-97143234 on Build GRCh38
rs908898133 Chr.6: 97143664-97143664 on Build GRCh38
rs978099308 Chr.6: 97143547-97143547 on Build GRCh38
rs940055984 Chr.6: 97143527-97143527 on Build GRCh38
rs970351252 Chr.6: 97143407-97143407 on Build GRCh38
rs11961600 Chr.6: 97143305-97143305 on Build GRCh38
rs544432304 Chr.6: 97143432-97143432 on Build GRCh38
rs1036499862 Chr.6: 97143370-97143370 on Build GRCh38
rs1008279604 Chr.6: 97143299-97143299 on Build GRCh38
rs879181390 Chr.6: 97143389-97143390 on Build GRCh38
rs1032126346 Chr.6: 97143355-97143356 on Build GRCh38
rs565089700 Chr.6: 97143588-97143588 on Build GRCh38
rs998386272 Chr.6: 97143386-97143386 on Build GRCh38
rs887405249 Chr.6: 97143364-97143364 on Build GRCh38
rs968428581 Chr.6: 97143493-97143493 on Build GRCh38
rs898332569 Chr.6: 97143250-97143250 on Build GRCh38
rs972835886 Chr.6: 97143536-97143538 on Build GRCh38
rs943971863 Chr.6: 97143625-97143625 on Build GRCh38
rs117592459 Chr.6: 97143668-97143668 on Build GRCh38
rs755642699 Chr.6: 97143401-97143401 on Build GRCh38
rs939210926 Chr.6: 97143275-97143275 on Build GRCh38
rs947979396 Chr.6: 97143640-97143640 on Build GRCh38
rs981817278 Chr.6: 97143421-97143421 on Build GRCh38
rs528877473 Chr.6: 97143646-97143646 on Build GRCh38
rs779627083 Chr.6: 97143516-97143516 on Build GRCh38
rs1022304827 Chr.6: 97143475-97143475 on Build GRCh38
rs112227421 Chr.6: 97143328-97143328 on Build GRCh38

View Full Product Details