Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 99546310-99546816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNC
Entrez Gene ID
892
Gene Name
cyclin C
Gene Aliases
CycC
UniGene
Hs.430646
Gene Chromosome Location
Chr.6: 99542387-99568973 on Build GRCh38
Transcripts
NM_001013399.1 NM_005190.3 XM_006715594.1

Gene Symbol
TSTD3
Entrez Gene ID
100130890
Gene Name
thiosulfate sulfurtransferase (rhodanese)-like domain containing 3
Gene Aliases
-
UniGene
Hs.634506
Gene Chromosome Location
Chr.6: 99424854-99579288 on Build GRCh38
Transcripts
NM_001013399.1 NM_005190.3 XM_006715594.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3949567 c.628C>T p.Q210* Chr.6: 99546445-99546445 on Build GRCh38
6107472 c.625C>T p.Q209* Chr.6: 99546448-99546448 on Build GRCh38
3080131 c.637G>A p.A213T Chr.6: 99546436-99546436 on Build GRCh38
1312466 c.634G>C p.D212H Chr.6: 99546439-99546439 on Build GRCh38
1082833 c.605T>C p.L202P Chr.6: 99546468-99546468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189565121 Chr.6: 99546680-99546680 on Build GRCh38
rs547996448 Chr.6: 99546719-99546720 on Build GRCh38
rs777556400 Chr.6: 99546376-99546376 on Build GRCh38
rs770254174 Chr.6: 99546494-99546494 on Build GRCh38
rs372641047 Chr.6: 99546525-99546525 on Build GRCh38
rs376505239 Chr.6: 99546366-99546366 on Build GRCh38
rs540253843 Chr.6: 99546380-99546380 on Build GRCh38
rs767255197 Chr.6: 99546346-99546346 on Build GRCh38
rs971257340 Chr.6: 99546712-99546712 on Build GRCh38
rs373726776 Chr.6: 99546561-99546561 on Build GRCh38
rs370991295 Chr.6: 99546538-99546538 on Build GRCh38
rs758122333 Chr.6: 99546498-99546498 on Build GRCh38
rs919338803 Chr.6: 99546694-99546694 on Build GRCh38
rs370895238 Chr.6: 99546757-99546757 on Build GRCh38
rs926612379 Chr.6: 99546334-99546334 on Build GRCh38
rs141737331 Chr.6: 99546630-99546630 on Build GRCh38
rs61753659 Chr.6: 99546441-99546441 on Build GRCh38
rs973912020 Chr.6: 99546656-99546656 on Build GRCh38
rs752452475 Chr.6: 99546355-99546355 on Build GRCh38
rs781634701 Chr.6: 99546460-99546460 on Build GRCh38
rs748455107 Chr.6: 99546482-99546482 on Build GRCh38
rs756015358 Chr.6: 99546367-99546367 on Build GRCh38
rs374765500 Chr.6: 99546552-99546552 on Build GRCh38
rs1010017363 Chr.6: 99546689-99546689 on Build GRCh38
rs891531887 Chr.6: 99546664-99546664 on Build GRCh38
rs531178060 Chr.6: 99546705-99546705 on Build GRCh38
rs769011849 Chr.6: 99546428-99546428 on Build GRCh38
rs752289569 Chr.6: 99546368-99546368 on Build GRCh38
rs1047873357 Chr.6: 99546493-99546493 on Build GRCh38
rs780307872 Chr.6: 99546381-99546381 on Build GRCh38
rs973827203 Chr.6: 99546337-99546337 on Build GRCh38
rs770388274 Chr.6: 99546524-99546524 on Build GRCh38
rs550733549 Chr.6: 99546639-99546639 on Build GRCh38
rs900758825 Chr.6: 99546593-99546593 on Build GRCh38
rs1054547925 Chr.6: 99546634-99546634 on Build GRCh38
rs530256650 Chr.6: 99546719-99546719 on Build GRCh38
rs929334313 Chr.6: 99546698-99546698 on Build GRCh38
rs1024135627 Chr.6: 99546702-99546702 on Build GRCh38
rs112973777 Chr.6: 99546491-99546491 on Build GRCh38
rs747179991 Chr.6: 99546403-99546403 on Build GRCh38
rs1021351124 Chr.6: 99546360-99546360 on Build GRCh38
rs374164863 Chr.6: 99546495-99546495 on Build GRCh38
rs928588063 Chr.6: 99546721-99546721 on Build GRCh38
rs9494694 Chr.6: 99546722-99546722 on Build GRCh38
rs764832664 Chr.6: 99546336-99546336 on Build GRCh38
rs1055674456 Chr.6: 99546736-99546736 on Build GRCh38
rs915745064 Chr.6: 99546740-99546740 on Build GRCh38
rs564821657 Chr.6: 99546508-99546508 on Build GRCh38
rs563565543 Chr.6: 99546623-99546623 on Build GRCh38
rs371337674 Chr.6: 99546378-99546378 on Build GRCh38
rs954934936 Chr.6: 99546504-99546504 on Build GRCh38
rs986366448 Chr.6: 99546595-99546595 on Build GRCh38
rs1018193146 Chr.6: 99546596-99546596 on Build GRCh38

View Full Product Details