Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 99551736-99552242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNC
Entrez Gene ID
892
Gene Name
cyclin C
Gene Aliases
CycC
UniGene
Hs.430646
Gene Chromosome Location
Chr.6: 99542387-99568973 on Build GRCh38
Transcripts
NM_001013399.1 NM_005190.3 XM_006715594.1 XM_011536232.2 XM_017011436.1

Gene Symbol
TSTD3
Entrez Gene ID
100130890
Gene Name
thiosulfate sulfurtransferase (rhodanese)-like domain containing 3
Gene Aliases
-
UniGene
Hs.634506
Gene Chromosome Location
Chr.6: 99424854-99579288 on Build GRCh38
Transcripts
NM_001013399.1 NM_005190.3 XM_006715594.1 XM_011536232.2 XM_017011436.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1082837 c.359T>G p.F120C Chr.6: 99551883-99551883 on Build GRCh38
1754381 c.400C>T p.H134Y Chr.6: 99551842-99551842 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs578242060 Chr.6: 99552067-99552067 on Build GRCh38
rs553737766 Chr.6: 99552029-99552029 on Build GRCh38
rs762926221 Chr.6: 99551840-99551840 on Build GRCh38
rs145717552 Chr.6: 99551966-99551966 on Build GRCh38
rs1016624749 Chr.6: 99552066-99552066 on Build GRCh38
rs768107666 Chr.6: 99551799-99551799 on Build GRCh38
rs113812281 Chr.6: 99552144-99552144 on Build GRCh38
rs972059994 Chr.6: 99552068-99552068 on Build GRCh38
rs968167958 Chr.6: 99552039-99552039 on Build GRCh38
rs545443685 Chr.6: 99552109-99552109 on Build GRCh38
rs140253488 Chr.6: 99551879-99551879 on Build GRCh38
rs768869214 Chr.6: 99551907-99551907 on Build GRCh38
rs947861151 Chr.6: 99551814-99551814 on Build GRCh38
rs1001040696 Chr.6: 99552041-99552041 on Build GRCh38
rs191842012 Chr.6: 99551910-99551910 on Build GRCh38
rs749077257 Chr.6: 99551906-99551907 on Build GRCh38
rs1029910667 Chr.6: 99552025-99552025 on Build GRCh38
rs561834082 Chr.6: 99551783-99551783 on Build GRCh38
rs776322557 Chr.6: 99551815-99551815 on Build GRCh38
rs752053894 Chr.6: 99551921-99551921 on Build GRCh38
rs1045292995 Chr.6: 99551847-99551847 on Build GRCh38
rs769674194 Chr.6: 99551831-99551831 on Build GRCh38
rs998076553 Chr.6: 99552012-99552012 on Build GRCh38
rs1042349322 Chr.6: 99551920-99551920 on Build GRCh38
rs748244281 Chr.6: 99551936-99551939 on Build GRCh38
rs779955958 Chr.6: 99551839-99551839 on Build GRCh38
rs75489406 Chr.6: 99552161-99552161 on Build GRCh38
rs763492462 Chr.6: 99551918-99551918 on Build GRCh38
rs981694147 Chr.6: 99552100-99552100 on Build GRCh38
rs969541104 Chr.6: 99552216-99552216 on Build GRCh38
rs747845760 Chr.6: 99551819-99551819 on Build GRCh38
rs990255074 Chr.6: 99552146-99552146 on Build GRCh38
rs937478025 Chr.6: 99552110-99552110 on Build GRCh38
rs372706393 Chr.6: 99552164-99552164 on Build GRCh38
rs950285438 Chr.6: 99551821-99551821 on Build GRCh38
rs754478669 Chr.6: 99551942-99551942 on Build GRCh38
rs1018449628 Chr.6: 99552030-99552030 on Build GRCh38
rs536699193 Chr.6: 99551908-99551908 on Build GRCh38
rs1024679231 Chr.6: 99552183-99552183 on Build GRCh38
rs914655674 Chr.6: 99552148-99552148 on Build GRCh38
rs760798435 Chr.6: 99551900-99551900 on Build GRCh38
rs773533886 Chr.6: 99551852-99551852 on Build GRCh38
rs370442761 Chr.6: 99551907-99551908 on Build GRCh38
rs917642558 Chr.6: 99551784-99551784 on Build GRCh38
rs957002441 Chr.6: 99552073-99552073 on Build GRCh38
rs750786536 Chr.6: 99551911-99551911 on Build GRCh38
rs758756123 Chr.6: 99551917-99551917 on Build GRCh38
rs550927032 Chr.6: 99551786-99551786 on Build GRCh38
rs764132423 Chr.6: 99551901-99551901 on Build GRCh38
rs535209145 Chr.6: 99552003-99552003 on Build GRCh38
rs746524334 Chr.6: 99551798-99551798 on Build GRCh38
rs994550736 Chr.6: 99551949-99551949 on Build GRCh38
rs367600036 Chr.6: 99551797-99551797 on Build GRCh38
rs754016354 Chr.6: 99551904-99551904 on Build GRCh38
rs546640445 Chr.6: 99552079-99552079 on Build GRCh38
rs1051125597 Chr.6: 99551952-99551952 on Build GRCh38

View Full Product Details