Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 99993839-99994341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCHR2
Entrez Gene ID
84539
Gene Name
melanin concentrating hormone receptor 2
Gene Aliases
GPR145, GPRv17, MCH-2R, MCH-R2, MCH2, MCH2R, MCHR-2, SLT
UniGene
Hs.591342
Gene Chromosome Location
Chr.6: 99919910-99994238 on Build GRCh38
Transcripts
NM_001040179.1 NM_032503.2 NR_038384.1

Gene Symbol
MCHR2-AS1
Entrez Gene ID
728012
Gene Name
MCHR2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.434366
Gene Chromosome Location
Chr.6: 99994038-100076419 on Build GRCh38
Transcripts
NM_001040179.1 NM_032503.2 NR_038384.1

SNP Detail

SNP ID Chromosome Location
rs996818102 Chr.6: 99994127-99994127 on Build GRCh38
rs559763335 Chr.6: 99994090-99994090 on Build GRCh38
rs201809052 Chr.6: 99994040-99994040 on Build GRCh38
rs991368775 Chr.6: 99994025-99994025 on Build GRCh38
rs1048122527 Chr.6: 99994150-99994150 on Build GRCh38
rs201894719 Chr.6: 99994076-99994076 on Build GRCh38
rs991701204 Chr.6: 99993918-99993918 on Build GRCh38
rs1011718518 Chr.6: 99993957-99993957 on Build GRCh38
rs969421239 Chr.6: 99993926-99993926 on Build GRCh38
rs1035373968 Chr.6: 99994271-99994271 on Build GRCh38
rs111275707 Chr.6: 99994208-99994208 on Build GRCh38
rs199972059 Chr.6: 99994099-99994099 on Build GRCh38
rs563680235 Chr.6: 99994170-99994170 on Build GRCh38
rs201972556 Chr.6: 99994133-99994133 on Build GRCh38
rs541296917 Chr.6: 99994073-99994073 on Build GRCh38
rs551751081 Chr.6: 99994158-99994158 on Build GRCh38
rs200393684 Chr.6: 99994224-99994224 on Build GRCh38
rs886880445 Chr.6: 99994196-99994196 on Build GRCh38
rs886413906 Chr.6: 99993866-99993866 on Build GRCh38
rs78827858 Chr.6: 99994047-99994047 on Build GRCh38
rs928597549 Chr.6: 99994004-99994004 on Build GRCh38
rs981327727 Chr.6: 99993987-99993987 on Build GRCh38
rs1022927366 Chr.6: 99994277-99994277 on Build GRCh38
rs745807715 Chr.6: 99994311-99994311 on Build GRCh38
rs1020263913 Chr.6: 99993860-99993882 on Build GRCh38
rs201517224 Chr.6: 99993895-99993895 on Build GRCh38
rs557354570 Chr.6: 99993956-99993956 on Build GRCh38
rs931517613 Chr.6: 99994149-99994149 on Build GRCh38
rs117399293 Chr.6: 99994066-99994066 on Build GRCh38
rs201874210 Chr.6: 99994215-99994215 on Build GRCh38
rs200512325 Chr.6: 99994057-99994057 on Build GRCh38
rs533486936 Chr.6: 99994101-99994101 on Build GRCh38
rs925393092 Chr.6: 99993976-99993976 on Build GRCh38
rs201028337 Chr.6: 99994030-99994030 on Build GRCh38
rs1004260936 Chr.6: 99994230-99994230 on Build GRCh38
rs958610014 Chr.6: 99994009-99994009 on Build GRCh38
rs148547635 Chr.6: 99994274-99994274 on Build GRCh38
rs200248313 Chr.6: 99993929-99993929 on Build GRCh38
rs968531996 Chr.6: 99994312-99994316 on Build GRCh38
rs201360449 Chr.6: 99994182-99994182 on Build GRCh38
rs200739667 Chr.6: 99994212-99994212 on Build GRCh38
rs1005149764 Chr.6: 99993876-99993876 on Build GRCh38
rs199560205 Chr.6: 99994034-99994034 on Build GRCh38
rs1019962252 Chr.6: 99993970-99993970 on Build GRCh38
rs556043373 Chr.6: 99993875-99993875 on Build GRCh38
rs201122564 Chr.6: 99994112-99994112 on Build GRCh38
rs568234085 Chr.6: 99993879-99993879 on Build GRCh38
rs200519724 Chr.6: 99994000-99994000 on Build GRCh38
rs2271116 Chr.6: 99993919-99993919 on Build GRCh38
rs199737403 Chr.6: 99993999-99993999 on Build GRCh38
rs112748388 Chr.6: 99994205-99994205 on Build GRCh38
rs562286486 Chr.6: 99994053-99994053 on Build GRCh38
rs201240762 Chr.6: 99993979-99993979 on Build GRCh38
rs199846924 Chr.6: 99994156-99994156 on Build GRCh38
rs893266319 Chr.6: 99994172-99994172 on Build GRCh38
rs940631746 Chr.6: 99994064-99994064 on Build GRCh38
rs996384728 Chr.6: 99993913-99993913 on Build GRCh38
rs765778272 Chr.6: 99993859-99993859 on Build GRCh38
rs6941007 Chr.6: 99994035-99994035 on Build GRCh38
rs776118730 Chr.6: 99993920-99993920 on Build GRCh38
rs1050634985 Chr.6: 99994157-99994157 on Build GRCh38

View Full Product Details