Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 113940433-113940933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC2
Entrez Gene ID
3066
Gene Name
histone deacetylase 2
Gene Aliases
HD2, RPD3, YAF1
UniGene
Hs.3352
Gene Chromosome Location
Chr.6: 113936156-113971195 on Build GRCh38
Transcripts
NM_001527.3 NR_033441.1 NR_073443.1 XM_011535788.1 XM_017010799.1

SNP Detail

SNP ID Chromosome Location
rs921092353 Chr.6: 113940801-113940801 on Build GRCh38
rs964987179 Chr.6: 113940554-113940554 on Build GRCh38
rs902848404 Chr.6: 113940646-113940646 on Build GRCh38
rs573333378 Chr.6: 113940581-113940581 on Build GRCh38
rs777941078 Chr.6: 113940696-113940696 on Build GRCh38
rs577117569 Chr.6: 113940543-113940543 on Build GRCh38
rs547359357 Chr.6: 113940835-113940835 on Build GRCh38
rs202124365 Chr.6: 113940640-113940640 on Build GRCh38
rs932250152 Chr.6: 113940576-113940576 on Build GRCh38
rs559711838 Chr.6: 113940788-113940790 on Build GRCh38
rs1046346782 Chr.6: 113940752-113940752 on Build GRCh38
rs923486425 Chr.6: 113940574-113940574 on Build GRCh38
rs352068 Chr.6: 113940688-113940688 on Build GRCh38
rs1027047457 Chr.6: 113940750-113940750 on Build GRCh38
rs144038528 Chr.6: 113940690-113940690 on Build GRCh38
rs1020597411 Chr.6: 113940551-113940551 on Build GRCh38
rs146500509 Chr.6: 113940827-113940827 on Build GRCh38
rs905402623 Chr.6: 113940725-113940725 on Build GRCh38
rs975745531 Chr.6: 113940800-113940801 on Build GRCh38
rs921103477 Chr.6: 113940631-113940631 on Build GRCh38
rs569950978 Chr.6: 113940500-113940500 on Build GRCh38
rs781552925 Chr.6: 113940516-113940516 on Build GRCh38
rs1002405304 Chr.6: 113940473-113940473 on Build GRCh38
rs115198269 Chr.6: 113940562-113940562 on Build GRCh38
rs965286032 Chr.6: 113940798-113940798 on Build GRCh38
rs144431877 Chr.6: 113940550-113940550 on Build GRCh38
rs997846447 Chr.6: 113940897-113940897 on Build GRCh38
rs949329873 Chr.6: 113940748-113940748 on Build GRCh38
rs1054054594 Chr.6: 113940780-113940780 on Build GRCh38
rs924386016 Chr.6: 113940869-113940869 on Build GRCh38
rs753035105 Chr.6: 113940531-113940531 on Build GRCh38
rs934338929 Chr.6: 113940663-113940663 on Build GRCh38
rs149673029 Chr.6: 113940475-113940475 on Build GRCh38
rs11391 Chr.6: 113940851-113940851 on Build GRCh38
rs905146368 Chr.6: 113940755-113940755 on Build GRCh38
rs909369367 Chr.6: 113940610-113940610 on Build GRCh38
rs576317609 Chr.6: 113940660-113940660 on Build GRCh38
rs1002116571 Chr.6: 113940763-113940763 on Build GRCh38
rs965018087 Chr.6: 113940894-113940894 on Build GRCh38
rs893618452 Chr.6: 113940791-113940791 on Build GRCh38
rs116225704 Chr.6: 113940529-113940529 on Build GRCh38
rs537114687 Chr.6: 113940523-113940523 on Build GRCh38
rs572303962 Chr.6: 113940567-113940567 on Build GRCh38
rs148438716 Chr.6: 113940642-113940642 on Build GRCh38

View Full Product Details