Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 122443256-122443755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERINC1
Entrez Gene ID
57515
Gene Name
serine incorporator 1
Gene Aliases
TDE1L, TDE2, TMS-2, TMS2
UniGene
Hs.146668
Gene Chromosome Location
Chr.6: 122443348-122471881 on Build GRCh38
Transcripts
NM_020755.3

SNP Detail

SNP ID Chromosome Location
rs913710700 Chr.6: 122443531-122443531 on Build GRCh38
rs552947243 Chr.6: 122443506-122443506 on Build GRCh38
rs981990988 Chr.6: 122443560-122443560 on Build GRCh38
rs551406832 Chr.6: 122443414-122443414 on Build GRCh38
rs922108074 Chr.6: 122443415-122443416 on Build GRCh38
rs180856260 Chr.6: 122443590-122443590 on Build GRCh38
rs887069372 Chr.6: 122443638-122443638 on Build GRCh38
rs1030629068 Chr.6: 122443312-122443312 on Build GRCh38
rs954323306 Chr.6: 122443397-122443397 on Build GRCh38
rs11559298 Chr.6: 122443583-122443583 on Build GRCh38
rs34359298 Chr.6: 122443435-122443436 on Build GRCh38
rs568940752 Chr.6: 122443329-122443329 on Build GRCh38
rs571472188 Chr.6: 122443430-122443430 on Build GRCh38
rs551946432 Chr.6: 122443492-122443492 on Build GRCh38
rs770233652 Chr.6: 122443353-122443353 on Build GRCh38
rs939194180 Chr.6: 122443619-122443619 on Build GRCh38
rs989195853 Chr.6: 122443539-122443539 on Build GRCh38
rs935563869 Chr.6: 122443497-122443497 on Build GRCh38
rs544746424 Chr.6: 122443711-122443711 on Build GRCh38
rs958045803 Chr.6: 122443482-122443482 on Build GRCh38
rs924245762 Chr.6: 122443494-122443494 on Build GRCh38
rs555407580 Chr.6: 122443604-122443604 on Build GRCh38
rs527363681 Chr.6: 122443631-122443631 on Build GRCh38
rs899124315 Chr.6: 122443700-122443700 on Build GRCh38
rs947786989 Chr.6: 122443591-122443591 on Build GRCh38
rs963530810 Chr.6: 122443390-122443390 on Build GRCh38
rs796935922 Chr.6: 122443302-122443302 on Build GRCh38
rs548997685 Chr.6: 122443705-122443705 on Build GRCh38
rs1044812371 Chr.6: 122443599-122443599 on Build GRCh38
rs989268142 Chr.6: 122443524-122443524 on Build GRCh38
rs987697010 Chr.6: 122443493-122443493 on Build GRCh38
rs966548974 Chr.6: 122443536-122443536 on Build GRCh38
rs763553188 Chr.6: 122443328-122443331 on Build GRCh38
rs186088087 Chr.6: 122443600-122443600 on Build GRCh38
rs1016276350 Chr.6: 122443661-122443661 on Build GRCh38
rs942099352 Chr.6: 122443723-122443723 on Build GRCh38
rs1005876669 Chr.6: 122443645-122443645 on Build GRCh38
rs35790440 Chr.6: 122443586-122443587 on Build GRCh38
rs531498271 Chr.6: 122443354-122443354 on Build GRCh38
rs566652375 Chr.6: 122443522-122443522 on Build GRCh38

View Full Product Details