Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 125751878-125752424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEY2
Entrez Gene ID
23493
Gene Name
hes related family bHLH transcription factor with YRPW motif 2
Gene Aliases
CHF1, GRIDLOCK, GRL, HERP1, HESR2, HRT2, bHLHb32
UniGene
Hs.144287
Gene Chromosome Location
Chr.6: 125747592-125762243 on Build GRCh38
Transcripts
NM_012259.2 XM_017010627.1 XM_017010628.1 XM_017010629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3828969 c.215G>A p.R72K Chr.6: 125752059-125752059 on Build GRCh38
6679600 c.188G>A p.R63Q Chr.6: 125752032-125752032 on Build GRCh38
3744909 c.182G>A p.R61Q Chr.6: 125752026-125752026 on Build GRCh38
1741411 c.246+6C>A p.? Chr.6: 125752096-125752096 on Build GRCh38
6029927 c.203T>C p.L68S Chr.6: 125752047-125752047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770161316 Chr.6: 125752120-125752120 on Build GRCh38
rs9398786 Chr.6: 125752361-125752361 on Build GRCh38
rs957255807 Chr.6: 125752309-125752309 on Build GRCh38
rs554381894 Chr.6: 125752103-125752103 on Build GRCh38
rs796768031 Chr.6: 125752311-125752311 on Build GRCh38
rs753434608 Chr.6: 125752101-125752102 on Build GRCh38
rs1010860335 Chr.6: 125752054-125752054 on Build GRCh38
rs757286685 Chr.6: 125752061-125752061 on Build GRCh38
rs755837991 Chr.6: 125751981-125751982 on Build GRCh38
rs781516744 Chr.6: 125751982-125751982 on Build GRCh38
rs868146450 Chr.6: 125752076-125752076 on Build GRCh38
rs533629589 Chr.6: 125752133-125752133 on Build GRCh38
rs764074174 Chr.6: 125752026-125752026 on Build GRCh38
rs556448688 Chr.6: 125752342-125752342 on Build GRCh38
rs983107246 Chr.6: 125752288-125752288 on Build GRCh38
rs928944617 Chr.6: 125752284-125752284 on Build GRCh38
rs1047446507 Chr.6: 125751937-125751937 on Build GRCh38
rs920864234 Chr.6: 125752257-125752257 on Build GRCh38
rs763035505 Chr.6: 125752004-125752004 on Build GRCh38
rs753077902 Chr.6: 125752028-125752028 on Build GRCh38
rs374782040 Chr.6: 125751919-125751919 on Build GRCh38
rs185432140 Chr.6: 125752214-125752214 on Build GRCh38
rs1023167322 Chr.6: 125752182-125752182 on Build GRCh38
rs960175717 Chr.6: 125752169-125752169 on Build GRCh38
rs189909373 Chr.6: 125752337-125752337 on Build GRCh38
rs763984618 Chr.6: 125752135-125752135 on Build GRCh38
rs542174585 Chr.6: 125752355-125752355 on Build GRCh38
rs766067177 Chr.6: 125752090-125752091 on Build GRCh38
rs778960437 Chr.6: 125752113-125752114 on Build GRCh38
rs760487306 Chr.6: 125752134-125752134 on Build GRCh38
rs201888758 Chr.6: 125752117-125752117 on Build GRCh38
rs113682501 Chr.6: 125752129-125752129 on Build GRCh38
rs970639929 Chr.6: 125752209-125752209 on Build GRCh38
rs181601319 Chr.6: 125752352-125752352 on Build GRCh38
rs752507444 Chr.6: 125752025-125752025 on Build GRCh38
rs774553825 Chr.6: 125752011-125752011 on Build GRCh38
rs372807209 Chr.6: 125751927-125751927 on Build GRCh38
rs141450342 Chr.6: 125752090-125752090 on Build GRCh38
rs993404112 Chr.6: 125752171-125752171 on Build GRCh38
rs771815981 Chr.6: 125751973-125751973 on Build GRCh38
rs911610505 Chr.6: 125752295-125752295 on Build GRCh38
rs776547365 Chr.6: 125752136-125752136 on Build GRCh38
rs772377585 Chr.6: 125752132-125752132 on Build GRCh38
rs945415950 Chr.6: 125751914-125751914 on Build GRCh38
rs78508125 Chr.6: 125752357-125752357 on Build GRCh38
rs759661730 Chr.6: 125752018-125752018 on Build GRCh38
rs112235907 Chr.6: 125752066-125752066 on Build GRCh38
rs1001491360 Chr.6: 125752179-125752179 on Build GRCh38
rs771523537 Chr.6: 125752128-125752128 on Build GRCh38
rs748007219 Chr.6: 125752139-125752139 on Build GRCh38
rs78771365 Chr.6: 125752113-125752113 on Build GRCh38
rs944524486 Chr.6: 125751993-125751993 on Build GRCh38
rs112677723 Chr.6: 125752098-125752098 on Build GRCh38
rs1025562153 Chr.6: 125752109-125752109 on Build GRCh38
rs905769933 Chr.6: 125752106-125752106 on Build GRCh38
rs531916278 Chr.6: 125752031-125752031 on Build GRCh38
rs745502281 Chr.6: 125751966-125751966 on Build GRCh38
rs778569906 Chr.6: 125751960-125751960 on Build GRCh38
rs946552776 Chr.6: 125752358-125752358 on Build GRCh38
rs373889651 Chr.6: 125751922-125751922 on Build GRCh38
rs761635639 Chr.6: 125752137-125752137 on Build GRCh38
rs773734737 Chr.6: 125751995-125751995 on Build GRCh38
rs1043638027 Chr.6: 125752359-125752359 on Build GRCh38
rs767207956 Chr.6: 125751913-125751913 on Build GRCh38
rs371231318 Chr.6: 125752114-125752125 on Build GRCh38
rs148304301 Chr.6: 125752079-125752079 on Build GRCh38
rs770916099 Chr.6: 125752006-125752006 on Build GRCh38
rs193035394 Chr.6: 125751994-125751994 on Build GRCh38
rs749750107 Chr.6: 125752125-125752125 on Build GRCh38
rs767898232 Chr.6: 125752021-125752021 on Build GRCh38
rs998839418 Chr.6: 125752366-125752366 on Build GRCh38
rs752594984 Chr.6: 125751918-125751918 on Build GRCh38
rs377597491 Chr.6: 125751925-125751925 on Build GRCh38
rs558751274 Chr.6: 125752139-125752139 on Build GRCh38
rs1031749606 Chr.6: 125752166-125752166 on Build GRCh38
rs370671117 Chr.6: 125751983-125751983 on Build GRCh38
rs750154365 Chr.6: 125751915-125751916 on Build GRCh38
rs778190656 Chr.6: 125752124-125752124 on Build GRCh38

View Full Product Details