Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155320707-155321209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RUSC1
Entrez Gene ID
23623
Gene Name
RUN and SH3 domain containing 1
Gene Aliases
NESCA
UniGene
Hs.226499
Gene Chromosome Location
Chr.1: 155320849-155331118 on Build GRCh38
Transcripts
NM_001105203.1 NM_001105204.1 XM_006711254.1 XM_006711256.1 NM_001039517.1

Gene Symbol
RUSC1-AS1
Entrez Gene ID
284618
Gene Name
RUSC1 antisense RNA 1
Gene Aliases
C1orf104
UniGene
Hs.650801
Gene Chromosome Location
Chr.1: 155320460-155324147 on Build GRCh38
Transcripts
NM_001105203.1 NM_001105204.1 XM_006711254.1 XM_006711256.1 NM_001039517.1

SNP Detail

SNP ID Chromosome Location
rs776338589 Chr.1: 155320735-155320735 on Build GRCh38
rs773191185 Chr.1: 155320743-155320743 on Build GRCh38
rs773419814 Chr.1: 155320941-155320941 on Build GRCh38
rs765618930 Chr.1: 155320939-155320940 on Build GRCh38
rs745696830 Chr.1: 155320927-155320927 on Build GRCh38
rs370918681 Chr.1: 155321000-155321000 on Build GRCh38
rs753247532 Chr.1: 155321053-155321053 on Build GRCh38
rs12407073 Chr.1: 155320781-155320781 on Build GRCh38
rs776030779 Chr.1: 155321136-155321136 on Build GRCh38
rs745963454 Chr.1: 155321170-155321170 on Build GRCh38
rs777551535 Chr.1: 155320915-155320915 on Build GRCh38
rs779955120 Chr.1: 155320996-155320996 on Build GRCh38
rs951216327 Chr.1: 155320900-155320920 on Build GRCh38
rs780554022 Chr.1: 155320795-155320795 on Build GRCh38
rs751318429 Chr.1: 155321150-155321150 on Build GRCh38
rs187876813 Chr.1: 155321117-155321117 on Build GRCh38
rs769524791 Chr.1: 155320817-155320817 on Build GRCh38
rs766720502 Chr.1: 155320889-155320889 on Build GRCh38
rs562568962 Chr.1: 155320920-155320920 on Build GRCh38
rs753312022 Chr.1: 155320956-155320956 on Build GRCh38
rs367859903 Chr.1: 155320861-155320861 on Build GRCh38
rs950214323 Chr.1: 155321022-155321022 on Build GRCh38
rs183376293 Chr.1: 155321090-155321090 on Build GRCh38
rs1023225289 Chr.1: 155320774-155320774 on Build GRCh38
rs780935853 Chr.1: 155321155-155321155 on Build GRCh38
rs753720409 Chr.1: 155320930-155320933 on Build GRCh38
rs769920357 Chr.1: 155320936-155320936 on Build GRCh38
rs1011898181 Chr.1: 155320962-155320962 on Build GRCh38
rs1030150032 Chr.1: 155320871-155320871 on Build GRCh38
rs554496067 Chr.1: 155320745-155320745 on Build GRCh38
rs769217918 Chr.1: 155321126-155321126 on Build GRCh38
rs750659410 Chr.1: 155320878-155320878 on Build GRCh38
rs765464736 Chr.1: 155320777-155320777 on Build GRCh38
rs776104992 Chr.1: 155321037-155321037 on Build GRCh38
rs961656333 Chr.1: 155320855-155320855 on Build GRCh38
rs766747488 Chr.1: 155320943-155320943 on Build GRCh38
rs374091551 Chr.1: 155320942-155320942 on Build GRCh38
rs1052882207 Chr.1: 155321004-155321004 on Build GRCh38
rs374302742 Chr.1: 155321018-155321018 on Build GRCh38
rs774639847 Chr.1: 155321023-155321023 on Build GRCh38
rs111493759 Chr.1: 155320805-155320805 on Build GRCh38
rs267598066 Chr.1: 155321076-155321076 on Build GRCh38
rs777156413 Chr.1: 155321128-155321128 on Build GRCh38
rs980835720 Chr.1: 155320818-155320818 on Build GRCh38
rs145119331 Chr.1: 155320770-155320770 on Build GRCh38
rs756810809 Chr.1: 155320974-155320974 on Build GRCh38
rs762068055 Chr.1: 155320775-155320775 on Build GRCh38
rs777602429 Chr.1: 155320825-155320825 on Build GRCh38
rs1048748737 Chr.1: 155320924-155320924 on Build GRCh38
rs957474365 Chr.1: 155320747-155320747 on Build GRCh38
rs749412035 Chr.1: 155321014-155321014 on Build GRCh38
rs939514707 Chr.1: 155320888-155320888 on Build GRCh38
rs761152095 Chr.1: 155321120-155321120 on Build GRCh38
rs774167260 Chr.1: 155320751-155320751 on Build GRCh38
rs972878619 Chr.1: 155320866-155320866 on Build GRCh38
rs780610953 Chr.1: 155320816-155320816 on Build GRCh38
rs755272023 Chr.1: 155320779-155320779 on Build GRCh38
rs775408370 Chr.1: 155320928-155320928 on Build GRCh38
rs749262734 Chr.1: 155321103-155321103 on Build GRCh38
rs753501973 Chr.1: 155320970-155320970 on Build GRCh38
rs568254069 Chr.1: 155320739-155320739 on Build GRCh38
rs759804232 Chr.1: 155321025-155321025 on Build GRCh38
rs760794361 Chr.1: 155320760-155320760 on Build GRCh38
rs758492513 Chr.1: 155321080-155321081 on Build GRCh38
rs758769020 Chr.1: 155320793-155320793 on Build GRCh38
rs370142160 Chr.1: 155320845-155320845 on Build GRCh38
rs542394224 Chr.1: 155321082-155321082 on Build GRCh38
rs771431669 Chr.1: 155321095-155321095 on Build GRCh38
rs774337682 Chr.1: 155320843-155320843 on Build GRCh38
rs764737054 Chr.1: 155321064-155321064 on Build GRCh38
rs778657731 Chr.1: 155320980-155320980 on Build GRCh38
rs750150497 Chr.1: 155321075-155321075 on Build GRCh38
rs763363706 Chr.1: 155320887-155320887 on Build GRCh38
rs752743632 Chr.1: 155321156-155321156 on Build GRCh38
rs778398403 Chr.1: 155320973-155320973 on Build GRCh38
rs529844870 Chr.1: 155321052-155321052 on Build GRCh38
rs1006918598 Chr.1: 155321104-155321104 on Build GRCh38
rs779218513 Chr.1: 155320841-155320841 on Build GRCh38
rs758992173 Chr.1: 155321144-155321144 on Build GRCh38
rs752217114 Chr.1: 155320905-155320905 on Build GRCh38
rs778622328 Chr.1: 155320921-155320921 on Build GRCh38
rs779095141 Chr.1: 155321168-155321168 on Build GRCh38
rs763769671 Chr.1: 155320803-155320803 on Build GRCh38
rs761910815 Chr.1: 155320868-155320868 on Build GRCh38
rs772538731 Chr.1: 155321114-155321114 on Build GRCh38
rs368269316 Chr.1: 155321045-155321045 on Build GRCh38
rs755696915 Chr.1: 155320914-155320914 on Build GRCh38
rs982630902 Chr.1: 155321082-155321083 on Build GRCh38
rs771059509 Chr.1: 155321109-155321109 on Build GRCh38
rs759431114 Chr.1: 155320844-155320844 on Build GRCh38
rs897186124 Chr.1: 155321085-155321085 on Build GRCh38
rs756795455 Chr.1: 155321057-155321057 on Build GRCh38
rs762110324 Chr.1: 155320935-155320935 on Build GRCh38
rs779865313 Chr.1: 155321083-155321083 on Build GRCh38
rs376418913 Chr.1: 155320950-155320950 on Build GRCh38
rs754775212 Chr.1: 155321151-155321151 on Build GRCh38
rs771022265 Chr.1: 155321019-155321019 on Build GRCh38
rs934312868 Chr.1: 155320979-155320979 on Build GRCh38
rs750012837 Chr.1: 155321142-155321142 on Build GRCh38
rs758205324 Chr.1: 155320992-155320992 on Build GRCh38
rs752079175 Chr.1: 155320893-155320893 on Build GRCh38
rs768057653 Chr.1: 155321032-155321032 on Build GRCh38
rs772378995 Chr.1: 155321174-155321174 on Build GRCh38
rs754024477 Chr.1: 155320771-155320771 on Build GRCh38
rs745581563 Chr.1: 155320982-155320982 on Build GRCh38
rs776642315 Chr.1: 155320859-155320859 on Build GRCh38
rs773333967 Chr.1: 155321009-155321009 on Build GRCh38
rs768072117 Chr.1: 155320953-155320953 on Build GRCh38
rs780202699 Chr.1: 155321141-155321175 on Build GRCh38
rs778996504 Chr.1: 155321112-155321112 on Build GRCh38
rs201236896 Chr.1: 155320894-155320894 on Build GRCh38
rs750745157 Chr.1: 155320783-155320783 on Build GRCh38
rs747017052 Chr.1: 155320997-155320997 on Build GRCh38
rs750675441 Chr.1: 155321072-155321076 on Build GRCh38
rs757250361 Chr.1: 155321162-155321162 on Build GRCh38
rs754865207 Chr.1: 155321093-155321093 on Build GRCh38
rs16836822 Chr.1: 155320796-155320796 on Build GRCh38
rs58624579 Chr.1: 155320740-155320740 on Build GRCh38
rs370805843 Chr.1: 155321175-155321175 on Build GRCh38
rs749030193 Chr.1: 155320831-155320831 on Build GRCh38
rs1053278750 Chr.1: 155320762-155320762 on Build GRCh38
rs772556955 Chr.1: 155320951-155320951 on Build GRCh38
rs544266286 Chr.1: 155320900-155320900 on Build GRCh38
rs895528084 Chr.1: 155321012-155321012 on Build GRCh38
rs748170311 Chr.1: 155320930-155320930 on Build GRCh38
rs747317253 Chr.1: 155321187-155321187 on Build GRCh38

View Full Product Details