Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155659326-155659854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YY1AP1
Entrez Gene ID
55249
Gene Name
YY1 associated protein 1
Gene Aliases
HCCA1, HCCA2, YAP, YY1AP
UniGene
Hs.584927
Gene Chromosome Location
Chr.1: 155659442-155689032 on Build GRCh38
Transcripts
NM_001198899.1 NM_001198900.1 NM_001198901.1 NM_001198902.1 NM_001198903.1 NM_001198904.1 NM_001198905.1 NM_001198906.1 NM_018253.3 NM_139118.2 NM_139119.2 NM_139121.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1270655 c.2266C>A p.Q756K Chr.1: 155659782-155659782 on Build GRCh38
6860592 c.2625G>A p.M875I Chr.1: 155659699-155659699 on Build GRCh38
5789323 c.2254G>C p.E752Q Chr.1: 155659794-155659794 on Build GRCh38
107657 c.2370G>A p.E790E Chr.1: 155659678-155659678 on Build GRCh38
340147 c.2365A>G p.T789A Chr.1: 155659683-155659683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772566717 Chr.1: 155659783-155659783 on Build GRCh38
rs865780038 Chr.1: 155659471-155659471 on Build GRCh38
rs918399127 Chr.1: 155659591-155659591 on Build GRCh38
rs773437691 Chr.1: 155659641-155659642 on Build GRCh38
rs768518995 Chr.1: 155659476-155659476 on Build GRCh38
rs892603033 Chr.1: 155659656-155659656 on Build GRCh38
rs1031260482 Chr.1: 155659651-155659651 on Build GRCh38
rs181513775 Chr.1: 155659478-155659478 on Build GRCh38
rs969369018 Chr.1: 155659485-155659485 on Build GRCh38
rs773059490 Chr.1: 155659442-155659442 on Build GRCh38
rs766452850 Chr.1: 155659739-155659739 on Build GRCh38
rs774215231 Chr.1: 155659790-155659790 on Build GRCh38
rs763116149 Chr.1: 155659659-155659662 on Build GRCh38
rs144591490 Chr.1: 155659786-155659786 on Build GRCh38
rs903960830 Chr.1: 155659640-155659640 on Build GRCh38
rs768940668 Chr.1: 155659747-155659747 on Build GRCh38
rs771844192 Chr.1: 155659371-155659371 on Build GRCh38
rs772307540 Chr.1: 155659638-155659638 on Build GRCh38
rs753215911 Chr.1: 155659793-155659795 on Build GRCh38
rs1025149759 Chr.1: 155659711-155659711 on Build GRCh38
rs762655678 Chr.1: 155659682-155659682 on Build GRCh38
rs749104341 Chr.1: 155659373-155659373 on Build GRCh38
rs148849021 Chr.1: 155659412-155659412 on Build GRCh38
rs199876978 Chr.1: 155659823-155659823 on Build GRCh38
rs974028880 Chr.1: 155659572-155659572 on Build GRCh38
rs770985845 Chr.1: 155659616-155659616 on Build GRCh38
rs755968877 Chr.1: 155659802-155659802 on Build GRCh38
rs779056591 Chr.1: 155659619-155659619 on Build GRCh38
rs759270293 Chr.1: 155659795-155659795 on Build GRCh38
rs751432312 Chr.1: 155659607-155659607 on Build GRCh38
rs758593196 Chr.1: 155659735-155659735 on Build GRCh38
rs770364572 Chr.1: 155659779-155659779 on Build GRCh38
rs764393982 Chr.1: 155659830-155659831 on Build GRCh38
rs767944417 Chr.1: 155659792-155659793 on Build GRCh38
rs1139870 Chr.1: 155659504-155659504 on Build GRCh38
rs918513816 Chr.1: 155659736-155659736 on Build GRCh38
rs558393470 Chr.1: 155659672-155659672 on Build GRCh38
rs945898661 Chr.1: 155659631-155659631 on Build GRCh38
rs1038718487 Chr.1: 155659590-155659590 on Build GRCh38
rs201532179 Chr.1: 155659615-155659615 on Build GRCh38
rs959678430 Chr.1: 155659436-155659436 on Build GRCh38
rs775796839 Chr.1: 155659639-155659639 on Build GRCh38
rs376798637 Chr.1: 155659807-155659807 on Build GRCh38
rs774471583 Chr.1: 155659742-155659743 on Build GRCh38
rs941301977 Chr.1: 155659571-155659571 on Build GRCh38
rs986637548 Chr.1: 155659798-155659798 on Build GRCh38
rs369033668 Chr.1: 155659831-155659831 on Build GRCh38
rs950849363 Chr.1: 155659472-155659472 on Build GRCh38
rs1051657149 Chr.1: 155659614-155659614 on Build GRCh38
rs974359926 Chr.1: 155659733-155659733 on Build GRCh38
rs376638223 Chr.1: 155659825-155659825 on Build GRCh38
rs146321739 Chr.1: 155659678-155659678 on Build GRCh38
rs138944459 Chr.1: 155659703-155659703 on Build GRCh38
rs761048449 Chr.1: 155659814-155659815 on Build GRCh38
rs761072735 Chr.1: 155659646-155659646 on Build GRCh38
rs766174671 Chr.1: 155659687-155659687 on Build GRCh38
rs1034078618 Chr.1: 155659435-155659435 on Build GRCh38
rs752876559 Chr.1: 155659814-155659814 on Build GRCh38
rs200664513 Chr.1: 155659726-155659726 on Build GRCh38
rs368438278 Chr.1: 155659742-155659742 on Build GRCh38
rs200816738 Chr.1: 155659709-155659709 on Build GRCh38
rs201827011 Chr.1: 155659809-155659809 on Build GRCh38
rs747366618 Chr.1: 155659744-155659744 on Build GRCh38
rs781329819 Chr.1: 155659609-155659609 on Build GRCh38
rs7539 Chr.1: 155659692-155659692 on Build GRCh38
rs368946647 Chr.1: 155659817-155659817 on Build GRCh38
rs1033700305 Chr.1: 155659666-155659666 on Build GRCh38
rs769136644 Chr.1: 155659647-155659647 on Build GRCh38
rs992344354 Chr.1: 155659462-155659462 on Build GRCh38
rs1018522369 Chr.1: 155659365-155659365 on Build GRCh38
rs866594441 Chr.1: 155659743-155659743 on Build GRCh38
rs185713547 Chr.1: 155659789-155659789 on Build GRCh38
rs755249698 Chr.1: 155659818-155659818 on Build GRCh38
rs752512078 Chr.1: 155659710-155659710 on Build GRCh38
rs890910947 Chr.1: 155659598-155659598 on Build GRCh38
rs1050769749 Chr.1: 155659596-155659596 on Build GRCh38
rs749970690 Chr.1: 155659674-155659674 on Build GRCh38
rs1031616627 Chr.1: 155659440-155659440 on Build GRCh38
rs529638610 Chr.1: 155659434-155659434 on Build GRCh38
rs116024191 Chr.1: 155659595-155659595 on Build GRCh38
rs372287277 Chr.1: 155659769-155659769 on Build GRCh38
rs1001023899 Chr.1: 155659433-155659433 on Build GRCh38
rs891605363 Chr.1: 155659348-155659348 on Build GRCh38
rs767162593 Chr.1: 155659797-155659797 on Build GRCh38
rs1025071692 Chr.1: 155659483-155659483 on Build GRCh38
rs918106345 Chr.1: 155659468-155659469 on Build GRCh38
rs942379058 Chr.1: 155659599-155659599 on Build GRCh38
rs777065440 Chr.1: 155659768-155659768 on Build GRCh38
rs760032262 Chr.1: 155659759-155659760 on Build GRCh38
rs752563562 Chr.1: 155659800-155659800 on Build GRCh38
rs375067522 Chr.1: 155659637-155659637 on Build GRCh38
rs556811436 Chr.1: 155659731-155659731 on Build GRCh38
rs143518160 Chr.1: 155659660-155659660 on Build GRCh38
rs780109936 Chr.1: 155659816-155659816 on Build GRCh38
rs980781936 Chr.1: 155659724-155659724 on Build GRCh38
rs927213545 Chr.1: 155659565-155659565 on Build GRCh38
rs371628973 Chr.1: 155659608-155659608 on Build GRCh38
rs778812731 Chr.1: 155659727-155659727 on Build GRCh38
rs775229522 Chr.1: 155659372-155659372 on Build GRCh38
rs981218796 Chr.1: 155659539-155659539 on Build GRCh38
rs550837957 Chr.1: 155659364-155659364 on Build GRCh38
rs762385069 Chr.1: 155659670-155659670 on Build GRCh38

View Full Product Details