Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155756568-155757086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GON4L
Entrez Gene ID
54856
Gene Name
gon-4 like
Gene Aliases
GON-4, GON4, YARP
UniGene
Hs.656361
Gene Chromosome Location
Chr.1: 155749658-155859400 on Build GRCh38
Transcripts
NM_001282856.1 NM_001282858.1 NM_001282860.1 XM_005245284.3 XM_005245286.3 XM_006711393.3 XM_006711394.3 XM_011509658.2 XM_011509659.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
896989 c.5496C>T p.I1832I Chr.1: 155756979-155756979 on Build GRCh38
4023090 c.5442G>A p.E1814E Chr.1: 155757033-155757033 on Build GRCh38
4946722 c.5424G>A p.L1808L Chr.1: 155757051-155757051 on Build GRCh38
4023091 c.5442G>A p.E1814E Chr.1: 155757033-155757033 on Build GRCh38
3360376 c.5418G>T p.V1806V Chr.1: 155757057-155757057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962055561 Chr.1: 155756935-155756935 on Build GRCh38
rs756015284 Chr.1: 155757014-155757014 on Build GRCh38
rs1054152328 Chr.1: 155756600-155756601 on Build GRCh38
rs1014894992 Chr.1: 155756706-155756706 on Build GRCh38
rs558284726 Chr.1: 155756833-155756833 on Build GRCh38
rs753156261 Chr.1: 155756979-155756979 on Build GRCh38
rs775520278 Chr.1: 155756916-155756916 on Build GRCh38
rs372043132 Chr.1: 155756975-155756975 on Build GRCh38
rs915619053 Chr.1: 155756879-155756879 on Build GRCh38
rs745620709 Chr.1: 155756911-155756911 on Build GRCh38
rs933854853 Chr.1: 155756751-155756751 on Build GRCh38
rs71591910 Chr.1: 155756877-155756878 on Build GRCh38
rs371209533 Chr.1: 155756762-155756762 on Build GRCh38
rs576652930 Chr.1: 155756844-155756844 on Build GRCh38
rs896722424 Chr.1: 155756843-155756843 on Build GRCh38
rs146685010 Chr.1: 155756691-155756691 on Build GRCh38
rs529877070 Chr.1: 155756942-155756942 on Build GRCh38
rs376547681 Chr.1: 155756668-155756668 on Build GRCh38
rs972665581 Chr.1: 155756712-155756712 on Build GRCh38
rs768046906 Chr.1: 155756982-155756982 on Build GRCh38
rs777350947 Chr.1: 155757053-155757053 on Build GRCh38
rs902136999 Chr.1: 155756813-155756813 on Build GRCh38
rs1012408825 Chr.1: 155756671-155756671 on Build GRCh38
rs1036815874 Chr.1: 155756715-155756715 on Build GRCh38
rs751989071 Chr.1: 155756998-155756999 on Build GRCh38
rs182274287 Chr.1: 155756890-155756890 on Build GRCh38
rs753140395 Chr.1: 155756994-155756994 on Build GRCh38
rs750913731 Chr.1: 155756943-155756946 on Build GRCh38
rs946457703 Chr.1: 155756794-155756794 on Build GRCh38
rs572362289 Chr.1: 155756725-155756725 on Build GRCh38
rs957133860 Chr.1: 155756848-155756848 on Build GRCh38
rs766464010 Chr.1: 155756772-155756772 on Build GRCh38
rs966693775 Chr.1: 155756826-155756826 on Build GRCh38
rs907072527 Chr.1: 155756697-155756697 on Build GRCh38
rs369973352 Chr.1: 155756966-155756966 on Build GRCh38
rs35875716 Chr.1: 155756950-155756951 on Build GRCh38
rs758882866 Chr.1: 155756990-155756991 on Build GRCh38
rs897341223 Chr.1: 155756699-155756699 on Build GRCh38
rs772186271 Chr.1: 155756915-155756915 on Build GRCh38
rs1043792790 Chr.1: 155756797-155756797 on Build GRCh38
rs1049152933 Chr.1: 155757032-155757032 on Build GRCh38
rs977621708 Chr.1: 155756837-155756837 on Build GRCh38
rs765266945 Chr.1: 155756939-155756942 on Build GRCh38
rs997457495 Chr.1: 155756950-155756950 on Build GRCh38
rs897941622 Chr.1: 155756735-155756735 on Build GRCh38
rs774270351 Chr.1: 155756910-155756910 on Build GRCh38
rs763292913 Chr.1: 155756968-155756968 on Build GRCh38
rs1056764035 Chr.1: 155756842-155756842 on Build GRCh38
rs1007597918 Chr.1: 155757045-155757045 on Build GRCh38
rs756975667 Chr.1: 155757051-155757051 on Build GRCh38
rs1015422209 Chr.1: 155756913-155756913 on Build GRCh38
rs953418932 Chr.1: 155756791-155756791 on Build GRCh38
rs541780927 Chr.1: 155756978-155756978 on Build GRCh38
rs755342308 Chr.1: 155756999-155757000 on Build GRCh38
rs922513743 Chr.1: 155756723-155756723 on Build GRCh38
rs948367647 Chr.1: 155756650-155756650 on Build GRCh38
rs1036848481 Chr.1: 155756976-155756976 on Build GRCh38
rs887869459 Chr.1: 155756883-155756883 on Build GRCh38
rs574964145 Chr.1: 155756649-155756649 on Build GRCh38
rs755225719 Chr.1: 155756896-155756896 on Build GRCh38
rs369298519 Chr.1: 155757033-155757033 on Build GRCh38
rs952140097 Chr.1: 155756695-155756695 on Build GRCh38
rs910979200 Chr.1: 155756786-155756786 on Build GRCh38
rs543924094 Chr.1: 155756859-155756859 on Build GRCh38
rs762014494 Chr.1: 155756946-155756946 on Build GRCh38
rs985209644 Chr.1: 155756696-155756696 on Build GRCh38
rs961594643 Chr.1: 155756709-155756709 on Build GRCh38
rs777940388 Chr.1: 155756928-155756929 on Build GRCh38
rs535802007 Chr.1: 155756690-155756690 on Build GRCh38
rs373074777 Chr.1: 155756606-155756606 on Build GRCh38
rs765341511 Chr.1: 155756957-155756957 on Build GRCh38
rs766717492 Chr.1: 155756991-155756992 on Build GRCh38
rs376378515 Chr.1: 155757010-155757010 on Build GRCh38
rs143188838 Chr.1: 155756608-155756608 on Build GRCh38
rs1012439766 Chr.1: 155756851-155756851 on Build GRCh38
rs1045666122 Chr.1: 155756693-155756693 on Build GRCh38
rs934980722 Chr.1: 155756819-155756819 on Build GRCh38

View Full Product Details