Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 134170941-134171447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGK1
Entrez Gene ID
6446
Gene Name
serum/glucocorticoid regulated kinase 1
Gene Aliases
SGK
UniGene
Hs.510078
Gene Chromosome Location
Chr.6: 134169246-134318112 on Build GRCh38
Transcripts
NM_001143676.1 NM_001143677.1 NM_001143678.1 NM_001291995.1 NM_005627.3 XM_011536071.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2155662 c.948C>T p.H316H Chr.6: 134171083-134171083 on Build GRCh38
6502927 c.1087G>C p.D363H Chr.6: 134171058-134171058 on Build GRCh38
3858437 c.1069A>G p.K357E Chr.6: 134171034-134171034 on Build GRCh38
3858439 c.997A>G p.K333E Chr.6: 134171034-134171034 on Build GRCh38
5014642 c.1120T>A p.F374I Chr.6: 134171025-134171025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776533497 Chr.6: 134171191-134171191 on Build GRCh38
rs60975684 Chr.6: 134171247-134171247 on Build GRCh38
rs867991403 Chr.6: 134170995-134170995 on Build GRCh38
rs368663146 Chr.6: 134171015-134171015 on Build GRCh38
rs1015298233 Chr.6: 134171280-134171280 on Build GRCh38
rs55932330 Chr.6: 134171036-134171036 on Build GRCh38
rs759833790 Chr.6: 134170998-134171000 on Build GRCh38
rs568024103 Chr.6: 134171225-134171225 on Build GRCh38
rs902460344 Chr.6: 134171245-134171245 on Build GRCh38
rs140184576 Chr.6: 134171092-134171092 on Build GRCh38
rs867978372 Chr.6: 134171324-134171324 on Build GRCh38
rs772324787 Chr.6: 134171091-134171091 on Build GRCh38
rs770097947 Chr.6: 134171007-134171007 on Build GRCh38
rs150335204 Chr.6: 134171098-134171098 on Build GRCh38
rs377197471 Chr.6: 134171403-134171403 on Build GRCh38
rs761768112 Chr.6: 134171131-134171131 on Build GRCh38
rs201165496 Chr.6: 134170983-134170983 on Build GRCh38
rs781575804 Chr.6: 134170973-134170973 on Build GRCh38
rs754957913 Chr.6: 134171382-134171382 on Build GRCh38
rs746968315 Chr.6: 134171259-134171259 on Build GRCh38
rs9493846 Chr.6: 134171358-134171358 on Build GRCh38
rs945373246 Chr.6: 134171093-134171093 on Build GRCh38
rs538127002 Chr.6: 134171143-134171143 on Build GRCh38
rs552502455 Chr.6: 134171072-134171072 on Build GRCh38
rs746430738 Chr.6: 134171360-134171361 on Build GRCh38
rs758343377 Chr.6: 134171162-134171162 on Build GRCh38
rs770988201 Chr.6: 134170987-134170987 on Build GRCh38
rs369841718 Chr.6: 134171243-134171248 on Build GRCh38
rs556332407 Chr.6: 134171354-134171354 on Build GRCh38
rs57787286 Chr.6: 134171255-134171255 on Build GRCh38
rs778918804 Chr.6: 134171154-134171154 on Build GRCh38
rs959527267 Chr.6: 134171348-134171365 on Build GRCh38
rs560627470 Chr.6: 134170988-134170988 on Build GRCh38
rs1008714863 Chr.6: 134171253-134171253 on Build GRCh38
rs1042795773 Chr.6: 134171233-134171233 on Build GRCh38
rs927417922 Chr.6: 134171371-134171371 on Build GRCh38
rs1046343840 Chr.6: 134171057-134171057 on Build GRCh38
rs745955657 Chr.6: 134171156-134171156 on Build GRCh38
rs1027732767 Chr.6: 134171300-134171300 on Build GRCh38
rs975200099 Chr.6: 134171269-134171269 on Build GRCh38
rs978042636 Chr.6: 134171383-134171383 on Build GRCh38
rs780434638 Chr.6: 134171163-134171163 on Build GRCh38
rs764689970 Chr.6: 134171132-134171132 on Build GRCh38
rs1010616694 Chr.6: 134171177-134171177 on Build GRCh38
rs869212004 Chr.6: 134171359-134171360 on Build GRCh38
rs113901059 Chr.6: 134171405-134171405 on Build GRCh38
rs778115907 Chr.6: 134170984-134170984 on Build GRCh38
rs761427803 Chr.6: 134171125-134171126 on Build GRCh38
rs372296178 Chr.6: 134170993-134170993 on Build GRCh38
rs747605647 Chr.6: 134170982-134170982 on Build GRCh38
rs749748729 Chr.6: 134171045-134171045 on Build GRCh38
rs767063374 Chr.6: 134171131-134171137 on Build GRCh38
rs369131581 Chr.6: 134171256-134171256 on Build GRCh38
rs539693807 Chr.6: 134171348-134171348 on Build GRCh38
rs748689783 Chr.6: 134170974-134170974 on Build GRCh38
rs770948952 Chr.6: 134171046-134171046 on Build GRCh38
rs908114220 Chr.6: 134171347-134171347 on Build GRCh38
rs766028121 Chr.6: 134171135-134171135 on Build GRCh38
rs138245764 Chr.6: 134171125-134171125 on Build GRCh38
rs764121588 Chr.6: 134170992-134170992 on Build GRCh38
rs576451068 Chr.6: 134171251-134171255 on Build GRCh38
rs918147584 Chr.6: 134171353-134171353 on Build GRCh38
rs999954731 Chr.6: 134170972-134170972 on Build GRCh38
rs143238056 Chr.6: 134170986-134170990 on Build GRCh38
rs777126637 Chr.6: 134170992-134170993 on Build GRCh38
rs768512667 Chr.6: 134171103-134171103 on Build GRCh38
rs756138821 Chr.6: 134171359-134171360 on Build GRCh38
rs778072365 Chr.6: 134171140-134171140 on Build GRCh38
rs995295582 Chr.6: 134171347-134171348 on Build GRCh38
rs375575573 Chr.6: 134170996-134170996 on Build GRCh38
rs190276895 Chr.6: 134171176-134171176 on Build GRCh38
rs569220878 Chr.6: 134171142-134171142 on Build GRCh38
rs140051664 Chr.6: 134171104-134171105 on Build GRCh38
rs756495408 Chr.6: 134171137-134171137 on Build GRCh38
rs113274174 Chr.6: 134171358-134171359 on Build GRCh38
rs751156921 Chr.6: 134171136-134171136 on Build GRCh38
rs748081018 Chr.6: 134171194-134171194 on Build GRCh38
rs778816776 Chr.6: 134171102-134171102 on Build GRCh38
rs374331599 Chr.6: 134171255-134171255 on Build GRCh38
rs60070447 Chr.6: 134171258-134171259 on Build GRCh38
rs996812808 Chr.6: 134170985-134170985 on Build GRCh38
rs769753666 Chr.6: 134171228-134171228 on Build GRCh38
rs762583569 Chr.6: 134171133-134171133 on Build GRCh38
rs962436698 Chr.6: 134171331-134171331 on Build GRCh38
rs1054879972 Chr.6: 134171379-134171379 on Build GRCh38
rs865981643 Chr.6: 134171101-134171102 on Build GRCh38
rs9493847 Chr.6: 134171360-134171360 on Build GRCh38
rs781472175 Chr.6: 134171014-134171014 on Build GRCh38
rs1020875822 Chr.6: 134171222-134171222 on Build GRCh38
rs376904641 Chr.6: 134171083-134171083 on Build GRCh38
rs775403686 Chr.6: 134170989-134170989 on Build GRCh38
rs937527269 Chr.6: 134171373-134171373 on Build GRCh38
rs770276025 Chr.6: 134170978-134170978 on Build GRCh38
rs758033352 Chr.6: 134171002-134171002 on Build GRCh38
rs78176488 Chr.6: 134171128-134171128 on Build GRCh38
rs146066740 Chr.6: 134171095-134171095 on Build GRCh38
rs770218929 Chr.6: 134171372-134171373 on Build GRCh38
rs57304640 Chr.6: 134171243-134171243 on Build GRCh38
rs748478722 Chr.6: 134171041-134171041 on Build GRCh38
rs760752811 Chr.6: 134170991-134170991 on Build GRCh38
rs983442545 Chr.6: 134171344-134171344 on Build GRCh38
rs925323869 Chr.6: 134171388-134171388 on Build GRCh38
rs774028416 Chr.6: 134171012-134171016 on Build GRCh38
rs142616527 Chr.6: 134171253-134171254 on Build GRCh38
rs371687628 Chr.6: 134171017-134171017 on Build GRCh38

View Full Product Details