Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136845423-136845920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX7
Entrez Gene ID
5191
Gene Name
peroxisomal biogenesis factor 7
Gene Aliases
PBD9B, PTS2R, RCDP1, RD
UniGene
Hs.280932
Gene Chromosome Location
Chr.6: 136822220-136913934 on Build GRCh38
Transcripts
NM_000288.3 XM_005267019.4 XM_006715502.1 XM_017010934.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1073765 c.378G>A p.Q126Q Chr.6: 136845653-136845653 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781067011 Chr.6: 136845721-136845721 on Build GRCh38
rs758421190 Chr.6: 136845587-136845587 on Build GRCh38
rs1043802368 Chr.6: 136845757-136845757 on Build GRCh38
rs746310218 Chr.6: 136845660-136845660 on Build GRCh38
rs1029742964 Chr.6: 136845601-136845601 on Build GRCh38
rs757320874 Chr.6: 136845575-136845575 on Build GRCh38
rs962705488 Chr.6: 136845887-136845887 on Build GRCh38
rs528317074 Chr.6: 136845607-136845607 on Build GRCh38
rs776675774 Chr.6: 136845683-136845683 on Build GRCh38
rs373278677 Chr.6: 136845795-136845795 on Build GRCh38
rs891992004 Chr.6: 136845794-136845794 on Build GRCh38
rs775519611 Chr.6: 136845857-136845857 on Build GRCh38
rs762113236 Chr.6: 136845688-136845688 on Build GRCh38
rs531174213 Chr.6: 136845541-136845541 on Build GRCh38
rs752887223 Chr.6: 136845565-136845565 on Build GRCh38
rs751370922 Chr.6: 136845716-136845716 on Build GRCh38
rs561801070 Chr.6: 136845814-136845814 on Build GRCh38
rs538621257 Chr.6: 136845824-136845824 on Build GRCh38
rs1011861471 Chr.6: 136845489-136845489 on Build GRCh38
rs544037188 Chr.6: 136845889-136845889 on Build GRCh38
rs7449595 Chr.6: 136845591-136845591 on Build GRCh38
rs965319414 Chr.6: 136845798-136845798 on Build GRCh38
rs1053370246 Chr.6: 136845779-136845779 on Build GRCh38
rs958184054 Chr.6: 136845584-136845584 on Build GRCh38
rs374892341 Chr.6: 136845578-136845578 on Build GRCh38
rs373876195 Chr.6: 136845733-136845733 on Build GRCh38
rs766476163 Chr.6: 136845701-136845701 on Build GRCh38
rs571896936 Chr.6: 136845534-136845534 on Build GRCh38
rs750200076 Chr.6: 136845740-136845740 on Build GRCh38
rs753932031 Chr.6: 136845564-136845564 on Build GRCh38
rs765168761 Chr.6: 136845692-136845692 on Build GRCh38
rs954126784 Chr.6: 136845823-136845823 on Build GRCh38
rs747418919 Chr.6: 136845847-136845847 on Build GRCh38
rs114626592 Chr.6: 136845839-136845839 on Build GRCh38
rs575174066 Chr.6: 136845855-136845855 on Build GRCh38
rs780751870 Chr.6: 136845614-136845614 on Build GRCh38
rs377394660 Chr.6: 136845778-136845778 on Build GRCh38
rs2295592 Chr.6: 136845512-136845512 on Build GRCh38
rs377509685 Chr.6: 136845735-136845735 on Build GRCh38
rs886829129 Chr.6: 136845788-136845788 on Build GRCh38
rs775817219 Chr.6: 136845665-136845665 on Build GRCh38
rs1056103459 Chr.6: 136845453-136845453 on Build GRCh38
rs567401960 Chr.6: 136845561-136845561 on Build GRCh38
rs927181 Chr.6: 136845544-136845544 on Build GRCh38
rs780790798 Chr.6: 136845452-136845452 on Build GRCh38
rs777262353 Chr.6: 136845741-136845741 on Build GRCh38
rs1023203401 Chr.6: 136845510-136845510 on Build GRCh38
rs912120745 Chr.6: 136845740-136845740 on Build GRCh38
rs780920376 Chr.6: 136845728-136845728 on Build GRCh38
rs1029774168 Chr.6: 136845813-136845813 on Build GRCh38
rs372212162 Chr.6: 136845468-136845468 on Build GRCh38
rs113268723 Chr.6: 136845652-136845652 on Build GRCh38
rs769137963 Chr.6: 136845648-136845648 on Build GRCh38
rs1032264188 Chr.6: 136845545-136845545 on Build GRCh38
rs746861382 Chr.6: 136845602-136845602 on Build GRCh38
rs982579812 Chr.6: 136845487-136845487 on Build GRCh38
rs913951873 Chr.6: 136845621-136845621 on Build GRCh38
rs758402722 Chr.6: 136845698-136845698 on Build GRCh38
rs61753242 Chr.6: 136845670-136845670 on Build GRCh38
rs373287524 Chr.6: 136845720-136845720 on Build GRCh38
rs750532429 Chr.6: 136845695-136845695 on Build GRCh38
rs956192334 Chr.6: 136845559-136845559 on Build GRCh38
rs761074779 Chr.6: 136845668-136845668 on Build GRCh38
rs1028201184 Chr.6: 136845831-136845831 on Build GRCh38
rs772635570 Chr.6: 136845662-136845662 on Build GRCh38
rs1046525626 Chr.6: 136845657-136845657 on Build GRCh38
rs267608255 Chr.6: 136845605-136845605 on Build GRCh38
rs750333842 Chr.6: 136845582-136845582 on Build GRCh38
rs559459893 Chr.6: 136845507-136845507 on Build GRCh38
rs779925128 Chr.6: 136845596-136845596 on Build GRCh38
rs62653606 Chr.6: 136845645-136845671 on Build GRCh38
rs551746653 Chr.6: 136845521-136845521 on Build GRCh38
rs121909154 Chr.6: 136845620-136845620 on Build GRCh38
rs769266228 Chr.6: 136845862-136845862 on Build GRCh38
rs764346452 Chr.6: 136845675-136845675 on Build GRCh38
rs758654715 Chr.6: 136845644-136845670 on Build GRCh38
rs745922509 Chr.6: 136845743-136845743 on Build GRCh38
rs903484455 Chr.6: 136845761-136845761 on Build GRCh38

View Full Product Details