Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138215919-138216437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFGEF3
Entrez Gene ID
57221
Gene Name
ARFGEF family member 3
Gene Aliases
A7322, BIG3, C6orf92, KIAA1244, PPP1R33, dJ171N11.1
UniGene
Hs.194408
Gene Chromosome Location
Chr.6: 138161690-138344663 on Build GRCh38
Transcripts
NM_020340.4 NM_021635.2

Gene Symbol
PBOV1
Entrez Gene ID
59351
Gene Name
prostate and breast cancer overexpressed 1
Gene Aliases
UC28, UROC28, dJ171N11.2
UniGene
Hs.302016
Gene Chromosome Location
Chr.6: 138215986-138218490 on Build GRCh38
Transcripts
NM_020340.4 NM_021635.2

SNP Detail

SNP ID Chromosome Location
rs540755847 Chr.6: 138215986-138215986 on Build GRCh38
rs935078528 Chr.6: 138216040-138216040 on Build GRCh38
rs556993751 Chr.6: 138216359-138216359 on Build GRCh38
rs979670318 Chr.6: 138216213-138216213 on Build GRCh38
rs1056957611 Chr.6: 138216164-138216164 on Build GRCh38
rs903484288 Chr.6: 138216031-138216031 on Build GRCh38
rs138652386 Chr.6: 138216072-138216072 on Build GRCh38
rs569759050 Chr.6: 138216151-138216151 on Build GRCh38
rs561863100 Chr.6: 138216015-138216015 on Build GRCh38
rs11967414 Chr.6: 138215983-138215983 on Build GRCh38
rs9402959 Chr.6: 138216340-138216340 on Build GRCh38
rs773318356 Chr.6: 138216284-138216285 on Build GRCh38
rs367947315 Chr.6: 138216223-138216223 on Build GRCh38
rs910009711 Chr.6: 138216337-138216337 on Build GRCh38
rs751363258 Chr.6: 138216071-138216071 on Build GRCh38
rs527913817 Chr.6: 138216091-138216091 on Build GRCh38
rs577837215 Chr.6: 138216011-138216011 on Build GRCh38
rs963133386 Chr.6: 138215955-138215955 on Build GRCh38
rs1025753306 Chr.6: 138216161-138216161 on Build GRCh38
rs965628168 Chr.6: 138216253-138216253 on Build GRCh38
rs915714333 Chr.6: 138216401-138216401 on Build GRCh38
rs1023457151 Chr.6: 138216086-138216086 on Build GRCh38
rs769227947 Chr.6: 138216127-138216127 on Build GRCh38
rs972790330 Chr.6: 138216358-138216358 on Build GRCh38
rs948570000 Chr.6: 138216413-138216413 on Build GRCh38
rs1057074395 Chr.6: 138216391-138216391 on Build GRCh38
rs562126386 Chr.6: 138216090-138216090 on Build GRCh38
rs185103880 Chr.6: 138216088-138216088 on Build GRCh38
rs535375128 Chr.6: 138216165-138216165 on Build GRCh38
rs1021393279 Chr.6: 138216208-138216208 on Build GRCh38
rs188103150 Chr.6: 138216138-138216138 on Build GRCh38
rs911361901 Chr.6: 138216200-138216200 on Build GRCh38
rs985059578 Chr.6: 138216003-138216003 on Build GRCh38
rs949095433 Chr.6: 138216313-138216313 on Build GRCh38
rs768930653 Chr.6: 138215981-138215983 on Build GRCh38
rs878997497 Chr.6: 138216304-138216304 on Build GRCh38
rs1013454354 Chr.6: 138216078-138216078 on Build GRCh38
rs201119348 Chr.6: 138216207-138216207 on Build GRCh38
rs11967442 Chr.6: 138216124-138216124 on Build GRCh38
rs565718683 Chr.6: 138216273-138216273 on Build GRCh38
rs938557306 Chr.6: 138216386-138216386 on Build GRCh38
rs911498067 Chr.6: 138215996-138215996 on Build GRCh38
rs972801774 Chr.6: 138215981-138215981 on Build GRCh38
rs148955106 Chr.6: 138215969-138215971 on Build GRCh38
rs555266477 Chr.6: 138215953-138215953 on Build GRCh38
rs533257315 Chr.6: 138216134-138216134 on Build GRCh38
rs769433972 Chr.6: 138216215-138216215 on Build GRCh38
rs199524611 Chr.6: 138215979-138215980 on Build GRCh38
rs201009832 Chr.6: 138215980-138215980 on Build GRCh38
rs573939781 Chr.6: 138215996-138215996 on Build GRCh38
rs932712680 Chr.6: 138216108-138216108 on Build GRCh38
rs375664430 Chr.6: 138216089-138216089 on Build GRCh38
rs57864443 Chr.6: 138216056-138216056 on Build GRCh38
rs771065862 Chr.6: 138216055-138216055 on Build GRCh38
rs374640173 Chr.6: 138215980-138215982 on Build GRCh38
rs9402958 Chr.6: 138216295-138216295 on Build GRCh38
rs550806084 Chr.6: 138215994-138215995 on Build GRCh38
rs942931647 Chr.6: 138216002-138216002 on Build GRCh38
rs779787471 Chr.6: 138216068-138216068 on Build GRCh38
rs374477877 Chr.6: 138216285-138216285 on Build GRCh38
rs577773607 Chr.6: 138216296-138216296 on Build GRCh38
rs11967441 Chr.6: 138216102-138216102 on Build GRCh38
rs867024811 Chr.6: 138216266-138216266 on Build GRCh38
rs904184050 Chr.6: 138216209-138216209 on Build GRCh38
rs986948337 Chr.6: 138216199-138216199 on Build GRCh38
rs938502616 Chr.6: 138216152-138216152 on Build GRCh38
rs374978243 Chr.6: 138216218-138216218 on Build GRCh38
rs1036687717 Chr.6: 138216135-138216135 on Build GRCh38
rs200688424 Chr.6: 138215982-138215983 on Build GRCh38
rs1052518664 Chr.6: 138216287-138216287 on Build GRCh38
rs750900708 Chr.6: 138215945-138215952 on Build GRCh38
rs1051189474 Chr.6: 138215990-138215990 on Build GRCh38
rs534792405 Chr.6: 138216274-138216274 on Build GRCh38
rs994201859 Chr.6: 138216404-138216404 on Build GRCh38

View Full Product Details