Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 142166040-142166547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VTA1
Entrez Gene ID
51534
Gene Name
vesicle trafficking 1
Gene Aliases
C6orf55, DRG-1, DRG1, HSPC228, LIP5, My012, SBP1
UniGene
Hs.431367
Gene Chromosome Location
Chr.6: 142147162-142220948 on Build GRCh38
Transcripts
NM_001286371.1 NM_001286372.1 NM_016485.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
150187 c.207+10C>A p.? Chr.6: 142166332-142166332 on Build GRCh38
7259291 c.129G>A p.M43I Chr.6: 142166244-142166244 on Build GRCh38
5869433 c.170G>A p.R57H Chr.6: 142166285-142166285 on Build GRCh38
740796 c.185A>G p.K62R Chr.6: 142166300-142166300 on Build GRCh38
4526796 c.141G>A p.M47I Chr.6: 142166256-142166256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762018505 Chr.6: 142166370-142166370 on Build GRCh38
rs776263301 Chr.6: 142166227-142166227 on Build GRCh38
rs767801276 Chr.6: 142166239-142166239 on Build GRCh38
rs779608507 Chr.6: 142166197-142166197 on Build GRCh38
rs1046252803 Chr.6: 142166171-142166171 on Build GRCh38
rs765290723 Chr.6: 142166267-142166267 on Build GRCh38
rs760198268 Chr.6: 142166211-142166214 on Build GRCh38
rs141564366 Chr.6: 142166276-142166276 on Build GRCh38
rs543577980 Chr.6: 142166384-142166384 on Build GRCh38
rs938726143 Chr.6: 142166496-142166496 on Build GRCh38
rs928710984 Chr.6: 142166172-142166172 on Build GRCh38
rs750309454 Chr.6: 142166147-142166147 on Build GRCh38
rs746610256 Chr.6: 142166203-142166203 on Build GRCh38
rs754490110 Chr.6: 142166189-142166189 on Build GRCh38
rs891804826 Chr.6: 142166210-142166210 on Build GRCh38
rs113182478 Chr.6: 142166182-142166182 on Build GRCh38
rs983972692 Chr.6: 142166401-142166401 on Build GRCh38
rs373907353 Chr.6: 142166300-142166300 on Build GRCh38
rs146846977 Chr.6: 142166515-142166518 on Build GRCh38
rs775987102 Chr.6: 142166343-142166343 on Build GRCh38
rs771469609 Chr.6: 142166231-142166231 on Build GRCh38
rs997574540 Chr.6: 142166411-142166411 on Build GRCh38
rs927830257 Chr.6: 142166488-142166488 on Build GRCh38
rs764422499 Chr.6: 142166366-142166366 on Build GRCh38
rs977148980 Chr.6: 142166250-142166250 on Build GRCh38
rs79577504 Chr.6: 142166385-142166385 on Build GRCh38
rs754727814 Chr.6: 142166289-142166289 on Build GRCh38
rs952398339 Chr.6: 142166392-142166392 on Build GRCh38
rs542577369 Chr.6: 142166285-142166285 on Build GRCh38
rs761226791 Chr.6: 142166230-142166230 on Build GRCh38
rs773907499 Chr.6: 142166104-142166104 on Build GRCh38
rs1053495260 Chr.6: 142166207-142166207 on Build GRCh38
rs969317821 Chr.6: 142166480-142166480 on Build GRCh38
rs947878858 Chr.6: 142166144-142166144 on Build GRCh38
rs191549526 Chr.6: 142166113-142166113 on Build GRCh38
rs559393068 Chr.6: 142166494-142166494 on Build GRCh38
rs561581686 Chr.6: 142166320-142166320 on Build GRCh38
rs774843539 Chr.6: 142166334-142166334 on Build GRCh38
rs746316520 Chr.6: 142166337-142166337 on Build GRCh38
rs555110000 Chr.6: 142166510-142166510 on Build GRCh38
rs750350264 Chr.6: 142166275-142166275 on Build GRCh38
rs768064815 Chr.6: 142166213-142166213 on Build GRCh38
rs554812192 Chr.6: 142166297-142166297 on Build GRCh38
rs774788611 Chr.6: 142166233-142166233 on Build GRCh38
rs982348832 Chr.6: 142166486-142166486 on Build GRCh38
rs745709412 Chr.6: 142166193-142166193 on Build GRCh38
rs761011756 Chr.6: 142166244-142166244 on Build GRCh38
rs225655 Chr.6: 142166126-142166126 on Build GRCh38
rs225656 Chr.6: 142166332-142166332 on Build GRCh38
rs574447625 Chr.6: 142166260-142166260 on Build GRCh38
rs772550011 Chr.6: 142166340-142166340 on Build GRCh38
rs756247482 Chr.6: 142166444-142166444 on Build GRCh38
rs779841639 Chr.6: 142166284-142166284 on Build GRCh38
rs568714142 Chr.6: 142166263-142166263 on Build GRCh38
rs755040442 Chr.6: 142166468-142166468 on Build GRCh38
rs771947281 Chr.6: 142166196-142166196 on Build GRCh38
rs776671306 Chr.6: 142166367-142166367 on Build GRCh38
rs780680109 Chr.6: 142166310-142166310 on Build GRCh38
rs757393774 Chr.6: 142166264-142166264 on Build GRCh38
rs796983025 Chr.6: 142166511-142166514 on Build GRCh38
rs541086140 Chr.6: 142166431-142166431 on Build GRCh38
rs760913838 Chr.6: 142166356-142166356 on Build GRCh38
rs1051870141 Chr.6: 142166445-142166445 on Build GRCh38
rs897950980 Chr.6: 142166067-142166067 on Build GRCh38
rs759923917 Chr.6: 142166238-142166238 on Build GRCh38
rs752875745 Chr.6: 142166242-142166242 on Build GRCh38
rs758277438 Chr.6: 142166280-142166280 on Build GRCh38
rs753755409 Chr.6: 142166376-142166376 on Build GRCh38
rs879182887 Chr.6: 142166364-142166364 on Build GRCh38
rs13213706 Chr.6: 142166283-142166283 on Build GRCh38
rs997605470 Chr.6: 142166070-142166070 on Build GRCh38
rs577003174 Chr.6: 142166404-142166404 on Build GRCh38

View Full Product Details