Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 149378328-149378837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAB2
Entrez Gene ID
23118
Gene Name
TGF-beta activated kinase 1/MAP3K7 binding protein 2
Gene Aliases
CHTD2, MAP3K7IP2, TAB-2
UniGene
Hs.269775
Gene Chromosome Location
Chr.6: 149217924-149411613 on Build GRCh38
Transcripts
NM_001292034.2 NM_001292035.2 NM_015093.5 XM_011535633.2 XM_017010591.1 XM_017010592.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4852455 c.518G>C p.R173T Chr.6: 149378433-149378433 on Build GRCh38
1728595 c.779C>T p.P260L Chr.6: 149378694-149378694 on Build GRCh38
1672824 c.745C>T p.Q249* Chr.6: 149378660-149378660 on Build GRCh38
1074392 c.562G>T p.G188C Chr.6: 149378477-149378477 on Build GRCh38
5061477 c.811C>T p.Q271* Chr.6: 149378726-149378726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528190406 Chr.6: 149378815-149378815 on Build GRCh38
rs774890725 Chr.6: 149378543-149378543 on Build GRCh38
rs146755707 Chr.6: 149378591-149378591 on Build GRCh38
rs267607100 Chr.6: 149378603-149378603 on Build GRCh38
rs150852637 Chr.6: 149378353-149378353 on Build GRCh38
rs953649650 Chr.6: 149378792-149378792 on Build GRCh38
rs763333319 Chr.6: 149378528-149378528 on Build GRCh38
rs772663796 Chr.6: 149378783-149378783 on Build GRCh38
rs369027697 Chr.6: 149378481-149378481 on Build GRCh38
rs962520265 Chr.6: 149378422-149378422 on Build GRCh38
rs998144680 Chr.6: 149378610-149378610 on Build GRCh38
rs772221255 Chr.6: 149378508-149378508 on Build GRCh38
rs149874061 Chr.6: 149378768-149378768 on Build GRCh38
rs753582412 Chr.6: 149378479-149378479 on Build GRCh38
rs541985795 Chr.6: 149378742-149378742 on Build GRCh38
rs369055635 Chr.6: 149378766-149378766 on Build GRCh38
rs267607101 Chr.6: 149378537-149378537 on Build GRCh38
rs764892237 Chr.6: 149378602-149378602 on Build GRCh38
rs573087020 Chr.6: 149378713-149378713 on Build GRCh38
rs752518088 Chr.6: 149378492-149378492 on Build GRCh38
rs758306321 Chr.6: 149378605-149378605 on Build GRCh38
rs766969196 Chr.6: 149378421-149378421 on Build GRCh38
rs764100688 Chr.6: 149378620-149378620 on Build GRCh38
rs751616426 Chr.6: 149378628-149378628 on Build GRCh38
rs770367619 Chr.6: 149378680-149378680 on Build GRCh38
rs1025859824 Chr.6: 149378804-149378804 on Build GRCh38
rs769996008 Chr.6: 149378514-149378514 on Build GRCh38
rs760222192 Chr.6: 149378425-149378425 on Build GRCh38
rs144874468 Chr.6: 149378773-149378773 on Build GRCh38
rs769063306 Chr.6: 149378674-149378674 on Build GRCh38
rs914680833 Chr.6: 149378535-149378535 on Build GRCh38
rs774833204 Chr.6: 149378675-149378675 on Build GRCh38
rs914695532 Chr.6: 149378489-149378489 on Build GRCh38
rs777958155 Chr.6: 149378504-149378504 on Build GRCh38
rs1002679542 Chr.6: 149378529-149378529 on Build GRCh38
rs765990978 Chr.6: 149378470-149378470 on Build GRCh38
rs973007353 Chr.6: 149378448-149378448 on Build GRCh38
rs140436062 Chr.6: 149378708-149378708 on Build GRCh38
rs781346597 Chr.6: 149378634-149378634 on Build GRCh38
rs372346677 Chr.6: 149378365-149378365 on Build GRCh38
rs749645606 Chr.6: 149378667-149378667 on Build GRCh38
rs780313015 Chr.6: 149378743-149378743 on Build GRCh38
rs927250962 Chr.6: 149378482-149378482 on Build GRCh38
rs773484017 Chr.6: 149378540-149378540 on Build GRCh38
rs749575189 Chr.6: 149378761-149378761 on Build GRCh38
rs777032310 Chr.6: 149378374-149378374 on Build GRCh38
rs748656776 Chr.6: 149378679-149378679 on Build GRCh38
rs756394050 Chr.6: 149378641-149378641 on Build GRCh38
rs748601861 Chr.6: 149378781-149378781 on Build GRCh38
rs746032729 Chr.6: 149378638-149378638 on Build GRCh38
rs922195786 Chr.6: 149378407-149378407 on Build GRCh38
rs757284171 Chr.6: 149378722-149378722 on Build GRCh38
rs971262944 Chr.6: 149378812-149378812 on Build GRCh38
rs754720551 Chr.6: 149378480-149378480 on Build GRCh38
rs79807560 Chr.6: 149378686-149378686 on Build GRCh38
rs559449485 Chr.6: 149378696-149378696 on Build GRCh38
rs775795498 Chr.6: 149378388-149378388 on Build GRCh38
rs181739891 Chr.6: 149378498-149378498 on Build GRCh38
rs377150853 Chr.6: 149378376-149378376 on Build GRCh38
rs750487045 Chr.6: 149378728-149378728 on Build GRCh38
rs990325896 Chr.6: 149378811-149378811 on Build GRCh38
rs775948663 Chr.6: 149378515-149378515 on Build GRCh38
rs775053250 Chr.6: 149378694-149378694 on Build GRCh38
rs757340629 Chr.6: 149378632-149378632 on Build GRCh38
rs767459040 Chr.6: 149378724-149378724 on Build GRCh38
rs760161262 Chr.6: 149378556-149378556 on Build GRCh38
rs115168915 Chr.6: 149378512-149378512 on Build GRCh38
rs758431534 Chr.6: 149378503-149378503 on Build GRCh38
rs765938014 Chr.6: 149378576-149378576 on Build GRCh38
rs781562724 Chr.6: 149378511-149378511 on Build GRCh38
rs368534604 Chr.6: 149378689-149378689 on Build GRCh38
rs139897844 Chr.6: 149378387-149378387 on Build GRCh38
rs372085948 Chr.6: 149378586-149378586 on Build GRCh38
rs776226341 Chr.6: 149378681-149378681 on Build GRCh38
rs747053355 Chr.6: 149378506-149378506 on Build GRCh38
rs749969260 Chr.6: 149378424-149378424 on Build GRCh38
rs780182449 Chr.6: 149378644-149378644 on Build GRCh38
rs745360232 Chr.6: 149378802-149378802 on Build GRCh38
rs1016733839 Chr.6: 149378413-149378413 on Build GRCh38
rs763533213 Chr.6: 149378400-149378400 on Build GRCh38
rs113049462 Chr.6: 149378789-149378789 on Build GRCh38

View Full Product Details