Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 149863241-149863733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRP11
Entrez Gene ID
84918
Gene Name
LDL receptor related protein 11
Gene Aliases
MANSC3, bA350J20.3
UniGene
Hs.408355
Gene Chromosome Location
Chr.6: 149818758-149864344 on Build GRCh38
Transcripts
NM_032832.5 XM_011536194.2 XM_017011380.1 NR_045126.1

Gene Symbol
RAET1E-AS1
Entrez Gene ID
100652739
Gene Name
RAET1E antisense RNA 1
Gene Aliases
RAET1G-AS1
UniGene
Hs.642734
Gene Chromosome Location
Chr.6: 149863498-149919508 on Build GRCh38
Transcripts
NM_032832.5 XM_011536194.2 XM_017011380.1 NR_045126.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4657398 c.435G>A p.V145V Chr.6: 149863586-149863586 on Build GRCh38
4657397 c.435G>A p.V145V Chr.6: 149863586-149863586 on Build GRCh38
4787971 c.564C>A p.R188R Chr.6: 149863457-149863457 on Build GRCh38
4790193 c.503G>T p.R168L Chr.6: 149863518-149863518 on Build GRCh38
4801494 c.498G>T p.T166T Chr.6: 149863523-149863523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746555321 Chr.6: 149863597-149863611 on Build GRCh38
rs781004944 Chr.6: 149863484-149863484 on Build GRCh38
rs1040856050 Chr.6: 149863384-149863384 on Build GRCh38
rs571328083 Chr.6: 149863569-149863569 on Build GRCh38
rs897123820 Chr.6: 149863659-149863659 on Build GRCh38
rs754973701 Chr.6: 149863651-149863651 on Build GRCh38
rs867763037 Chr.6: 149863507-149863507 on Build GRCh38
rs754346516 Chr.6: 149863421-149863421 on Build GRCh38
rs937021151 Chr.6: 149863591-149863591 on Build GRCh38
rs374706154 Chr.6: 149863367-149863367 on Build GRCh38
rs781094949 Chr.6: 149863370-149863370 on Build GRCh38
rs762505450 Chr.6: 149863434-149863434 on Build GRCh38
rs868592263 Chr.6: 149863546-149863546 on Build GRCh38
rs913808325 Chr.6: 149863342-149863342 on Build GRCh38
rs988335823 Chr.6: 149863493-149863493 on Build GRCh38
rs761082756 Chr.6: 149863414-149863414 on Build GRCh38
rs1045367068 Chr.6: 149863355-149863355 on Build GRCh38
rs945682168 Chr.6: 149863343-149863343 on Build GRCh38
rs749355223 Chr.6: 149863399-149863399 on Build GRCh38
rs570558607 Chr.6: 149863329-149863329 on Build GRCh38
rs772283204 Chr.6: 149863412-149863412 on Build GRCh38
rs913611938 Chr.6: 149863557-149863557 on Build GRCh38
rs756823292 Chr.6: 149863659-149863675 on Build GRCh38
rs767921707 Chr.6: 149863444-149863444 on Build GRCh38
rs936312015 Chr.6: 149863299-149863299 on Build GRCh38
rs750230007 Chr.6: 149863576-149863576 on Build GRCh38
rs1032446416 Chr.6: 149863483-149863483 on Build GRCh38
rs187967019 Chr.6: 149863586-149863586 on Build GRCh38
rs546611868 Chr.6: 149863475-149863475 on Build GRCh38
rs761980271 Chr.6: 149863411-149863411 on Build GRCh38
rs1015635206 Chr.6: 149863430-149863430 on Build GRCh38
rs779922098 Chr.6: 149863397-149863397 on Build GRCh38
rs769796237 Chr.6: 149863393-149863393 on Build GRCh38
rs749128783 Chr.6: 149863600-149863600 on Build GRCh38
rs768665678 Chr.6: 149863400-149863400 on Build GRCh38
rs941464878 Chr.6: 149863655-149863655 on Build GRCh38
rs748237888 Chr.6: 149863691-149863691 on Build GRCh38
rs778788219 Chr.6: 149863686-149863686 on Build GRCh38
rs1053408684 Chr.6: 149863305-149863305 on Build GRCh38
rs888518957 Chr.6: 149863636-149863636 on Build GRCh38
rs926348942 Chr.6: 149863541-149863541 on Build GRCh38
rs751112522 Chr.6: 149863450-149863450 on Build GRCh38
rs866428918 Chr.6: 149863518-149863518 on Build GRCh38
rs932457376 Chr.6: 149863588-149863588 on Build GRCh38
rs957812597 Chr.6: 149863548-149863551 on Build GRCh38
rs995367852 Chr.6: 149863420-149863420 on Build GRCh38
rs982351879 Chr.6: 149863585-149863585 on Build GRCh38
rs975876598 Chr.6: 149863482-149863482 on Build GRCh38
rs949634308 Chr.6: 149863563-149863563 on Build GRCh38
rs774509201 Chr.6: 149863401-149863401 on Build GRCh38
rs777127823 Chr.6: 149863549-149863577 on Build GRCh38
rs914107506 Chr.6: 149863560-149863560 on Build GRCh38
rs780017760 Chr.6: 149863592-149863592 on Build GRCh38
rs745664321 Chr.6: 149863385-149863385 on Build GRCh38
rs926407357 Chr.6: 149863284-149863284 on Build GRCh38
rs931105590 Chr.6: 149863701-149863701 on Build GRCh38
rs143869691 Chr.6: 149863465-149863465 on Build GRCh38
rs936869839 Chr.6: 149863272-149863272 on Build GRCh38
rs766870969 Chr.6: 149863416-149863416 on Build GRCh38
rs906916481 Chr.6: 149863366-149863366 on Build GRCh38
rs989098347 Chr.6: 149863550-149863550 on Build GRCh38
rs898247037 Chr.6: 149863666-149863666 on Build GRCh38
rs773503734 Chr.6: 149863413-149863413 on Build GRCh38
rs895214132 Chr.6: 149863400-149863410 on Build GRCh38
rs1053091789 Chr.6: 149863280-149863280 on Build GRCh38
rs866664792 Chr.6: 149863529-149863529 on Build GRCh38
rs756994423 Chr.6: 149863369-149863369 on Build GRCh38
rs1025941691 Chr.6: 149863432-149863432 on Build GRCh38
rs762474773 Chr.6: 149863278-149863278 on Build GRCh38

View Full Product Details