Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 158661184-158661625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYTL3
Entrez Gene ID
94120
Gene Name
synaptotagmin like 3
Gene Aliases
SLP3
UniGene
Hs.436977
Gene Chromosome Location
Chr.6: 158644881-158766298 on Build GRCh38
Transcripts
NM_001009991.3 NM_001242384.1 NM_001242394.1 NM_001242395.1 XM_005267215.4 XM_005267218.4 XM_006715605.3 XM_006715606.3 XM_006715611.3 XM_011536254.2 XM_017011495.1 XM_017011496.1 XM_017011497.1 XM_017011498.1 XM_017011499.1

SNP Detail

SNP ID Chromosome Location
rs975434203 Chr.6: 158661527-158661527 on Build GRCh38
rs118082531 Chr.6: 158661551-158661551 on Build GRCh38
rs896595132 Chr.6: 158661531-158661531 on Build GRCh38
rs905076362 Chr.6: 158661570-158661570 on Build GRCh38
rs944555717 Chr.6: 158661353-158661353 on Build GRCh38
rs1017147590 Chr.6: 158661497-158661497 on Build GRCh38
rs148978250 Chr.6: 158661300-158661300 on Build GRCh38
rs148220872 Chr.6: 158661409-158661409 on Build GRCh38
rs554633370 Chr.6: 158661599-158661599 on Build GRCh38
rs988251684 Chr.6: 158661557-158661557 on Build GRCh38
rs905125176 Chr.6: 158661347-158661347 on Build GRCh38
rs538942521 Chr.6: 158661508-158661508 on Build GRCh38
rs896178979 Chr.6: 158661601-158661601 on Build GRCh38
rs937959702 Chr.6: 158661577-158661577 on Build GRCh38
rs563448774 Chr.6: 158661280-158661280 on Build GRCh38
rs74691371 Chr.6: 158661249-158661249 on Build GRCh38
rs913998588 Chr.6: 158661560-158661560 on Build GRCh38
rs145837886 Chr.6: 158661240-158661240 on Build GRCh38
rs76948983 Chr.6: 158661540-158661540 on Build GRCh38
rs578105150 Chr.6: 158661248-158661248 on Build GRCh38
rs925649709 Chr.6: 158661302-158661302 on Build GRCh38
rs1001121830 Chr.6: 158661588-158661588 on Build GRCh38
rs911700545 Chr.6: 158661345-158661345 on Build GRCh38
rs919155027 Chr.6: 158661385-158661385 on Build GRCh38
rs375441691 Chr.6: 158661276-158661276 on Build GRCh38
rs113292554 Chr.6: 158661543-158661543 on Build GRCh38
rs528431543 Chr.6: 158661374-158661374 on Build GRCh38
rs951521359 Chr.6: 158661473-158661473 on Build GRCh38
rs2306749 Chr.6: 158661241-158661241 on Build GRCh38
rs186195177 Chr.6: 158661484-158661484 on Build GRCh38
rs189446135 Chr.6: 158661565-158661565 on Build GRCh38
rs143742164 Chr.6: 158661388-158661388 on Build GRCh38
rs1035105530 Chr.6: 158661386-158661386 on Build GRCh38
rs964654967 Chr.6: 158661524-158661524 on Build GRCh38
rs1043619855 Chr.6: 158661341-158661341 on Build GRCh38
rs1043950935 Chr.6: 158661567-158661567 on Build GRCh38
rs1025755786 Chr.6: 158661411-158661411 on Build GRCh38
rs905202946 Chr.6: 158661418-158661418 on Build GRCh38
rs1040114872 Chr.6: 158661227-158661227 on Build GRCh38
rs868696309 Chr.6: 158661550-158661550 on Build GRCh38
rs1021608358 Chr.6: 158661253-158661253 on Build GRCh38
rs569725700 Chr.6: 158661355-158661355 on Build GRCh38
rs368459439 Chr.6: 158661228-158661228 on Build GRCh38
rs560921295 Chr.6: 158661368-158661368 on Build GRCh38
rs542643738 Chr.6: 158661322-158661322 on Build GRCh38

View Full Product Details