Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 160122138-160122630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A1
Entrez Gene ID
6580
Gene Name
solute carrier family 22 member 1
Gene Aliases
HOCT1, OCT1, oct1_cds
UniGene
Hs.117367
Gene Chromosome Location
Chr.6: 160121789-160159201 on Build GRCh38
Transcripts
NM_003057.2 NM_153187.1 XM_005267102.4 XM_005267103.1 XM_005267105.4 XM_006715552.1 XM_011536074.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3023615 c.235C>G p.P79A Chr.6: 160122170-160122170 on Build GRCh38
4726652 c.390C>T p.P130P Chr.6: 160122325-160122325 on Build GRCh38
1075627 c.269G>A p.R90H Chr.6: 160122204-160122204 on Build GRCh38
4726651 c.263G>A p.C88Y Chr.6: 160122198-160122198 on Build GRCh38
6437839 c.378G>T p.V126V Chr.6: 160122313-160122313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044519923 Chr.6: 160122346-160122346 on Build GRCh38
rs913262231 Chr.6: 160122457-160122457 on Build GRCh38
rs559672598 Chr.6: 160122325-160122325 on Build GRCh38
rs992478353 Chr.6: 160122571-160122571 on Build GRCh38
rs371964408 Chr.6: 160122201-160122201 on Build GRCh38
rs75309601 Chr.6: 160122601-160122601 on Build GRCh38
rs145740120 Chr.6: 160122174-160122174 on Build GRCh38
rs965158163 Chr.6: 160122235-160122235 on Build GRCh38
rs55918055 Chr.6: 160122197-160122197 on Build GRCh38
rs368225370 Chr.6: 160122178-160122178 on Build GRCh38
rs925740888 Chr.6: 160122594-160122594 on Build GRCh38
rs138558984 Chr.6: 160122480-160122480 on Build GRCh38
rs767174948 Chr.6: 160122261-160122261 on Build GRCh38
rs141775735 Chr.6: 160122228-160122228 on Build GRCh38
rs111863691 Chr.6: 160122296-160122297 on Build GRCh38
rs563134940 Chr.6: 160122247-160122247 on Build GRCh38
rs913323076 Chr.6: 160122206-160122206 on Build GRCh38
rs750183710 Chr.6: 160122326-160122326 on Build GRCh38
rs769151008 Chr.6: 160122207-160122207 on Build GRCh38
rs758799074 Chr.6: 160122363-160122363 on Build GRCh38
rs138109329 Chr.6: 160122280-160122280 on Build GRCh38
rs762147370 Chr.6: 160122300-160122301 on Build GRCh38
rs766605382 Chr.6: 160122330-160122330 on Build GRCh38
rs185697157 Chr.6: 160122311-160122311 on Build GRCh38
rs900552380 Chr.6: 160122470-160122470 on Build GRCh38
rs73598470 Chr.6: 160122463-160122463 on Build GRCh38
rs772259161 Chr.6: 160122224-160122224 on Build GRCh38
rs560632505 Chr.6: 160122286-160122286 on Build GRCh38
rs769565823 Chr.6: 160122309-160122309 on Build GRCh38
rs994580790 Chr.6: 160122537-160122537 on Build GRCh38
rs200716785 Chr.6: 160122203-160122203 on Build GRCh38
rs756936346 Chr.6: 160122385-160122385 on Build GRCh38
rs912148254 Chr.6: 160122504-160122504 on Build GRCh38
rs201620700 Chr.6: 160122229-160122229 on Build GRCh38
rs753283796 Chr.6: 160122335-160122335 on Build GRCh38
rs1034911558 Chr.6: 160122413-160122413 on Build GRCh38
rs201731508 Chr.6: 160122279-160122279 on Build GRCh38
rs200865946 Chr.6: 160122298-160122298 on Build GRCh38
rs149299457 Chr.6: 160122499-160122499 on Build GRCh38
rs750709201 Chr.6: 160122264-160122264 on Build GRCh38
rs111781079 Chr.6: 160122367-160122367 on Build GRCh38
rs556555770 Chr.6: 160122586-160122586 on Build GRCh38
rs143215289 Chr.6: 160122172-160122172 on Build GRCh38
rs144580927 Chr.6: 160122502-160122502 on Build GRCh38
rs199642943 Chr.6: 160122175-160122175 on Build GRCh38
rs760315741 Chr.6: 160122173-160122173 on Build GRCh38
rs35546288 Chr.6: 160122188-160122188 on Build GRCh38
rs747364135 Chr.6: 160122371-160122371 on Build GRCh38
rs200684404 Chr.6: 160122285-160122285 on Build GRCh38
rs112536204 Chr.6: 160122548-160122548 on Build GRCh38
rs532477464 Chr.6: 160122338-160122338 on Build GRCh38
rs916921193 Chr.6: 160122576-160122576 on Build GRCh38
rs374886881 Chr.6: 160122226-160122226 on Build GRCh38
rs781487444 Chr.6: 160122200-160122200 on Build GRCh38
rs758386386 Chr.6: 160122185-160122185 on Build GRCh38
rs374066285 Chr.6: 160122366-160122366 on Build GRCh38
rs569062438 Chr.6: 160122448-160122448 on Build GRCh38
rs748560150 Chr.6: 160122222-160122222 on Build GRCh38
rs748905423 Chr.6: 160122398-160122398 on Build GRCh38
rs778210538 Chr.6: 160122543-160122543 on Build GRCh38
rs960278363 Chr.6: 160122440-160122440 on Build GRCh38
rs752668758 Chr.6: 160122383-160122384 on Build GRCh38
rs765176277 Chr.6: 160122342-160122342 on Build GRCh38
rs146272144 Chr.6: 160122276-160122276 on Build GRCh38
rs944991496 Chr.6: 160122506-160122506 on Build GRCh38
rs535669545 Chr.6: 160122230-160122230 on Build GRCh38
rs200409072 Chr.6: 160122171-160122171 on Build GRCh38
rs200083538 Chr.6: 160122321-160122321 on Build GRCh38
rs753993715 Chr.6: 160122337-160122337 on Build GRCh38
rs779853405 Chr.6: 160122390-160122390 on Build GRCh38
rs979810270 Chr.6: 160122583-160122583 on Build GRCh38
rs765183333 Chr.6: 160122342-160122342 on Build GRCh38
rs761669870 Chr.6: 160122251-160122251 on Build GRCh38
rs371895747 Chr.6: 160122394-160122394 on Build GRCh38
rs1034862401 Chr.6: 160122579-160122579 on Build GRCh38
rs188474761 Chr.6: 160122189-160122189 on Build GRCh38
rs4709400 Chr.6: 160122578-160122578 on Build GRCh38
rs548372769 Chr.6: 160122490-160122490 on Build GRCh38
rs759376914 Chr.6: 160122183-160122186 on Build GRCh38
rs138657404 Chr.6: 160122204-160122204 on Build GRCh38
rs186584856 Chr.6: 160122407-160122407 on Build GRCh38
rs461473 Chr.6: 160122530-160122530 on Build GRCh38
rs779207613 Chr.6: 160122211-160122211 on Build GRCh38
rs774374480 Chr.6: 160122322-160122322 on Build GRCh38
rs936203627 Chr.6: 160122383-160122383 on Build GRCh38
rs779713536 Chr.6: 160122517-160122517 on Build GRCh38
rs764109170 Chr.6: 160122240-160122240 on Build GRCh38
rs752701668 Chr.6: 160122179-160122179 on Build GRCh38
rs748565339 Chr.6: 160122282-160122282 on Build GRCh38
rs749749067 Chr.6: 160122163-160122163 on Build GRCh38
rs369823928 Chr.6: 160122357-160122357 on Build GRCh38
rs996656164 Chr.6: 160122265-160122265 on Build GRCh38

View Full Product Details