Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161135210-161135716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT4
Entrez Gene ID
56895
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 4
Gene Aliases
1-AGPAT4, LPAAT-delta, dJ473J16.2
UniGene
Hs.353175
Gene Chromosome Location
Chr.6: 161129980-161274108 on Build GRCh38
Transcripts
NM_020133.2 XM_005267053.3 XM_006715514.3 XM_017011059.1

SNP Detail

SNP ID Chromosome Location
rs1035599273 Chr.6: 161135666-161135666 on Build GRCh38
rs759217230 Chr.6: 161135623-161135623 on Build GRCh38
rs935566398 Chr.6: 161135528-161135528 on Build GRCh38
rs565434636 Chr.6: 161135478-161135478 on Build GRCh38
rs56073227 Chr.6: 161135379-161135379 on Build GRCh38
rs55978942 Chr.6: 161135561-161135561 on Build GRCh38
rs1014850348 Chr.6: 161135446-161135446 on Build GRCh38
rs138972524 Chr.6: 161135593-161135593 on Build GRCh38
rs76477931 Chr.6: 161135641-161135641 on Build GRCh38
rs1018497466 Chr.6: 161135575-161135575 on Build GRCh38
rs375715087 Chr.6: 161135525-161135525 on Build GRCh38
rs536370778 Chr.6: 161135368-161135368 on Build GRCh38
rs979585110 Chr.6: 161135518-161135518 on Build GRCh38
rs181349672 Chr.6: 161135424-161135424 on Build GRCh38
rs916697493 Chr.6: 161135483-161135483 on Build GRCh38
rs186311537 Chr.6: 161135602-161135602 on Build GRCh38
rs780133585 Chr.6: 161135326-161135326 on Build GRCh38
rs985697036 Chr.6: 161135334-161135334 on Build GRCh38
rs912216032 Chr.6: 161135545-161135545 on Build GRCh38
rs895427713 Chr.6: 161135441-161135441 on Build GRCh38
rs935583379 Chr.6: 161135258-161135258 on Build GRCh38
rs532363582 Chr.6: 161135655-161135655 on Build GRCh38
rs896482899 Chr.6: 161135694-161135694 on Build GRCh38
rs768697318 Chr.6: 161135353-161135353 on Build GRCh38
rs575919786 Chr.6: 161135508-161135508 on Build GRCh38
rs1052703938 Chr.6: 161135544-161135544 on Build GRCh38
rs369862930 Chr.6: 161135274-161135274 on Build GRCh38
rs1039499072 Chr.6: 161135347-161135347 on Build GRCh38
rs545027782 Chr.6: 161135453-161135453 on Build GRCh38
rs371650050 Chr.6: 161135288-161135288 on Build GRCh38
rs761908687 Chr.6: 161135423-161135423 on Build GRCh38
rs951082606 Chr.6: 161135607-161135607 on Build GRCh38
rs767310808 Chr.6: 161135646-161135646 on Build GRCh38
rs150586454 Chr.6: 161135666-161135667 on Build GRCh38
rs550131892 Chr.6: 161135369-161135371 on Build GRCh38
rs931059944 Chr.6: 161135601-161135601 on Build GRCh38
rs886955393 Chr.6: 161135624-161135624 on Build GRCh38
rs376258236 Chr.6: 161135312-161135312 on Build GRCh38
rs1052064640 Chr.6: 161135267-161135267 on Build GRCh38
rs1032470477 Chr.6: 161135543-161135544 on Build GRCh38
rs770232782 Chr.6: 161135436-161135436 on Build GRCh38
rs925449364 Chr.6: 161135527-161135527 on Build GRCh38
rs563893292 Chr.6: 161135356-161135356 on Build GRCh38
rs16892183 Chr.6: 161135246-161135246 on Build GRCh38
rs769049526 Chr.6: 161135350-161135351 on Build GRCh38
rs1032184941 Chr.6: 161135302-161135302 on Build GRCh38
rs573767680 Chr.6: 161135322-161135322 on Build GRCh38
rs546690940 Chr.6: 161135605-161135605 on Build GRCh38
rs909627574 Chr.6: 161135412-161135412 on Build GRCh38
rs55799452 Chr.6: 161135597-161135597 on Build GRCh38
rs983748658 Chr.6: 161135648-161135648 on Build GRCh38
rs899652981 Chr.6: 161135358-161135358 on Build GRCh38
rs966804087 Chr.6: 161135284-161135284 on Build GRCh38
rs976456598 Chr.6: 161135354-161135354 on Build GRCh38
rs1039466481 Chr.6: 161135591-161135591 on Build GRCh38
rs761279606 Chr.6: 161135631-161135632 on Build GRCh38
rs761290519 Chr.6: 161135484-161135484 on Build GRCh38
rs549709158 Chr.6: 161135566-161135568 on Build GRCh38
rs560776366 Chr.6: 161135395-161135395 on Build GRCh38
rs960713624 Chr.6: 161135448-161135448 on Build GRCh38
rs995388882 Chr.6: 161135372-161135372 on Build GRCh38
rs34041989 Chr.6: 161135457-161135458 on Build GRCh38
rs768339314 Chr.6: 161135337-161135337 on Build GRCh38
rs751151652 Chr.6: 161135610-161135610 on Build GRCh38

View Full Product Details