Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 162727461-162727981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PACRG
Entrez Gene ID
135138
Gene Name
PARK2 coregulated
Gene Aliases
GLUP, HAK005771, PARK2CRG
UniGene
Hs.25791
Gene Chromosome Location
Chr.6: 162726870-163315492 on Build GRCh38
Transcripts
NM_001080378.1 NM_001080379.1 NM_152410.2 XM_005266825.4 XM_005266826.4 XM_006715344.3 XM_006715345.3 XM_011535461.2 XM_011535462.2 XM_011535466.1 XM_011535468.2 XM_011535469.2 XM_011535471.1 XM_017010275.1 XM_017010276.1 XM_017010277.1 XM_017010278.1 XM_017010279.1 XM_017010280.1 NM_004562.2 NM_013987.2 NM_013988.2 XM_011535863.1 XM_017010909.1

Gene Symbol
PARK2
Entrez Gene ID
5071
Gene Name
parkin RBR E3 ubiquitin protein ligase
Gene Aliases
AR-JP, LPRS2, PDJ, PRKN
UniGene
Hs.132954
Gene Chromosome Location
Chr.6: 161347417-162727802 on Build GRCh38
Transcripts
NM_001080378.1 NM_001080379.1 NM_152410.2 XM_005266825.4 XM_005266826.4 XM_006715344.3 XM_006715345.3 XM_011535461.2 XM_011535462.2 XM_011535466.1 XM_011535468.2 XM_011535469.2 XM_011535471.1 XM_017010275.1 XM_017010276.1 XM_017010277.1 XM_017010278.1 XM_017010279.1 XM_017010280.1 NM_004562.2 NM_013987.2 NM_013988.2 XM_011535863.1 XM_017010909.1

SNP Detail

SNP ID Chromosome Location
rs534060238 Chr.6: 162727705-162727705 on Build GRCh38
rs1006067815 Chr.6: 162727539-162727539 on Build GRCh38
rs968217811 Chr.6: 162727957-162727957 on Build GRCh38
rs750626864 Chr.6: 162727725-162727725 on Build GRCh38
rs777729617 Chr.6: 162727736-162727736 on Build GRCh38
rs948590167 Chr.6: 162727870-162727870 on Build GRCh38
rs200065081 Chr.6: 162727698-162727698 on Build GRCh38
rs977031398 Chr.6: 162727776-162727776 on Build GRCh38
rs779917361 Chr.6: 162727625-162727625 on Build GRCh38
rs765760680 Chr.6: 162727679-162727679 on Build GRCh38
rs747682986 Chr.6: 162727665-162727665 on Build GRCh38
rs770336825 Chr.6: 162727723-162727725 on Build GRCh38
rs532978581 Chr.6: 162727558-162727558 on Build GRCh38
rs576793884 Chr.6: 162727711-162727711 on Build GRCh38
rs1008526612 Chr.6: 162727923-162727923 on Build GRCh38
rs771236536 Chr.6: 162727674-162727674 on Build GRCh38
rs770519631 Chr.6: 162727662-162727663 on Build GRCh38
rs954255204 Chr.6: 162727813-162727813 on Build GRCh38
rs771586218 Chr.6: 162727667-162727667 on Build GRCh38
rs756731328 Chr.6: 162727745-162727745 on Build GRCh38
rs762055309 Chr.6: 162727721-162727721 on Build GRCh38
rs974897520 Chr.6: 162727738-162727738 on Build GRCh38
rs922956128 Chr.6: 162727713-162727713 on Build GRCh38
rs778081992 Chr.6: 162727616-162727616 on Build GRCh38
rs778787823 Chr.6: 162727659-162727659 on Build GRCh38
rs751054800 Chr.6: 162727743-162727743 on Build GRCh38
rs1000207353 Chr.6: 162727960-162727960 on Build GRCh38
rs908838998 Chr.6: 162727829-162727829 on Build GRCh38
rs550065161 Chr.6: 162727573-162727573 on Build GRCh38
rs886106457 Chr.6: 162727902-162727902 on Build GRCh38
rs763058481 Chr.6: 162727686-162727686 on Build GRCh38
rs755930574 Chr.6: 162727620-162727620 on Build GRCh38
rs774538391 Chr.6: 162727717-162727717 on Build GRCh38
rs922924770 Chr.6: 162727497-162727497 on Build GRCh38
rs1039273628 Chr.6: 162727483-162727483 on Build GRCh38
rs931760147 Chr.6: 162727527-162727527 on Build GRCh38
rs369756736 Chr.6: 162727926-162727926 on Build GRCh38
rs112155221 Chr.6: 162727733-162727733 on Build GRCh38
rs764973407 Chr.6: 162727730-162727730 on Build GRCh38
rs897643603 Chr.6: 162727739-162727739 on Build GRCh38
rs201003791 Chr.6: 162727614-162727614 on Build GRCh38
rs1046186030 Chr.6: 162727950-162727950 on Build GRCh38
rs765761156 Chr.6: 162727746-162727746 on Build GRCh38
rs554458446 Chr.6: 162727886-162727887 on Build GRCh38
rs746211069 Chr.6: 162727707-162727707 on Build GRCh38
rs996368259 Chr.6: 162727947-162727947 on Build GRCh38
rs368314176 Chr.6: 162727706-162727706 on Build GRCh38
rs539607525 Chr.6: 162727961-162727961 on Build GRCh38
rs1040071219 Chr.6: 162727491-162727491 on Build GRCh38
rs754739638 Chr.6: 162727726-162727726 on Build GRCh38
rs1042482259 Chr.6: 162727523-162727523 on Build GRCh38
rs1022815802 Chr.6: 162727605-162727605 on Build GRCh38
rs200039995 Chr.6: 162727685-162727685 on Build GRCh38
rs541100410 Chr.6: 162727941-162727941 on Build GRCh38
rs950518517 Chr.6: 162727660-162727660 on Build GRCh38
rs745536814 Chr.6: 162727757-162727757 on Build GRCh38
rs547402532 Chr.6: 162727531-162727531 on Build GRCh38
rs528342807 Chr.6: 162727554-162727554 on Build GRCh38
rs540154030 Chr.6: 162727577-162727577 on Build GRCh38
rs757527178 Chr.6: 162727695-162727695 on Build GRCh38
rs746696823 Chr.6: 162727672-162727672 on Build GRCh38
rs1022460605 Chr.6: 162727951-162727951 on Build GRCh38
rs755667363 Chr.6: 162727500-162727500 on Build GRCh38
rs1013419206 Chr.6: 162727579-162727579 on Build GRCh38
rs770057490 Chr.6: 162727712-162727712 on Build GRCh38
rs369389244 Chr.6: 162727714-162727714 on Build GRCh38
rs994878069 Chr.6: 162727635-162727635 on Build GRCh38
rs749081702 Chr.6: 162727658-162727658 on Build GRCh38
rs112850281 Chr.6: 162727710-162727710 on Build GRCh38
rs978494336 Chr.6: 162727704-162727704 on Build GRCh38
rs397518439 Chr.6: 162727661-162727661 on Build GRCh38
rs542048864 Chr.6: 162727808-162727808 on Build GRCh38
rs939345880 Chr.6: 162727775-162727780 on Build GRCh38
rs965519063 Chr.6: 162727775-162727775 on Build GRCh38
rs549204834 Chr.6: 162727677-162727677 on Build GRCh38
rs111508614 Chr.6: 162727585-162727585 on Build GRCh38
rs776461846 Chr.6: 162727685-162727686 on Build GRCh38
rs886061239 Chr.6: 162727775-162727775 on Build GRCh38
rs949207875 Chr.6: 162727935-162727935 on Build GRCh38
rs1021162902 Chr.6: 162727774-162727774 on Build GRCh38
rs767771305 Chr.6: 162727724-162727724 on Build GRCh38
rs114231735 Chr.6: 162727551-162727551 on Build GRCh38
rs941821250 Chr.6: 162727835-162727835 on Build GRCh38
rs751780181 Chr.6: 162727692-162727692 on Build GRCh38
rs998737437 Chr.6: 162727565-162727565 on Build GRCh38
rs576721855 Chr.6: 162727791-162727791 on Build GRCh38
rs919118921 Chr.6: 162727903-162727903 on Build GRCh38
rs772786691 Chr.6: 162727668-162727668 on Build GRCh38
rs752584426 Chr.6: 162727731-162727731 on Build GRCh38
rs968685704 Chr.6: 162727751-162727751 on Build GRCh38
rs767676008 Chr.6: 162727696-162727696 on Build GRCh38
rs1047616333 Chr.6: 162727899-162727899 on Build GRCh38
rs764297261 Chr.6: 162727690-162727690 on Build GRCh38
rs900248963 Chr.6: 162727936-162727937 on Build GRCh38
rs985799098 Chr.6: 162727819-162727819 on Build GRCh38
rs561919613 Chr.6: 162727708-162727708 on Build GRCh38
rs916630380 Chr.6: 162727820-162727820 on Build GRCh38
rs914170852 Chr.6: 162727487-162727487 on Build GRCh38
rs1044207674 Chr.6: 162727871-162727871 on Build GRCh38
rs1046279256 Chr.6: 162727744-162727744 on Build GRCh38
rs556535340 Chr.6: 162727753-162727753 on Build GRCh38
rs1057350942 Chr.6: 162727575-162727575 on Build GRCh38
rs371617209 Chr.6: 162727707-162727707 on Build GRCh38
rs1032734680 Chr.6: 162727748-162727748 on Build GRCh38
rs572151187 Chr.6: 162727892-162727892 on Build GRCh38
rs1024963107 Chr.6: 162727581-162727581 on Build GRCh38
rs1036584200 Chr.6: 162727542-162727542 on Build GRCh38
rs887637359 Chr.6: 162727534-162727534 on Build GRCh38
rs751833256 Chr.6: 162727617-162727617 on Build GRCh38
rs1010059378 Chr.6: 162727769-162727769 on Build GRCh38
rs985766495 Chr.6: 162727719-162727719 on Build GRCh38
rs111721255 Chr.6: 162727887-162727887 on Build GRCh38
rs1036204101 Chr.6: 162727735-162727735 on Build GRCh38
rs944264003 Chr.6: 162727490-162727490 on Build GRCh38

View Full Product Details