Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 166329910-166330407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFT2D1
Entrez Gene ID
113402
Gene Name
SFT2 domain containing 1
Gene Aliases
C6orf83, pRGR1
UniGene
Hs.487143
Gene Chromosome Location
Chr.6: 166319728-166342606 on Build GRCh38
Transcripts
NM_145169.2 NR_130112.1 NR_130113.1 NR_130114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1442164 c.138C>A p.F46L Chr.6: 166330173-166330173 on Build GRCh38
4725377 c.124G>A p.V42I Chr.6: 166330187-166330187 on Build GRCh38
6398032 c.86G>A p.S29N Chr.6: 166330225-166330225 on Build GRCh38
277251 c.133G>A p.V45I Chr.6: 166330178-166330178 on Build GRCh38
4586464 c.64G>A p.V22I Chr.6: 166330247-166330247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757166819 Chr.6: 166330231-166330231 on Build GRCh38
rs750764998 Chr.6: 166329958-166329958 on Build GRCh38
rs1025424594 Chr.6: 166330361-166330361 on Build GRCh38
rs112844068 Chr.6: 166330134-166330134 on Build GRCh38
rs376245676 Chr.6: 166330235-166330235 on Build GRCh38
rs746052408 Chr.6: 166330166-166330166 on Build GRCh38
rs770028138 Chr.6: 166330175-166330175 on Build GRCh38
rs9348119 Chr.6: 166330272-166330272 on Build GRCh38
rs748205196 Chr.6: 166330206-166330206 on Build GRCh38
rs753133044 Chr.6: 166330131-166330131 on Build GRCh38
rs747099422 Chr.6: 166330143-166330143 on Build GRCh38
rs759122007 Chr.6: 166330279-166330279 on Build GRCh38
rs146041790 Chr.6: 166330179-166330179 on Build GRCh38
rs533249665 Chr.6: 166330241-166330241 on Build GRCh38
rs368692038 Chr.6: 166330297-166330297 on Build GRCh38
rs150012159 Chr.6: 166329999-166329999 on Build GRCh38
rs771809236 Chr.6: 166330268-166330268 on Build GRCh38
rs767665399 Chr.6: 166330118-166330118 on Build GRCh38
rs1010547891 Chr.6: 166329960-166329960 on Build GRCh38
rs139810206 Chr.6: 166330218-166330218 on Build GRCh38
rs561353559 Chr.6: 166329936-166329936 on Build GRCh38
rs893748232 Chr.6: 166329940-166329940 on Build GRCh38
rs991788468 Chr.6: 166330310-166330310 on Build GRCh38
rs902707095 Chr.6: 166329998-166329998 on Build GRCh38
rs764327004 Chr.6: 166330293-166330293 on Build GRCh38
rs962393077 Chr.6: 166330296-166330296 on Build GRCh38
rs143114200 Chr.6: 166330247-166330247 on Build GRCh38
rs756266337 Chr.6: 166330262-166330262 on Build GRCh38
rs117696269 Chr.6: 166330070-166330070 on Build GRCh38
rs1039014603 Chr.6: 166329944-166329945 on Build GRCh38
rs942629984 Chr.6: 166330187-166330187 on Build GRCh38
rs1035314104 Chr.6: 166330025-166330025 on Build GRCh38
rs572393571 Chr.6: 166330057-166330057 on Build GRCh38
rs778211426 Chr.6: 166330138-166330138 on Build GRCh38
rs1029833666 Chr.6: 166330058-166330058 on Build GRCh38
rs757331230 Chr.6: 166330144-166330144 on Build GRCh38
rs1017079423 Chr.6: 166330290-166330290 on Build GRCh38
rs376707135 Chr.6: 166330150-166330150 on Build GRCh38
rs769557631 Chr.6: 166330280-166330280 on Build GRCh38
rs772038544 Chr.6: 166330180-166330180 on Build GRCh38
rs528847565 Chr.6: 166329963-166329963 on Build GRCh38
rs373817495 Chr.6: 166330291-166330291 on Build GRCh38
rs375086661 Chr.6: 166330311-166330311 on Build GRCh38
rs565025799 Chr.6: 166330285-166330285 on Build GRCh38
rs201063880 Chr.6: 166330178-166330178 on Build GRCh38
rs991428763 Chr.6: 166330120-166330120 on Build GRCh38
rs201463834 Chr.6: 166330260-166330260 on Build GRCh38
rs779437566 Chr.6: 166330168-166330168 on Build GRCh38
rs910107791 Chr.6: 166330093-166330093 on Build GRCh38
rs770922561 Chr.6: 166330275-166330275 on Build GRCh38
rs752452116 Chr.6: 166330049-166330049 on Build GRCh38
rs371122813 Chr.6: 166330298-166330298 on Build GRCh38
rs368744851 Chr.6: 166330110-166330110 on Build GRCh38
rs982930639 Chr.6: 166330072-166330072 on Build GRCh38
rs757816564 Chr.6: 166330245-166330245 on Build GRCh38
rs371105029 Chr.6: 166330188-166330188 on Build GRCh38
rs780317547 Chr.6: 166330263-166330263 on Build GRCh38
rs773077255 Chr.6: 166330185-166330185 on Build GRCh38
rs370358469 Chr.6: 166330266-166330266 on Build GRCh38
rs181987230 Chr.6: 166330115-166330115 on Build GRCh38
rs772199367 Chr.6: 166330226-166330227 on Build GRCh38
rs752048093 Chr.6: 166330244-166330244 on Build GRCh38
rs1044259786 Chr.6: 166329971-166329971 on Build GRCh38
rs375032637 Chr.6: 166330212-166330212 on Build GRCh38
rs763285588 Chr.6: 166330236-166330236 on Build GRCh38
rs960359877 Chr.6: 166330046-166330046 on Build GRCh38
rs973867233 Chr.6: 166330100-166330100 on Build GRCh38
rs916158693 Chr.6: 166330317-166330317 on Build GRCh38
rs189112772 Chr.6: 166330277-166330277 on Build GRCh38
rs900562629 Chr.6: 166329953-166329956 on Build GRCh38

View Full Product Details