Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 170575844-170576356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDCD2
Entrez Gene ID
5134
Gene Name
programmed cell death 2
Gene Aliases
RP8, ZMYND7
UniGene
Hs.367900
Gene Chromosome Location
Chr.6: 170575572-170584692 on Build GRCh38
Transcripts
NM_001199462.1 NM_002598.3

SNP Detail

SNP ID Chromosome Location
rs12212765 Chr.6: 170576132-170576132 on Build GRCh38
rs753774949 Chr.6: 170576054-170576054 on Build GRCh38
rs781444862 Chr.6: 170576159-170576159 on Build GRCh38
rs367749345 Chr.6: 170576263-170576263 on Build GRCh38
rs577216277 Chr.6: 170576133-170576133 on Build GRCh38
rs59965148 Chr.6: 170576231-170576231 on Build GRCh38
rs1039195338 Chr.6: 170575885-170575885 on Build GRCh38
rs1007159035 Chr.6: 170576190-170576190 on Build GRCh38
rs993164771 Chr.6: 170576329-170576329 on Build GRCh38
rs746120487 Chr.6: 170576219-170576219 on Build GRCh38
rs766305251 Chr.6: 170576045-170576045 on Build GRCh38
rs954179760 Chr.6: 170575924-170575924 on Build GRCh38
rs956561893 Chr.6: 170576089-170576089 on Build GRCh38
rs778815174 Chr.6: 170576023-170576023 on Build GRCh38
rs900734485 Chr.6: 170575888-170575888 on Build GRCh38
rs565750767 Chr.6: 170576001-170576001 on Build GRCh38
rs6913308 Chr.6: 170576103-170576103 on Build GRCh38
rs149241428 Chr.6: 170576105-170576105 on Build GRCh38
rs963383300 Chr.6: 170575897-170575897 on Build GRCh38
rs913192182 Chr.6: 170575974-170575974 on Build GRCh38
rs115384362 Chr.6: 170575961-170575961 on Build GRCh38
rs35273447 Chr.6: 170576175-170576176 on Build GRCh38
rs746881516 Chr.6: 170576250-170576250 on Build GRCh38
rs927243878 Chr.6: 170576041-170576041 on Build GRCh38
rs994532951 Chr.6: 170576116-170576116 on Build GRCh38
rs778785938 Chr.6: 170576098-170576098 on Build GRCh38
rs1003408583 Chr.6: 170576264-170576264 on Build GRCh38
rs1012060619 Chr.6: 170576030-170576030 on Build GRCh38
rs574565450 Chr.6: 170576279-170576279 on Build GRCh38
rs193293086 Chr.6: 170576155-170576155 on Build GRCh38
rs188753274 Chr.6: 170575935-170575935 on Build GRCh38
rs767182647 Chr.6: 170575938-170575938 on Build GRCh38
rs548659610 Chr.6: 170576115-170576115 on Build GRCh38
rs899216744 Chr.6: 170576218-170576218 on Build GRCh38
rs376837597 Chr.6: 170575889-170575889 on Build GRCh38
rs982435504 Chr.6: 170576055-170576055 on Build GRCh38
rs116818292 Chr.6: 170576110-170576110 on Build GRCh38
rs770094038 Chr.6: 170576245-170576245 on Build GRCh38
rs1004916896 Chr.6: 170575947-170575947 on Build GRCh38
rs754975298 Chr.6: 170576097-170576097 on Build GRCh38
rs1020822464 Chr.6: 170576234-170576234 on Build GRCh38
rs752399456 Chr.6: 170576040-170576041 on Build GRCh38
rs111784233 Chr.6: 170575922-170575922 on Build GRCh38
rs978642476 Chr.6: 170576027-170576027 on Build GRCh38
rs570732769 Chr.6: 170576228-170576228 on Build GRCh38
rs968678557 Chr.6: 170576249-170576249 on Build GRCh38
rs144504968 Chr.6: 170576164-170576164 on Build GRCh38
rs1052436907 Chr.6: 170576121-170576121 on Build GRCh38
rs1031318823 Chr.6: 170576311-170576311 on Build GRCh38
rs985647211 Chr.6: 170575933-170575934 on Build GRCh38
rs1005269863 Chr.6: 170576157-170576157 on Build GRCh38
rs558795635 Chr.6: 170576059-170576059 on Build GRCh38
rs553163279 Chr.6: 170576253-170576253 on Build GRCh38
rs901489978 Chr.6: 170575887-170575887 on Build GRCh38
rs1019746195 Chr.6: 170576213-170576213 on Build GRCh38
rs995721557 Chr.6: 170575914-170575914 on Build GRCh38

View Full Product Details