Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156019935-156020444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSR2
Entrez Gene ID
6746
Gene Name
signal sequence receptor subunit 2
Gene Aliases
HSD25, TLAP, TRAP-BETA, TRAPB
UniGene
Hs.74564
Gene Chromosome Location
Chr.1: 156009048-156020992 on Build GRCh38
Transcripts
NM_003145.3 XM_011509923.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3475131 c.138C>T p.I46I Chr.1: 156020030-156020030 on Build GRCh38
1637936 c.99C>T p.Y33Y Chr.1: 156020069-156020069 on Build GRCh38
4816508 c.113G>A p.R38Q Chr.1: 156020055-156020055 on Build GRCh38
1334753 c.81A>C p.K27N Chr.1: 156020087-156020087 on Build GRCh38
3802181 c.31C>G p.L11V Chr.1: 156020137-156020137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374101844 Chr.1: 156020424-156020424 on Build GRCh38
rs771969846 Chr.1: 156020284-156020285 on Build GRCh38
rs973703937 Chr.1: 156020199-156020199 on Build GRCh38
rs376141987 Chr.1: 156019981-156019981 on Build GRCh38
rs924834570 Chr.1: 156020325-156020325 on Build GRCh38
rs184347581 Chr.1: 156020415-156020415 on Build GRCh38
rs939014981 Chr.1: 156020088-156020088 on Build GRCh38
rs754172358 Chr.1: 156020107-156020107 on Build GRCh38
rs745343478 Chr.1: 156019978-156019978 on Build GRCh38
rs374155755 Chr.1: 156020064-156020064 on Build GRCh38
rs377705476 Chr.1: 156020092-156020092 on Build GRCh38
rs768641620 Chr.1: 156020048-156020048 on Build GRCh38
rs185686947 Chr.1: 156020177-156020177 on Build GRCh38
rs778195793 Chr.1: 156020250-156020250 on Build GRCh38
rs775481673 Chr.1: 156020243-156020243 on Build GRCh38
rs770522519 Chr.1: 156020067-156020067 on Build GRCh38
rs754704677 Chr.1: 156020338-156020338 on Build GRCh38
rs990468414 Chr.1: 156020406-156020406 on Build GRCh38
rs1048843377 Chr.1: 156020167-156020167 on Build GRCh38
rs769718930 Chr.1: 156020065-156020065 on Build GRCh38
rs954076010 Chr.1: 156020317-156020317 on Build GRCh38
rs779728589 Chr.1: 156020324-156020324 on Build GRCh38
rs768370244 Chr.1: 156020187-156020187 on Build GRCh38
rs912058598 Chr.1: 156020306-156020306 on Build GRCh38
rs139854901 Chr.1: 156020379-156020379 on Build GRCh38
rs948798611 Chr.1: 156020413-156020413 on Build GRCh38
rs773914131 Chr.1: 156020174-156020174 on Build GRCh38
rs939634772 Chr.1: 156020404-156020404 on Build GRCh38
rs1028362128 Chr.1: 156020314-156020314 on Build GRCh38
rs745838065 Chr.1: 156020229-156020230 on Build GRCh38
rs760900867 Chr.1: 156020244-156020244 on Build GRCh38
rs764446062 Chr.1: 156020101-156020101 on Build GRCh38
rs767559812 Chr.1: 156020069-156020069 on Build GRCh38
rs752972978 Chr.1: 156020083-156020083 on Build GRCh38
rs989565504 Chr.1: 156020318-156020318 on Build GRCh38
rs149974874 Chr.1: 156020166-156020166 on Build GRCh38
rs774322271 Chr.1: 156020066-156020066 on Build GRCh38
rs878876479 Chr.1: 156020169-156020169 on Build GRCh38
rs764261705 Chr.1: 156020258-156020258 on Build GRCh38
rs11541383 Chr.1: 156020080-156020080 on Build GRCh38
rs769064155 Chr.1: 156020409-156020409 on Build GRCh38
rs192384622 Chr.1: 156020305-156020305 on Build GRCh38
rs567472407 Chr.1: 156020162-156020162 on Build GRCh38
rs894892826 Chr.1: 156020114-156020114 on Build GRCh38
rs879203903 Chr.1: 156020172-156020172 on Build GRCh38
rs571563268 Chr.1: 156020278-156020278 on Build GRCh38
rs747377886 Chr.1: 156020389-156020389 on Build GRCh38
rs779710271 Chr.1: 156020183-156020183 on Build GRCh38
rs978692157 Chr.1: 156020380-156020380 on Build GRCh38
rs778309648 Chr.1: 156019966-156019966 on Build GRCh38
rs1005179765 Chr.1: 156020234-156020234 on Build GRCh38
rs146742067 Chr.1: 156020137-156020137 on Build GRCh38
rs773247311 Chr.1: 156020242-156020242 on Build GRCh38
rs758911329 Chr.1: 156020311-156020311 on Build GRCh38
rs907285581 Chr.1: 156020417-156020417 on Build GRCh38
rs776506406 Chr.1: 156020060-156020060 on Build GRCh38
rs781153769 Chr.1: 156020195-156020195 on Build GRCh38
rs746667049 Chr.1: 156020184-156020184 on Build GRCh38
rs1015884925 Chr.1: 156020158-156020158 on Build GRCh38
rs769442300 Chr.1: 156020237-156020237 on Build GRCh38
rs977620624 Chr.1: 156020316-156020316 on Build GRCh38
rs568729816 Chr.1: 156020331-156020331 on Build GRCh38
rs545091940 Chr.1: 156020309-156020309 on Build GRCh38
rs753977773 Chr.1: 156020286-156020286 on Build GRCh38
rs746648955 Chr.1: 156020040-156020040 on Build GRCh38
rs762288878 Chr.1: 156020291-156020291 on Build GRCh38
rs930373058 Chr.1: 156020419-156020419 on Build GRCh38
rs750875111 Chr.1: 156020163-156020163 on Build GRCh38
rs189114352 Chr.1: 156020297-156020297 on Build GRCh38
rs920961427 Chr.1: 156020269-156020269 on Build GRCh38
rs747930857 Chr.1: 156020236-156020236 on Build GRCh38
rs779902655 Chr.1: 156020006-156020006 on Build GRCh38
rs759699925 Chr.1: 156020068-156020068 on Build GRCh38
rs546919651 Chr.1: 156020274-156020274 on Build GRCh38

View Full Product Details