Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156337072-156337583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT3
Entrez Gene ID
7203
Gene Name
chaperonin containing TCP1 subunit 3
Gene Aliases
CCT-gamma, CCTG, PIG48, TCP-1-gamma, TRIC5
UniGene
Hs.491494
Gene Chromosome Location
Chr.1: 156308961-156338415 on Build GRCh38
Transcripts
NM_001008800.2 NM_005998.4 NR_036564.1 NR_036565.1 NM_144627.4

Gene Symbol
TSACC
Entrez Gene ID
128229
Gene Name
TSSK6 activating cochaperone
Gene Aliases
C1orf182, SIP, SSTK-IP
UniGene
Hs.534539
Gene Chromosome Location
Chr.1: 156337314-156346995 on Build GRCh38
Transcripts
NM_001008800.2 NM_005998.4 NR_036564.1 NR_036565.1 NM_144627.4

SNP Detail

SNP ID Chromosome Location
rs553471511 Chr.1: 156337202-156337202 on Build GRCh38
rs922507580 Chr.1: 156337545-156337545 on Build GRCh38
rs551938933 Chr.1: 156337501-156337501 on Build GRCh38
rs369362506 Chr.1: 156337200-156337200 on Build GRCh38
rs1055535006 Chr.1: 156337110-156337110 on Build GRCh38
rs970222896 Chr.1: 156337492-156337492 on Build GRCh38
rs750643584 Chr.1: 156337462-156337462 on Build GRCh38
rs909685669 Chr.1: 156337199-156337199 on Build GRCh38
rs904675872 Chr.1: 156337343-156337343 on Build GRCh38
rs1022740161 Chr.1: 156337470-156337470 on Build GRCh38
rs183559338 Chr.1: 156337463-156337463 on Build GRCh38
rs574076002 Chr.1: 156337420-156337421 on Build GRCh38
rs777889482 Chr.1: 156337540-156337541 on Build GRCh38
rs1023130534 Chr.1: 156337156-156337156 on Build GRCh38
rs552555009 Chr.1: 156337106-156337106 on Build GRCh38
rs144069451 Chr.1: 156337176-156337176 on Build GRCh38
rs755579063 Chr.1: 156337180-156337180 on Build GRCh38
rs1043219948 Chr.1: 156337319-156337319 on Build GRCh38
rs943570308 Chr.1: 156337308-156337308 on Build GRCh38
rs1005279250 Chr.1: 156337515-156337515 on Build GRCh38
rs576186828 Chr.1: 156337452-156337452 on Build GRCh38
rs546176959 Chr.1: 156337416-156337416 on Build GRCh38
rs536096750 Chr.1: 156337501-156337502 on Build GRCh38
rs564479425 Chr.1: 156337433-156337433 on Build GRCh38
rs910610950 Chr.1: 156337301-156337301 on Build GRCh38
rs1016780447 Chr.1: 156337522-156337522 on Build GRCh38
rs1053268239 Chr.1: 156337414-156337414 on Build GRCh38
rs933308643 Chr.1: 156337237-156337237 on Build GRCh38
rs972678211 Chr.1: 156337540-156337540 on Build GRCh38
rs963494369 Chr.1: 156337266-156337266 on Build GRCh38
rs919272406 Chr.1: 156337216-156337216 on Build GRCh38
rs1016498725 Chr.1: 156337227-156337227 on Build GRCh38
rs556249107 Chr.1: 156337168-156337168 on Build GRCh38
rs113526416 Chr.1: 156337162-156337162 on Build GRCh38
rs1051894699 Chr.1: 156337263-156337263 on Build GRCh38
rs1029023745 Chr.1: 156337391-156337391 on Build GRCh38
rs972606100 Chr.1: 156337203-156337203 on Build GRCh38
rs11264476 Chr.1: 156337389-156337389 on Build GRCh38
rs752731728 Chr.1: 156337440-156337440 on Build GRCh38
rs1014317271 Chr.1: 156337469-156337469 on Build GRCh38
rs570654080 Chr.1: 156337125-156337125 on Build GRCh38
rs73008701 Chr.1: 156337528-156337528 on Build GRCh38
rs11264475 Chr.1: 156337127-156337127 on Build GRCh38
rs763483004 Chr.1: 156337515-156337515 on Build GRCh38
rs967674222 Chr.1: 156337428-156337428 on Build GRCh38

View Full Product Details