Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156736542-156737045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRNAD1
Entrez Gene ID
51093
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
Gene Aliases
C1orf66, CGI-41, METTL25B
UniGene
Hs.512597
Gene Chromosome Location
Chr.1: 156728471-156736960 on Build GRCh38
Transcripts
NM_001142560.1 NM_015997.3 XM_005245234.4 XM_005245235.4 XM_005245236.4 XM_005245237.4 XM_011509600.1 XM_017001404.1 XM_017001405.1 XM_017001406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
212098 c.1411G>T p.E471* Chr.1: 156736736-156736736 on Build GRCh38
6868767 c.1368G>T p.L456L Chr.1: 156736693-156736693 on Build GRCh38
5473156 c.1348T>G p.S450A Chr.1: 156736673-156736673 on Build GRCh38
274251 c.1309T>A p.F437I Chr.1: 156736634-156736634 on Build GRCh38
6019577 c.1399C>G p.L467V Chr.1: 156736724-156736724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957963509 Chr.1: 156736780-156736780 on Build GRCh38
rs750252048 Chr.1: 156736677-156736677 on Build GRCh38
rs944230421 Chr.1: 156737008-156737008 on Build GRCh38
rs768035988 Chr.1: 156736772-156736772 on Build GRCh38
rs919915728 Chr.1: 156736941-156736941 on Build GRCh38
rs752782468 Chr.1: 156736712-156736712 on Build GRCh38
rs928743623 Chr.1: 156736833-156736833 on Build GRCh38
rs746253249 Chr.1: 156736765-156736765 on Build GRCh38
rs559429776 Chr.1: 156736829-156736829 on Build GRCh38
rs190757969 Chr.1: 156736783-156736783 on Build GRCh38
rs759120773 Chr.1: 156736610-156736610 on Build GRCh38
rs1032005754 Chr.1: 156736769-156736769 on Build GRCh38
rs368220718 Chr.1: 156736619-156736619 on Build GRCh38
rs931548513 Chr.1: 156736654-156736654 on Build GRCh38
rs1008403511 Chr.1: 156736981-156736981 on Build GRCh38
rs750199052 Chr.1: 156736790-156736790 on Build GRCh38
rs988042279 Chr.1: 156736795-156736795 on Build GRCh38
rs763696807 Chr.1: 156736644-156736644 on Build GRCh38
rs974444553 Chr.1: 156736570-156736570 on Build GRCh38
rs759901013 Chr.1: 156736977-156736977 on Build GRCh38
rs749789707 Chr.1: 156736753-156736753 on Build GRCh38
rs372196050 Chr.1: 156736646-156736646 on Build GRCh38
rs761902145 Chr.1: 156736650-156736650 on Build GRCh38
rs755174216 Chr.1: 156736702-156736702 on Build GRCh38
rs371938329 Chr.1: 156737019-156737019 on Build GRCh38
rs1012609200 Chr.1: 156736824-156736824 on Build GRCh38
rs1019365880 Chr.1: 156736740-156736740 on Build GRCh38
rs1047250104 Chr.1: 156736661-156736661 on Build GRCh38
rs937360386 Chr.1: 156736565-156736565 on Build GRCh38
rs8658 Chr.1: 156736767-156736767 on Build GRCh38
rs764705985 Chr.1: 156736820-156736820 on Build GRCh38
rs760421283 Chr.1: 156736768-156736768 on Build GRCh38
rs776349607 Chr.1: 156736773-156736773 on Build GRCh38
rs374747953 Chr.1: 156736724-156736724 on Build GRCh38
rs1047322306 Chr.1: 156736988-156736988 on Build GRCh38
rs970604715 Chr.1: 156736811-156736811 on Build GRCh38
rs147105055 Chr.1: 156736992-156736992 on Build GRCh38
rs528097731 Chr.1: 156736886-156736886 on Build GRCh38
rs530443556 Chr.1: 156736670-156736671 on Build GRCh38
rs142171344 Chr.1: 156736651-156736651 on Build GRCh38
rs747939658 Chr.1: 156736595-156736595 on Build GRCh38
rs1031104171 Chr.1: 156736923-156736923 on Build GRCh38
rs368419419 Chr.1: 156736628-156736628 on Build GRCh38
rs759719007 Chr.1: 156736715-156736715 on Build GRCh38
rs937420016 Chr.1: 156736912-156736912 on Build GRCh38
rs1043728698 Chr.1: 156736827-156736827 on Build GRCh38
rs779190533 Chr.1: 156736762-156736762 on Build GRCh38
rs771635690 Chr.1: 156736611-156736611 on Build GRCh38
rs374022799 Chr.1: 156736804-156736804 on Build GRCh38
rs371732973 Chr.1: 156736893-156736896 on Build GRCh38
rs903875399 Chr.1: 156736873-156736873 on Build GRCh38
rs778175002 Chr.1: 156736592-156736592 on Build GRCh38
rs529190645 Chr.1: 156736991-156737005 on Build GRCh38
rs568875979 Chr.1: 156736980-156736980 on Build GRCh38
rs547880000 Chr.1: 156736990-156736990 on Build GRCh38
rs775011822 Chr.1: 156736620-156736620 on Build GRCh38
rs761321745 Chr.1: 156736778-156736778 on Build GRCh38
rs180671078 Chr.1: 156736987-156736987 on Build GRCh38
rs991472969 Chr.1: 156736940-156736940 on Build GRCh38
rs765282483 Chr.1: 156736786-156736786 on Build GRCh38
rs775705992 Chr.1: 156736666-156736667 on Build GRCh38
rs139211741 Chr.1: 156736709-156736709 on Build GRCh38
rs774762010 Chr.1: 156736596-156736596 on Build GRCh38
rs752182232 Chr.1: 156736897-156736897 on Build GRCh38
rs150264626 Chr.1: 156736890-156736893 on Build GRCh38
rs749835869 Chr.1: 156736593-156736593 on Build GRCh38
rs777960533 Chr.1: 156736741-156736741 on Build GRCh38
rs866586384 Chr.1: 156736918-156736918 on Build GRCh38

View Full Product Details