Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156841864-156842402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSRR
Entrez Gene ID
3645
Gene Name
insulin receptor related receptor
Gene Aliases
IRR
UniGene
Hs.248138
Gene Chromosome Location
Chr.1: 156840873-156858920 on Build GRCh38
Transcripts
NM_014215.2 NM_001007792.1

Gene Symbol
NTRK1
Entrez Gene ID
4914
Gene Name
neurotrophic receptor tyrosine kinase 1
Gene Aliases
MTC, TRK, TRK1, TRKA, Trk-A, p140-TrkA
UniGene
Hs.406293
Gene Chromosome Location
Chr.1: 156815750-156881850 on Build GRCh38
Transcripts
NM_014215.2 NM_001007792.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6459420 c.3249G>T p.G1083G Chr.1: 156842260-156842260 on Build GRCh38
3740717 c.52A>G p.T18A Chr.1: 156842123-156842123 on Build GRCh38
99212 c.13G>A p.A5T Chr.1: 156842084-156842084 on Build GRCh38
5843998 c.83G>A p.R28Q Chr.1: 156842154-156842154 on Build GRCh38
336600 c.3346C>A p.L1116I Chr.1: 156842163-156842163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755716153 Chr.1: 156842153-156842153 on Build GRCh38
rs866237041 Chr.1: 156842036-156842036 on Build GRCh38
rs1054768557 Chr.1: 156842372-156842372 on Build GRCh38
rs532005876 Chr.1: 156842286-156842286 on Build GRCh38
rs758128799 Chr.1: 156842161-156842161 on Build GRCh38
rs572654538 Chr.1: 156842284-156842284 on Build GRCh38
rs1035402024 Chr.1: 156842124-156842124 on Build GRCh38
rs547897400 Chr.1: 156841948-156841948 on Build GRCh38
rs759794005 Chr.1: 156842127-156842127 on Build GRCh38
rs778917490 Chr.1: 156842090-156842090 on Build GRCh38
rs201742278 Chr.1: 156842233-156842233 on Build GRCh38
rs1039164689 Chr.1: 156842070-156842070 on Build GRCh38
rs145596747 Chr.1: 156842104-156842104 on Build GRCh38
rs745647077 Chr.1: 156842091-156842091 on Build GRCh38
rs1000057562 Chr.1: 156842361-156842361 on Build GRCh38
rs771088422 Chr.1: 156842205-156842205 on Build GRCh38
rs924544018 Chr.1: 156841991-156841991 on Build GRCh38
rs765290647 Chr.1: 156842035-156842035 on Build GRCh38
rs772504661 Chr.1: 156842318-156842318 on Build GRCh38
rs755433838 Chr.1: 156842051-156842051 on Build GRCh38
rs745868240 Chr.1: 156842179-156842179 on Build GRCh38
rs145039687 Chr.1: 156842238-156842238 on Build GRCh38
rs982208257 Chr.1: 156842211-156842211 on Build GRCh38
rs770719969 Chr.1: 156842085-156842085 on Build GRCh38
rs543738366 Chr.1: 156842340-156842340 on Build GRCh38
rs746766711 Chr.1: 156842206-156842206 on Build GRCh38
rs777419942 Chr.1: 156842154-156842154 on Build GRCh38
rs878923534 Chr.1: 156841972-156841972 on Build GRCh38
rs771963500 Chr.1: 156842182-156842182 on Build GRCh38
rs944775599 Chr.1: 156842333-156842333 on Build GRCh38
rs1048256058 Chr.1: 156842087-156842087 on Build GRCh38
rs771085993 Chr.1: 156842290-156842290 on Build GRCh38
rs767907631 Chr.1: 156842130-156842130 on Build GRCh38
rs756395014 Chr.1: 156842277-156842277 on Build GRCh38
rs756700664 Chr.1: 156842243-156842243 on Build GRCh38
rs545349562 Chr.1: 156842276-156842276 on Build GRCh38
rs761017753 Chr.1: 156842349-156842349 on Build GRCh38
rs761488096 Chr.1: 156842374-156842374 on Build GRCh38
rs970859133 Chr.1: 156841956-156841956 on Build GRCh38
rs780805763 Chr.1: 156842055-156842055 on Build GRCh38
rs770161044 Chr.1: 156842108-156842108 on Build GRCh38
rs376720827 Chr.1: 156842100-156842100 on Build GRCh38
rs774437818 Chr.1: 156842300-156842300 on Build GRCh38
rs779943666 Chr.1: 156842190-156842190 on Build GRCh38
rs79020702 Chr.1: 156842014-156842014 on Build GRCh38
rs542461394 Chr.1: 156842065-156842066 on Build GRCh38
rs1032968973 Chr.1: 156842164-156842164 on Build GRCh38
rs180942214 Chr.1: 156842068-156842068 on Build GRCh38
rs749935027 Chr.1: 156842259-156842259 on Build GRCh38
rs13376034 Chr.1: 156842353-156842353 on Build GRCh38
rs1031996312 Chr.1: 156842273-156842273 on Build GRCh38
rs988965088 Chr.1: 156841945-156841945 on Build GRCh38
rs777563118 Chr.1: 156842185-156842185 on Build GRCh38
rs558577855 Chr.1: 156842020-156842020 on Build GRCh38
rs748854792 Chr.1: 156842084-156842084 on Build GRCh38
rs772332648 Chr.1: 156842174-156842174 on Build GRCh38
rs745962645 Chr.1: 156842311-156842311 on Build GRCh38
rs201633542 Chr.1: 156842195-156842195 on Build GRCh38
rs776116756 Chr.1: 156842362-156842362 on Build GRCh38
rs974057880 Chr.1: 156842278-156842278 on Build GRCh38
rs752185710 Chr.1: 156841974-156841974 on Build GRCh38
rs1038903772 Chr.1: 156842110-156842110 on Build GRCh38
rs955767102 Chr.1: 156841905-156841905 on Build GRCh38
rs200688061 Chr.1: 156842199-156842199 on Build GRCh38
rs749311435 Chr.1: 156842203-156842203 on Build GRCh38
rs372048671 Chr.1: 156842040-156842040 on Build GRCh38
rs768036718 Chr.1: 156842218-156842218 on Build GRCh38
rs764821124 Chr.1: 156842376-156842376 on Build GRCh38
rs139555558 Chr.1: 156842095-156842095 on Build GRCh38
rs200176126 Chr.1: 156842202-156842202 on Build GRCh38
rs758351138 Chr.1: 156842266-156842266 on Build GRCh38
rs763736073 Chr.1: 156842224-156842224 on Build GRCh38
rs777227609 Chr.1: 156842083-156842083 on Build GRCh38
rs148424816 Chr.1: 156842082-156842082 on Build GRCh38
rs199647144 Chr.1: 156842109-156842109 on Build GRCh38
rs761036015 Chr.1: 156842220-156842220 on Build GRCh38
rs753089317 Chr.1: 156842140-156842140 on Build GRCh38
rs566121796 Chr.1: 156841994-156841994 on Build GRCh38
rs374119198 Chr.1: 156841889-156841889 on Build GRCh38
rs189837619 Chr.1: 156842285-156842285 on Build GRCh38
rs753223410 Chr.1: 156842168-156842168 on Build GRCh38
rs752162923 Chr.1: 156842122-156842122 on Build GRCh38
rs752394184 Chr.1: 156842067-156842067 on Build GRCh38
rs756862919 Chr.1: 156842169-156842169 on Build GRCh38
rs35265289 Chr.1: 156841994-156841995 on Build GRCh38
rs147703846 Chr.1: 156842117-156842117 on Build GRCh38
rs150557473 Chr.1: 156842244-156842244 on Build GRCh38
rs759993634 Chr.1: 156842217-156842217 on Build GRCh38
rs978648819 Chr.1: 156841989-156841990 on Build GRCh38

View Full Product Details