Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27101046-27101537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA2
Entrez Gene ID
3199
Gene Name
homeobox A2
Gene Aliases
HOX1K, MCOHI
UniGene
Hs.445239
Gene Chromosome Location
Chr.7: 27100354-27102775 on Build GRCh38
Transcripts
NM_006735.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7312314 c.793C>A p.H265N Chr.7: 27101064-27101064 on Build GRCh38
4361803 c.661G>A p.E221K Chr.7: 27101196-27101196 on Build GRCh38
3027088 c.507C>A p.C169* Chr.7: 27101350-27101350 on Build GRCh38
7211334 c.660C>A p.S220S Chr.7: 27101197-27101197 on Build GRCh38
107066 c.519G>A p.R173R Chr.7: 27101338-27101338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373239286 Chr.7: 27101182-27101182 on Build GRCh38
rs746566874 Chr.7: 27101164-27101164 on Build GRCh38
rs758386563 Chr.7: 27101249-27101249 on Build GRCh38
rs752578788 Chr.7: 27101247-27101247 on Build GRCh38
rs746654616 Chr.7: 27101066-27101066 on Build GRCh38
rs770656809 Chr.7: 27101067-27101067 on Build GRCh38
rs762796761 Chr.7: 27101359-27101359 on Build GRCh38
rs773679395 Chr.7: 27101180-27101182 on Build GRCh38
rs372854321 Chr.7: 27101151-27101151 on Build GRCh38
rs762323908 Chr.7: 27101092-27101092 on Build GRCh38
rs779221330 Chr.7: 27101275-27101275 on Build GRCh38
rs772611071 Chr.7: 27101198-27101198 on Build GRCh38
rs771410610 Chr.7: 27101445-27101445 on Build GRCh38
rs748564970 Chr.7: 27101303-27101303 on Build GRCh38
rs758659237 Chr.7: 27101517-27101517 on Build GRCh38
rs776308114 Chr.7: 27101170-27101170 on Build GRCh38
rs763328645 Chr.7: 27101070-27101070 on Build GRCh38
rs995483551 Chr.7: 27101093-27101093 on Build GRCh38
rs967958117 Chr.7: 27101109-27101109 on Build GRCh38
rs201139490 Chr.7: 27101494-27101494 on Build GRCh38
rs772364626 Chr.7: 27101439-27101439 on Build GRCh38
rs769998504 Chr.7: 27101468-27101468 on Build GRCh38
rs761096280 Chr.7: 27101323-27101323 on Build GRCh38
rs775695374 Chr.7: 27101069-27101069 on Build GRCh38
rs758474437 Chr.7: 27101161-27101161 on Build GRCh38
rs917703849 Chr.7: 27101230-27101230 on Build GRCh38
rs762305952 Chr.7: 27101197-27101197 on Build GRCh38
rs749965554 Chr.7: 27101426-27101426 on Build GRCh38
rs767574059 Chr.7: 27101095-27101095 on Build GRCh38
rs116885108 Chr.7: 27101167-27101167 on Build GRCh38
rs756721464 Chr.7: 27101255-27101255 on Build GRCh38
rs1011085616 Chr.7: 27101431-27101431 on Build GRCh38
rs773638633 Chr.7: 27101320-27101320 on Build GRCh38
rs777386300 Chr.7: 27101162-27101162 on Build GRCh38
rs751457659 Chr.7: 27101395-27101395 on Build GRCh38
rs372030111 Chr.7: 27101510-27101510 on Build GRCh38
rs766323058 Chr.7: 27101492-27101497 on Build GRCh38
rs756301122 Chr.7: 27101102-27101102 on Build GRCh38
rs748975641 Chr.7: 27101437-27101437 on Build GRCh38
rs761803005 Chr.7: 27101503-27101503 on Build GRCh38
rs780582283 Chr.7: 27101258-27101258 on Build GRCh38
rs951464048 Chr.7: 27101115-27101115 on Build GRCh38
rs757201406 Chr.7: 27101408-27101408 on Build GRCh38
rs752989766 Chr.7: 27101356-27101356 on Build GRCh38
rs766677700 Chr.7: 27101108-27101108 on Build GRCh38
rs1051989126 Chr.7: 27101223-27101223 on Build GRCh38
rs765075132 Chr.7: 27101234-27101234 on Build GRCh38
rs755775462 Chr.7: 27101432-27101432 on Build GRCh38
rs751579648 Chr.7: 27101251-27101251 on Build GRCh38
rs780752179 Chr.7: 27101417-27101417 on Build GRCh38
rs760763670 Chr.7: 27101215-27101215 on Build GRCh38
rs887991299 Chr.7: 27101324-27101324 on Build GRCh38
rs763986379 Chr.7: 27101365-27101365 on Build GRCh38
rs119489104 Chr.7: 27101301-27101301 on Build GRCh38
rs184109877 Chr.7: 27101260-27101260 on Build GRCh38
rs767409984 Chr.7: 27101507-27101507 on Build GRCh38
rs377466867 Chr.7: 27101227-27101227 on Build GRCh38
rs959149385 Chr.7: 27101163-27101163 on Build GRCh38
rs775814582 Chr.7: 27101487-27101487 on Build GRCh38
rs772381717 Chr.7: 27101438-27101438 on Build GRCh38
rs760839440 Chr.7: 27101236-27101247 on Build GRCh38
rs754731827 Chr.7: 27101142-27101142 on Build GRCh38
rs745512420 Chr.7: 27101172-27101172 on Build GRCh38
rs769008688 Chr.7: 27101178-27101178 on Build GRCh38
rs750572076 Chr.7: 27101097-27101097 on Build GRCh38
rs140982794 Chr.7: 27101338-27101338 on Build GRCh38
rs774052606 Chr.7: 27101501-27101501 on Build GRCh38
rs754033891 Chr.7: 27101125-27101125 on Build GRCh38
rs139504171 Chr.7: 27101463-27101463 on Build GRCh38
rs398122360 Chr.7: 27101154-27101154 on Build GRCh38
rs763957850 Chr.7: 27101497-27101497 on Build GRCh38
rs61740490 Chr.7: 27101248-27101248 on Build GRCh38
rs776638973 Chr.7: 27101336-27101336 on Build GRCh38
rs991740115 Chr.7: 27101165-27101165 on Build GRCh38
rs778583563 Chr.7: 27101149-27101149 on Build GRCh38
rs968963087 Chr.7: 27101196-27101196 on Build GRCh38
rs558922282 Chr.7: 27101263-27101263 on Build GRCh38
rs774856311 Chr.7: 27101087-27101087 on Build GRCh38
rs764398309 Chr.7: 27101079-27101079 on Build GRCh38
rs765603928 Chr.7: 27101344-27101344 on Build GRCh38
rs779619613 Chr.7: 27101436-27101436 on Build GRCh38
rs766370249 Chr.7: 27101218-27101218 on Build GRCh38
rs146316132 Chr.7: 27101202-27101202 on Build GRCh38
rs776319129 Chr.7: 27101484-27101506 on Build GRCh38
rs201289702 Chr.7: 27101179-27101179 on Build GRCh38
rs747522109 Chr.7: 27101440-27101440 on Build GRCh38
rs776760448 Chr.7: 27101446-27101446 on Build GRCh38

View Full Product Details