Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27739140-27739661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAX1BP1
Entrez Gene ID
8887
Gene Name
Tax1 binding protein 1
Gene Aliases
CALCOCO3, T6BP, TXBP151
UniGene
Hs.34576
Gene Chromosome Location
Chr.7: 27739373-27829767 on Build GRCh38
Transcripts
NM_001206901.1

SNP Detail

SNP ID Chromosome Location
rs770683405 Chr.7: 27739394-27739394 on Build GRCh38
rs141551414 Chr.7: 27739626-27739626 on Build GRCh38
rs1050779015 Chr.7: 27739547-27739547 on Build GRCh38
rs980248323 Chr.7: 27739264-27739264 on Build GRCh38
rs17553448 Chr.7: 27739338-27739338 on Build GRCh38
rs546425993 Chr.7: 27739439-27739439 on Build GRCh38
rs1051261162 Chr.7: 27739322-27739322 on Build GRCh38
rs971503631 Chr.7: 27739405-27739405 on Build GRCh38
rs554637030 Chr.7: 27739188-27739188 on Build GRCh38
rs1040644158 Chr.7: 27739549-27739549 on Build GRCh38
rs1024834534 Chr.7: 27739403-27739403 on Build GRCh38
rs544965352 Chr.7: 27739615-27739615 on Build GRCh38
rs540325190 Chr.7: 27739259-27739259 on Build GRCh38
rs892078636 Chr.7: 27739319-27739319 on Build GRCh38
rs1029108700 Chr.7: 27739589-27739589 on Build GRCh38
rs538479331 Chr.7: 27739453-27739453 on Build GRCh38
rs116394648 Chr.7: 27739612-27739612 on Build GRCh38
rs17155727 Chr.7: 27739323-27739323 on Build GRCh38
rs1000756865 Chr.7: 27739616-27739616 on Build GRCh38
rs2237337 Chr.7: 27739266-27739266 on Build GRCh38
rs371829224 Chr.7: 27739311-27739311 on Build GRCh38
rs573601510 Chr.7: 27739285-27739285 on Build GRCh38
rs542517339 Chr.7: 27739320-27739320 on Build GRCh38
rs980300727 Chr.7: 27739433-27739433 on Build GRCh38
rs79867569 Chr.7: 27739332-27739332 on Build GRCh38
rs947928084 Chr.7: 27739430-27739430 on Build GRCh38
rs375225509 Chr.7: 27739539-27739539 on Build GRCh38
rs917436724 Chr.7: 27739204-27739204 on Build GRCh38
rs750352519 Chr.7: 27739287-27739287 on Build GRCh38
rs75146008 Chr.7: 27739586-27739586 on Build GRCh38
rs776751695 Chr.7: 27739249-27739253 on Build GRCh38
rs966245698 Chr.7: 27739326-27739326 on Build GRCh38
rs780576820 Chr.7: 27739206-27739206 on Build GRCh38
rs6952476 Chr.7: 27739361-27739361 on Build GRCh38
rs902070212 Chr.7: 27739571-27739571 on Build GRCh38
rs1052806430 Chr.7: 27739622-27739622 on Build GRCh38
rs1039501926 Chr.7: 27739336-27739336 on Build GRCh38
rs912333258 Chr.7: 27739569-27739569 on Build GRCh38
rs946005808 Chr.7: 27739483-27739483 on Build GRCh38
rs577557363 Chr.7: 27739281-27739281 on Build GRCh38
rs750562053 Chr.7: 27739196-27739196 on Build GRCh38
rs757104196 Chr.7: 27739205-27739214 on Build GRCh38
rs755244839 Chr.7: 27739288-27739288 on Build GRCh38
rs1000697854 Chr.7: 27739351-27739351 on Build GRCh38
rs917796561 Chr.7: 27739404-27739404 on Build GRCh38
rs184855260 Chr.7: 27739503-27739503 on Build GRCh38
rs868069508 Chr.7: 27739468-27739468 on Build GRCh38
rs570990115 Chr.7: 27739449-27739449 on Build GRCh38
rs917759758 Chr.7: 27739172-27739172 on Build GRCh38
rs1054501109 Chr.7: 27739473-27739473 on Build GRCh38
rs927413503 Chr.7: 27739438-27739438 on Build GRCh38
rs1038052071 Chr.7: 27739303-27739303 on Build GRCh38
rs1054448384 Chr.7: 27739297-27739297 on Build GRCh38
rs748824988 Chr.7: 27739375-27739375 on Build GRCh38
rs148614766 Chr.7: 27739479-27739479 on Build GRCh38
rs773984410 Chr.7: 27739536-27739536 on Build GRCh38
rs866870380 Chr.7: 27739269-27739269 on Build GRCh38
rs951785211 Chr.7: 27739333-27739333 on Build GRCh38
rs573289513 Chr.7: 27739214-27739214 on Build GRCh38
rs887567258 Chr.7: 27739608-27739608 on Build GRCh38

View Full Product Details