Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27830456-27830978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAZF1
Entrez Gene ID
221895
Gene Name
JAZF zinc finger 1
Gene Aliases
TIP27, ZNF802
UniGene
Hs.368944
Gene Chromosome Location
Chr.7: 27830573-28180818 on Build GRCh38
Transcripts
NM_175061.3 XM_006715656.1 XM_017011831.1 XM_017011832.1

SNP Detail

SNP ID Chromosome Location
rs993979580 Chr.7: 27830695-27830695 on Build GRCh38
rs780514293 Chr.7: 27830482-27830482 on Build GRCh38
rs182116025 Chr.7: 27830610-27830610 on Build GRCh38
rs1044432330 Chr.7: 27830585-27830585 on Build GRCh38
rs1050898152 Chr.7: 27830920-27830920 on Build GRCh38
rs542828592 Chr.7: 27830551-27830552 on Build GRCh38
rs891081512 Chr.7: 27830941-27830941 on Build GRCh38
rs879536788 Chr.7: 27830769-27830769 on Build GRCh38
rs1038091424 Chr.7: 27830660-27830660 on Build GRCh38
rs528070280 Chr.7: 27830924-27830924 on Build GRCh38
rs949208255 Chr.7: 27830729-27830729 on Build GRCh38
rs891035621 Chr.7: 27830716-27830716 on Build GRCh38
rs373101809 Chr.7: 27830844-27830848 on Build GRCh38
rs917468203 Chr.7: 27830561-27830561 on Build GRCh38
rs997726449 Chr.7: 27830562-27830565 on Build GRCh38
rs1029599149 Chr.7: 27830699-27830699 on Build GRCh38
rs963401027 Chr.7: 27830683-27830683 on Build GRCh38
rs979829468 Chr.7: 27830810-27830810 on Build GRCh38
rs918313273 Chr.7: 27830900-27830900 on Build GRCh38
rs544647241 Chr.7: 27830842-27830846 on Build GRCh38
rs1009334372 Chr.7: 27830484-27830484 on Build GRCh38
rs926962412 Chr.7: 27830813-27830813 on Build GRCh38
rs544991264 Chr.7: 27830606-27830606 on Build GRCh38
rs544785594 Chr.7: 27830719-27830719 on Build GRCh38
rs189942592 Chr.7: 27830495-27830495 on Build GRCh38
rs772801836 Chr.7: 27830775-27830775 on Build GRCh38
rs549013181 Chr.7: 27830806-27830806 on Build GRCh38
rs917520509 Chr.7: 27830791-27830791 on Build GRCh38
rs774857678 Chr.7: 27830746-27830746 on Build GRCh38
rs1043364 Chr.7: 27830576-27830576 on Build GRCh38
rs1007227184 Chr.7: 27830942-27830942 on Build GRCh38
rs3182941 Chr.7: 27830697-27830697 on Build GRCh38
rs903335494 Chr.7: 27830525-27830526 on Build GRCh38
rs764397070 Chr.7: 27830846-27830847 on Build GRCh38
rs1030993163 Chr.7: 27830570-27830570 on Build GRCh38
rs1038547820 Chr.7: 27830877-27830877 on Build GRCh38
rs1007821599 Chr.7: 27830908-27830908 on Build GRCh38
rs573984334 Chr.7: 27830572-27830572 on Build GRCh38
rs972051928 Chr.7: 27830793-27830793 on Build GRCh38
rs929824600 Chr.7: 27830913-27830913 on Build GRCh38
rs989039349 Chr.7: 27830518-27830518 on Build GRCh38
rs998090706 Chr.7: 27830885-27830885 on Build GRCh38
rs988924664 Chr.7: 27830785-27830785 on Build GRCh38
rs1020442525 Chr.7: 27830519-27830519 on Build GRCh38
rs140941341 Chr.7: 27830889-27830889 on Build GRCh38
rs896994784 Chr.7: 27830675-27830675 on Build GRCh38
rs981998615 Chr.7: 27830768-27830768 on Build GRCh38
rs563040904 Chr.7: 27830720-27830720 on Build GRCh38
rs775537665 Chr.7: 27830904-27830905 on Build GRCh38
rs183893860 Chr.7: 27830770-27830770 on Build GRCh38
rs1018014854 Chr.7: 27830743-27830743 on Build GRCh38
rs940997996 Chr.7: 27830814-27830814 on Build GRCh38

View Full Product Details