Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 33177234-33177743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBS9
Entrez Gene ID
27241
Gene Name
Bardet-Biedl syndrome 9
Gene Aliases
B1, C18, D1, PTHB1
UniGene
Hs.372360
Gene Chromosome Location
Chr.7: 33129244-33637238 on Build GRCh38
Transcripts
NM_001033604.1 NM_001033605.1 NM_014451.3 NM_198428.2 XM_005249700.3 XM_005249701.2 XM_011515264.2 XM_011515265.2 XM_011515266.2 XM_011515267.2 XM_011515268.1 XM_011515269.2 XM_011515270.2 XM_017011990.1 XM_017011991.1 XM_017011992.1 XM_017011993.1 XM_017011994.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4566595 c.442_442+1GG>AA p.? Chr.7: 33177591-33177592 on Build GRCh38
6109977 c.354G>T p.G118G Chr.7: 33177503-33177503 on Build GRCh38
23436 c.404C>T p.A135V Chr.7: 33177553-33177553 on Build GRCh38
5432923 c.329-6C>T p.? Chr.7: 33177472-33177472 on Build GRCh38
4362106 c.335T>C p.L112S Chr.7: 33177484-33177484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009533507 Chr.7: 33177402-33177402 on Build GRCh38
rs749403663 Chr.7: 33177507-33177507 on Build GRCh38
rs748506036 Chr.7: 33177633-33177633 on Build GRCh38
rs74591112 Chr.7: 33177543-33177543 on Build GRCh38
rs753552530 Chr.7: 33177460-33177462 on Build GRCh38
rs780063415 Chr.7: 33177498-33177498 on Build GRCh38
rs760353562 Chr.7: 33177538-33177538 on Build GRCh38
rs545925422 Chr.7: 33177591-33177591 on Build GRCh38
rs369955057 Chr.7: 33177621-33177621 on Build GRCh38
rs57972085 Chr.7: 33177327-33177327 on Build GRCh38
rs768515615 Chr.7: 33177510-33177510 on Build GRCh38
rs918281836 Chr.7: 33177258-33177258 on Build GRCh38
rs895945972 Chr.7: 33177365-33177365 on Build GRCh38
rs372412756 Chr.7: 33177585-33177585 on Build GRCh38
rs797044763 Chr.7: 33177471-33177472 on Build GRCh38
rs764726613 Chr.7: 33177429-33177429 on Build GRCh38
rs899691671 Chr.7: 33177362-33177362 on Build GRCh38
rs771893428 Chr.7: 33177525-33177525 on Build GRCh38
rs908452923 Chr.7: 33177275-33177275 on Build GRCh38
rs763565455 Chr.7: 33177653-33177653 on Build GRCh38
rs1055993716 Chr.7: 33177360-33177360 on Build GRCh38
rs144340890 Chr.7: 33177582-33177582 on Build GRCh38
rs377615534 Chr.7: 33177717-33177717 on Build GRCh38
rs556340713 Chr.7: 33177520-33177520 on Build GRCh38
rs558819061 Chr.7: 33177386-33177386 on Build GRCh38
rs1000158795 Chr.7: 33177411-33177411 on Build GRCh38
rs759120113 Chr.7: 33177546-33177546 on Build GRCh38
rs982857414 Chr.7: 33177271-33177271 on Build GRCh38
rs1041854136 Chr.7: 33177632-33177637 on Build GRCh38
rs773396443 Chr.7: 33177652-33177652 on Build GRCh38
rs781476639 Chr.7: 33177474-33177474 on Build GRCh38
rs752108930 Chr.7: 33177559-33177559 on Build GRCh38
rs974374718 Chr.7: 33177715-33177715 on Build GRCh38
rs772915882 Chr.7: 33177530-33177530 on Build GRCh38
rs1006732457 Chr.7: 33177420-33177420 on Build GRCh38
rs183421903 Chr.7: 33177401-33177401 on Build GRCh38
rs144093303 Chr.7: 33177291-33177291 on Build GRCh38
rs762019329 Chr.7: 33177459-33177460 on Build GRCh38
rs547292414 Chr.7: 33177387-33177387 on Build GRCh38
rs904098347 Chr.7: 33177484-33177484 on Build GRCh38
rs549959412 Chr.7: 33177700-33177700 on Build GRCh38
rs774614867 Chr.7: 33177460-33177460 on Build GRCh38
rs375694520 Chr.7: 33177620-33177620 on Build GRCh38
rs76037216 Chr.7: 33177263-33177263 on Build GRCh38
rs191915870 Chr.7: 33177642-33177642 on Build GRCh38
rs976567513 Chr.7: 33177471-33177471 on Build GRCh38
rs755035606 Chr.7: 33177615-33177615 on Build GRCh38
rs1038191863 Chr.7: 33177351-33177351 on Build GRCh38
rs779028713 Chr.7: 33177616-33177616 on Build GRCh38
rs746132711 Chr.7: 33177349-33177349 on Build GRCh38
rs746809700 Chr.7: 33177634-33177634 on Build GRCh38
rs1001554781 Chr.7: 33177491-33177491 on Build GRCh38
rs757628818 Chr.7: 33177442-33177442 on Build GRCh38
rs778832586 Chr.7: 33177512-33177512 on Build GRCh38
rs766234410 Chr.7: 33177460-33177461 on Build GRCh38
rs753859931 Chr.7: 33177613-33177613 on Build GRCh38
rs762267565 Chr.7: 33177568-33177568 on Build GRCh38
rs1042177209 Chr.7: 33177390-33177393 on Build GRCh38
rs78283245 Chr.7: 33177539-33177539 on Build GRCh38
rs751916651 Chr.7: 33177436-33177436 on Build GRCh38
rs768926505 Chr.7: 33177414-33177414 on Build GRCh38
rs587777811 Chr.7: 33177592-33177592 on Build GRCh38
rs375243042 Chr.7: 33177548-33177548 on Build GRCh38
rs932492917 Chr.7: 33177372-33177372 on Build GRCh38
rs756343574 Chr.7: 33177493-33177493 on Build GRCh38
rs137852857 Chr.7: 33177570-33177570 on Build GRCh38
rs542324847 Chr.7: 33177544-33177544 on Build GRCh38
rs763503968 Chr.7: 33177427-33177427 on Build GRCh38
rs886062284 Chr.7: 33177534-33177534 on Build GRCh38
rs1023969420 Chr.7: 33177578-33177578 on Build GRCh38
rs568621158 Chr.7: 33177610-33177610 on Build GRCh38
rs750627869 Chr.7: 33177481-33177481 on Build GRCh38
rs188817948 Chr.7: 33177567-33177567 on Build GRCh38

View Full Product Details