Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 39706804-39707322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RALA
Entrez Gene ID
5898
Gene Name
RALA Ras like proto-oncogene A
Gene Aliases
RAL
UniGene
Hs.6906
Gene Chromosome Location
Chr.7: 39622955-39708124 on Build GRCh38
Transcripts
NM_005402.3 XM_006715762.2 XM_011515466.1

SNP Detail

SNP ID Chromosome Location
rs961184755 Chr.7: 39706906-39706906 on Build GRCh38
rs962404717 Chr.7: 39707068-39707068 on Build GRCh38
rs777070201 Chr.7: 39706894-39706897 on Build GRCh38
rs973699001 Chr.7: 39707095-39707095 on Build GRCh38
rs11555198 Chr.7: 39706899-39706899 on Build GRCh38
rs751155100 Chr.7: 39706988-39706988 on Build GRCh38
rs768619430 Chr.7: 39706868-39706868 on Build GRCh38
rs929687371 Chr.7: 39707168-39707168 on Build GRCh38
rs991075564 Chr.7: 39707035-39707035 on Build GRCh38
rs367963792 Chr.7: 39706907-39706907 on Build GRCh38
rs912382806 Chr.7: 39707296-39707296 on Build GRCh38
rs567438331 Chr.7: 39707063-39707063 on Build GRCh38
rs761525420 Chr.7: 39707117-39707117 on Build GRCh38
rs4723892 Chr.7: 39706867-39706867 on Build GRCh38
rs190755590 Chr.7: 39707212-39707212 on Build GRCh38
rs762751747 Chr.7: 39706848-39706848 on Build GRCh38
rs761370931 Chr.7: 39707175-39707175 on Build GRCh38
rs918269191 Chr.7: 39707100-39707100 on Build GRCh38
rs767305220 Chr.7: 39707225-39707225 on Build GRCh38
rs577695275 Chr.7: 39707064-39707064 on Build GRCh38
rs773845335 Chr.7: 39706989-39706989 on Build GRCh38
rs902574647 Chr.7: 39706851-39706851 on Build GRCh38
rs879511861 Chr.7: 39707045-39707045 on Build GRCh38
rs550529127 Chr.7: 39706910-39706910 on Build GRCh38
rs571666307 Chr.7: 39706832-39706832 on Build GRCh38
rs1024342596 Chr.7: 39707059-39707059 on Build GRCh38

View Full Product Details