Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 41964335-41964838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLI3
Entrez Gene ID
2737
Gene Name
GLI family zinc finger 3
Gene Aliases
ACLS, GCPS, GLI3-190, GLI3FL, PAP-A, PAPA, PAPA1, PAPB, PHS, PPDIV
UniGene
Hs.21509
Gene Chromosome Location
Chr.7: 41960949-42237019 on Build GRCh38
Transcripts
NM_000168.5 XM_011515274.2 XM_017011997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6040859 c.4291C>A p.P1431T Chr.7: 41964782-41964782 on Build GRCh38
4687140 c.4302T>C p.S1434S Chr.7: 41964771-41964771 on Build GRCh38
7082297 c.4483G>A p.D1495N Chr.7: 41964590-41964590 on Build GRCh38
3698390 c.4624C>T p.R1542W Chr.7: 41964449-41964449 on Build GRCh38
5450809 c.4665C>T p.T1555T Chr.7: 41964408-41964408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755885558 Chr.7: 41964615-41964615 on Build GRCh38
rs79625212 Chr.7: 41964513-41964513 on Build GRCh38
rs1025821723 Chr.7: 41964506-41964506 on Build GRCh38
rs769269390 Chr.7: 41964796-41964796 on Build GRCh38
rs780314759 Chr.7: 41964425-41964425 on Build GRCh38
rs745631926 Chr.7: 41964590-41964590 on Build GRCh38
rs373789617 Chr.7: 41964433-41964433 on Build GRCh38
rs372691047 Chr.7: 41964548-41964548 on Build GRCh38
rs767338817 Chr.7: 41964700-41964700 on Build GRCh38
rs986970840 Chr.7: 41964788-41964788 on Build GRCh38
rs746798261 Chr.7: 41964463-41964463 on Build GRCh38
rs764701766 Chr.7: 41964412-41964412 on Build GRCh38
rs199887017 Chr.7: 41964703-41964703 on Build GRCh38
rs762445898 Chr.7: 41964475-41964475 on Build GRCh38
rs145419251 Chr.7: 41964563-41964563 on Build GRCh38
rs560716408 Chr.7: 41964537-41964537 on Build GRCh38
rs377535957 Chr.7: 41964717-41964717 on Build GRCh38
rs149817170 Chr.7: 41964369-41964369 on Build GRCh38
rs745490483 Chr.7: 41964517-41964517 on Build GRCh38
rs143247740 Chr.7: 41964385-41964385 on Build GRCh38
rs776751129 Chr.7: 41964545-41964545 on Build GRCh38
rs746243175 Chr.7: 41964765-41964765 on Build GRCh38
rs201112421 Chr.7: 41964549-41964549 on Build GRCh38
rs763199119 Chr.7: 41964774-41964774 on Build GRCh38
rs760962930 Chr.7: 41964539-41964539 on Build GRCh38
rs537550919 Chr.7: 41964792-41964792 on Build GRCh38
rs767497727 Chr.7: 41964789-41964789 on Build GRCh38
rs778838449 Chr.7: 41964450-41964450 on Build GRCh38
rs776279429 Chr.7: 41964465-41964465 on Build GRCh38
rs755909964 Chr.7: 41964708-41964708 on Build GRCh38
rs890797962 Chr.7: 41964441-41964441 on Build GRCh38
rs770587326 Chr.7: 41964395-41964395 on Build GRCh38
rs775186351 Chr.7: 41964521-41964521 on Build GRCh38
rs754037065 Chr.7: 41964499-41964499 on Build GRCh38
rs560064684 Chr.7: 41964414-41964414 on Build GRCh38
rs140436882 Chr.7: 41964420-41964420 on Build GRCh38
rs750815703 Chr.7: 41964493-41964493 on Build GRCh38
rs775026941 Chr.7: 41964474-41964474 on Build GRCh38
rs781662679 Chr.7: 41964757-41964757 on Build GRCh38
rs566409623 Chr.7: 41964681-41964681 on Build GRCh38
rs149543947 Chr.7: 41964691-41964691 on Build GRCh38
rs886042309 Chr.7: 41964724-41964724 on Build GRCh38
rs752743828 Chr.7: 41964505-41964505 on Build GRCh38
rs775718988 Chr.7: 41964772-41964772 on Build GRCh38
rs367855810 Chr.7: 41964477-41964477 on Build GRCh38
rs748031460 Chr.7: 41964454-41964454 on Build GRCh38
rs755074584 Chr.7: 41964501-41964501 on Build GRCh38
rs765262389 Chr.7: 41964502-41964502 on Build GRCh38
rs770095761 Chr.7: 41964767-41964767 on Build GRCh38
rs372000475 Chr.7: 41964689-41964689 on Build GRCh38
rs769537011 Chr.7: 41964519-41964519 on Build GRCh38
rs759881277 Chr.7: 41964660-41964660 on Build GRCh38
rs748815553 Chr.7: 41964629-41964629 on Build GRCh38
rs755022753 Chr.7: 41964449-41964449 on Build GRCh38
rs138100963 Chr.7: 41964646-41964646 on Build GRCh38
rs777586456 Chr.7: 41964461-41964461 on Build GRCh38
rs374978412 Chr.7: 41964721-41964721 on Build GRCh38
rs761856178 Chr.7: 41964693-41964693 on Build GRCh38
rs771886604 Chr.7: 41964458-41964458 on Build GRCh38
rs1003879310 Chr.7: 41964421-41964421 on Build GRCh38
rs1057519504 Chr.7: 41964637-41964637 on Build GRCh38
rs371464477 Chr.7: 41964688-41964688 on Build GRCh38
rs750332096 Chr.7: 41964790-41964790 on Build GRCh38
rs577522844 Chr.7: 41964363-41964363 on Build GRCh38
rs761099870 Chr.7: 41964497-41964497 on Build GRCh38
rs747590659 Chr.7: 41964742-41964742 on Build GRCh38
rs555888138 Chr.7: 41964361-41964361 on Build GRCh38
rs41305933 Chr.7: 41964364-41964364 on Build GRCh38
rs758379697 Chr.7: 41964509-41964509 on Build GRCh38
rs923793927 Chr.7: 41964664-41964664 on Build GRCh38
rs756335435 Chr.7: 41964424-41964424 on Build GRCh38
rs907289973 Chr.7: 41964618-41964618 on Build GRCh38
rs912292896 Chr.7: 41964755-41964755 on Build GRCh38
rs777156738 Chr.7: 41964560-41964560 on Build GRCh38
rs1034910492 Chr.7: 41964404-41964404 on Build GRCh38
rs770007515 Chr.7: 41964673-41964673 on Build GRCh38
rs748790609 Chr.7: 41964526-41964526 on Build GRCh38
rs377186629 Chr.7: 41964555-41964555 on Build GRCh38
rs2079451 Chr.7: 41964478-41964478 on Build GRCh38
rs145513625 Chr.7: 41964540-41964540 on Build GRCh38
rs758980438 Chr.7: 41964466-41964466 on Build GRCh38
rs111446996 Chr.7: 41964426-41964426 on Build GRCh38
rs886062335 Chr.7: 41964483-41964483 on Build GRCh38
rs975131125 Chr.7: 41964444-41964444 on Build GRCh38
rs138272666 Chr.7: 41964468-41964468 on Build GRCh38
rs141759179 Chr.7: 41964552-41964552 on Build GRCh38
rs905011458 Chr.7: 41964375-41964375 on Build GRCh38
rs1035767184 Chr.7: 41964553-41964553 on Build GRCh38
rs775631287 Chr.7: 41964676-41964676 on Build GRCh38
rs760205624 Chr.7: 41964393-41964393 on Build GRCh38
rs75313224 Chr.7: 41964467-41964467 on Build GRCh38
rs753656456 Chr.7: 41964815-41964815 on Build GRCh38
rs776061573 Chr.7: 41964406-41964406 on Build GRCh38
rs779851650 Chr.7: 41964628-41964628 on Build GRCh38
rs370009188 Chr.7: 41964653-41964653 on Build GRCh38
rs151254859 Chr.7: 41964419-41964419 on Build GRCh38
rs763951987 Chr.7: 41964554-41964554 on Build GRCh38
rs35765130 Chr.7: 41964368-41964368 on Build GRCh38
rs771312835 Chr.7: 41964649-41964649 on Build GRCh38
rs747943668 Chr.7: 41964370-41964370 on Build GRCh38
rs759082153 Chr.7: 41964409-41964409 on Build GRCh38
rs772874812 Chr.7: 41964386-41964386 on Build GRCh38
rs956536432 Chr.7: 41964732-41964732 on Build GRCh38
rs532584303 Chr.7: 41964586-41964586 on Build GRCh38
rs778526954 Chr.7: 41964731-41964731 on Build GRCh38
rs1016873543 Chr.7: 41964709-41964709 on Build GRCh38
rs771527522 Chr.7: 41964751-41964751 on Build GRCh38
rs753494256 Chr.7: 41964720-41964720 on Build GRCh38
rs768735366 Chr.7: 41964777-41964777 on Build GRCh38
rs28396689 Chr.7: 41964780-41964780 on Build GRCh38
rs144128064 Chr.7: 41964781-41964781 on Build GRCh38
rs898743387 Chr.7: 41964495-41964495 on Build GRCh38
rs41311769 Chr.7: 41964470-41964470 on Build GRCh38
rs780729219 Chr.7: 41964516-41964516 on Build GRCh38
rs773617369 Chr.7: 41964645-41964645 on Build GRCh38
rs759795415 Chr.7: 41964546-41964546 on Build GRCh38
rs772777499 Chr.7: 41964535-41964535 on Build GRCh38
rs768158142 Chr.7: 41964630-41964630 on Build GRCh38

View Full Product Details