Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44140141-44140679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYL7
Entrez Gene ID
58498
Gene Name
myosin light chain 7
Gene Aliases
MYL2A, MYLC2A
UniGene
Hs.75636
Gene Chromosome Location
Chr.7: 44138864-44141392 on Build GRCh38
Transcripts
NM_021223.2 XM_005249817.4 XM_011515463.2 XM_011515464.2 XM_011515465.2 XM_017012478.1 XM_017012479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1330612 c.270C>A p.F90L Chr.7: 44140351-44140351 on Build GRCh38
5011744 c.217G>A p.E73K Chr.7: 44140404-44140404 on Build GRCh38
3638883 c.270C>T p.F90F Chr.7: 44140351-44140351 on Build GRCh38
485361 c.270C>G p.F90L Chr.7: 44140351-44140351 on Build GRCh38
1089917 c.246G>T p.K82N Chr.7: 44140375-44140375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755314842 Chr.7: 44140489-44140489 on Build GRCh38
rs1053471513 Chr.7: 44140179-44140179 on Build GRCh38
rs546109215 Chr.7: 44140256-44140256 on Build GRCh38
rs761808767 Chr.7: 44140380-44140380 on Build GRCh38
rs773977622 Chr.7: 44140336-44140336 on Build GRCh38
rs889459368 Chr.7: 44140568-44140568 on Build GRCh38
rs746637258 Chr.7: 44140533-44140533 on Build GRCh38
rs200873384 Chr.7: 44140291-44140291 on Build GRCh38
rs997399513 Chr.7: 44140576-44140576 on Build GRCh38
rs1046276538 Chr.7: 44140438-44140438 on Build GRCh38
rs993489030 Chr.7: 44140408-44140408 on Build GRCh38
rs577969475 Chr.7: 44140413-44140413 on Build GRCh38
rs950252145 Chr.7: 44140411-44140411 on Build GRCh38
rs993949734 Chr.7: 44140601-44140601 on Build GRCh38
rs748745826 Chr.7: 44140322-44140322 on Build GRCh38
rs913656346 Chr.7: 44140272-44140272 on Build GRCh38
rs562583838 Chr.7: 44140401-44140401 on Build GRCh38
rs924803628 Chr.7: 44140634-44140634 on Build GRCh38
rs55908146 Chr.7: 44140627-44140627 on Build GRCh38
rs959081100 Chr.7: 44140623-44140623 on Build GRCh38
rs974378405 Chr.7: 44140194-44140204 on Build GRCh38
rs771599510 Chr.7: 44140521-44140521 on Build GRCh38
rs150940284 Chr.7: 44140395-44140395 on Build GRCh38
rs755832896 Chr.7: 44140370-44140370 on Build GRCh38
rs945125828 Chr.7: 44140289-44140289 on Build GRCh38
rs748660037 Chr.7: 44140437-44140437 on Build GRCh38
rs562192334 Chr.7: 44140545-44140545 on Build GRCh38
rs566096262 Chr.7: 44140641-44140641 on Build GRCh38
rs775224242 Chr.7: 44140447-44140447 on Build GRCh38
rs62460073 Chr.7: 44140279-44140279 on Build GRCh38
rs750433248 Chr.7: 44140473-44140473 on Build GRCh38
rs368416439 Chr.7: 44140498-44140498 on Build GRCh38
rs923665849 Chr.7: 44140495-44140495 on Build GRCh38
rs769242692 Chr.7: 44140315-44140315 on Build GRCh38
rs781726299 Chr.7: 44140422-44140422 on Build GRCh38
rs760377990 Chr.7: 44140449-44140449 on Build GRCh38
rs185765373 Chr.7: 44140177-44140177 on Build GRCh38
rs753763649 Chr.7: 44140373-44140373 on Build GRCh38
rs775108527 Chr.7: 44140350-44140350 on Build GRCh38
rs201923002 Chr.7: 44140320-44140320 on Build GRCh38
rs768437070 Chr.7: 44140453-44140453 on Build GRCh38
rs1053422376 Chr.7: 44140553-44140553 on Build GRCh38
rs1025456463 Chr.7: 44140618-44140618 on Build GRCh38
rs757056514 Chr.7: 44140248-44140248 on Build GRCh38
rs113051531 Chr.7: 44140429-44140429 on Build GRCh38
rs546714202 Chr.7: 44140544-44140544 on Build GRCh38
rs573687519 Chr.7: 44140388-44140388 on Build GRCh38
rs889689028 Chr.7: 44140217-44140217 on Build GRCh38
rs966316656 Chr.7: 44140566-44140566 on Build GRCh38
rs766451902 Chr.7: 44140484-44140484 on Build GRCh38
rs751815052 Chr.7: 44140487-44140487 on Build GRCh38
rs1019411708 Chr.7: 44140264-44140264 on Build GRCh38
rs757342607 Chr.7: 44140283-44140283 on Build GRCh38
rs148474161 Chr.7: 44140325-44140325 on Build GRCh38
rs776454713 Chr.7: 44140454-44140454 on Build GRCh38
rs768349318 Chr.7: 44140538-44140538 on Build GRCh38
rs398004624 Chr.7: 44140482-44140483 on Build GRCh38
rs760469703 Chr.7: 44140356-44140356 on Build GRCh38
rs781661363 Chr.7: 44140490-44140490 on Build GRCh38
rs765245085 Chr.7: 44140463-44140463 on Build GRCh38
rs747426370 Chr.7: 44140310-44140310 on Build GRCh38
rs763962748 Chr.7: 44140357-44140357 on Build GRCh38
rs142668114 Chr.7: 44140346-44140346 on Build GRCh38
rs199928989 Chr.7: 44140391-44140391 on Build GRCh38
rs745311942 Chr.7: 44140517-44140517 on Build GRCh38
rs551142776 Chr.7: 44140578-44140578 on Build GRCh38
rs775697077 Chr.7: 44140406-44140406 on Build GRCh38
rs5883888 Chr.7: 44140480-44140481 on Build GRCh38
rs753849950 Chr.7: 44140282-44140282 on Build GRCh38
rs769547866 Chr.7: 44140549-44140549 on Build GRCh38
rs75267165 Chr.7: 44140445-44140445 on Build GRCh38
rs778349184 Chr.7: 44140510-44140510 on Build GRCh38
rs751131902 Chr.7: 44140316-44140316 on Build GRCh38
rs780263101 Chr.7: 44140298-44140298 on Build GRCh38
rs767246348 Chr.7: 44140348-44140348 on Build GRCh38
rs1007945125 Chr.7: 44140525-44140525 on Build GRCh38
rs1017878193 Chr.7: 44140184-44140184 on Build GRCh38
rs529411463 Chr.7: 44140569-44140569 on Build GRCh38
rs564448793 Chr.7: 44140491-44140491 on Build GRCh38
rs869104936 Chr.7: 44140481-44140482 on Build GRCh38
rs916165134 Chr.7: 44140651-44140651 on Build GRCh38
rs533528999 Chr.7: 44140655-44140655 on Build GRCh38
rs190422753 Chr.7: 44140600-44140600 on Build GRCh38
rs779607333 Chr.7: 44140530-44140530 on Build GRCh38
rs741033 Chr.7: 44140166-44140166 on Build GRCh38
rs976286872 Chr.7: 44140254-44140254 on Build GRCh38
rs763096097 Chr.7: 44140474-44140474 on Build GRCh38
rs73691404 Chr.7: 44140621-44140621 on Build GRCh38
rs202150864 Chr.7: 44140462-44140462 on Build GRCh38
rs758690673 Chr.7: 44140296-44140296 on Build GRCh38
rs867398556 Chr.7: 44140435-44140435 on Build GRCh38
rs755402823 Chr.7: 44140418-44140418 on Build GRCh38
rs746077715 Chr.7: 44140293-44140293 on Build GRCh38
rs374534131 Chr.7: 44140396-44140396 on Build GRCh38
rs770372940 Chr.7: 44140440-44140440 on Build GRCh38
rs1041357411 Chr.7: 44140588-44140588 on Build GRCh38

View Full Product Details