Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44801565-44802106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPIA
Entrez Gene ID
5478
Gene Name
peptidylprolyl isomerase A
Gene Aliases
CYPA, CYPH, HEL-S-69p
UniGene
Hs.356331
Gene Chromosome Location
Chr.7: 44796636-44803123 on Build GRCh38
Transcripts
NM_001300981.1 NM_021130.4

SNP Detail

SNP ID Chromosome Location
rs776433516 Chr.7: 44801792-44801792 on Build GRCh38
rs890905522 Chr.7: 44801754-44801754 on Build GRCh38
rs764864527 Chr.7: 44802031-44802031 on Build GRCh38
rs907551542 Chr.7: 44801722-44801722 on Build GRCh38
rs1037618214 Chr.7: 44802000-44802000 on Build GRCh38
rs754803033 Chr.7: 44802023-44802023 on Build GRCh38
rs111735829 Chr.7: 44801784-44801784 on Build GRCh38
rs931498228 Chr.7: 44802025-44802025 on Build GRCh38
rs948252420 Chr.7: 44801625-44801625 on Build GRCh38
rs1037648992 Chr.7: 44801765-44801765 on Build GRCh38
rs765003104 Chr.7: 44802037-44802037 on Build GRCh38
rs980871301 Chr.7: 44801921-44801921 on Build GRCh38
rs912191920 Chr.7: 44802014-44802014 on Build GRCh38
rs985569558 Chr.7: 44802004-44802004 on Build GRCh38
rs1057172010 Chr.7: 44801714-44801714 on Build GRCh38
rs143350466 Chr.7: 44801908-44801909 on Build GRCh38
rs397704534 Chr.7: 44801740-44801741 on Build GRCh38
rs144790995 Chr.7: 44801822-44801822 on Build GRCh38
rs1027263268 Chr.7: 44801844-44801844 on Build GRCh38
rs397816218 Chr.7: 44801739-44801740 on Build GRCh38
rs890315706 Chr.7: 44802005-44802005 on Build GRCh38
rs533067408 Chr.7: 44801927-44801927 on Build GRCh38
rs927084957 Chr.7: 44801655-44801655 on Build GRCh38
rs890504644 Chr.7: 44801815-44801815 on Build GRCh38
rs895785244 Chr.7: 44802074-44802074 on Build GRCh38
rs960033981 Chr.7: 44801967-44801967 on Build GRCh38
rs1031398871 Chr.7: 44801851-44801851 on Build GRCh38
rs945300886 Chr.7: 44801620-44801620 on Build GRCh38
rs969525332 Chr.7: 44801921-44801925 on Build GRCh38
rs897287659 Chr.7: 44801813-44801813 on Build GRCh38
rs1001568402 Chr.7: 44801879-44801879 on Build GRCh38
rs988875659 Chr.7: 44801900-44801900 on Build GRCh38
rs17860087 Chr.7: 44801732-44801733 on Build GRCh38
rs928149740 Chr.7: 44801949-44801949 on Build GRCh38
rs566320533 Chr.7: 44801637-44801637 on Build GRCh38
rs139105425 Chr.7: 44801816-44801816 on Build GRCh38
rs940583222 Chr.7: 44801723-44801723 on Build GRCh38
rs1043597514 Chr.7: 44801628-44801628 on Build GRCh38
rs11984372 Chr.7: 44802047-44802047 on Build GRCh38
rs1050256694 Chr.7: 44801787-44801787 on Build GRCh38
rs938452146 Chr.7: 44801684-44801684 on Build GRCh38
rs17860088 Chr.7: 44801969-44801969 on Build GRCh38
rs1010901698 Chr.7: 44802044-44802044 on Build GRCh38
rs1049373004 Chr.7: 44801664-44801664 on Build GRCh38
rs17860090 Chr.7: 44802003-44802005 on Build GRCh38
rs541358110 Chr.7: 44801991-44801991 on Build GRCh38
rs8177829 Chr.7: 44801594-44801596 on Build GRCh38
rs574645132 Chr.7: 44802056-44802056 on Build GRCh38
rs759041932 Chr.7: 44802003-44802003 on Build GRCh38
rs924969682 Chr.7: 44801631-44801631 on Build GRCh38
rs753476943 Chr.7: 44802020-44802020 on Build GRCh38
rs386544857 Chr.7: 44801649-44801650 on Build GRCh38
rs1035691758 Chr.7: 44801897-44801897 on Build GRCh38
rs1021218140 Chr.7: 44801957-44801957 on Build GRCh38
rs8177830 Chr.7: 44801650-44801651 on Build GRCh38
rs943793404 Chr.7: 44802017-44802017 on Build GRCh38
rs191837591 Chr.7: 44801758-44801758 on Build GRCh38
rs141838679 Chr.7: 44802061-44802061 on Build GRCh38
rs906100303 Chr.7: 44801863-44801863 on Build GRCh38
rs374908469 Chr.7: 44801806-44801806 on Build GRCh38
rs11547708 Chr.7: 44801890-44801890 on Build GRCh38
rs758334118 Chr.7: 44802071-44802071 on Build GRCh38
rs17860086 Chr.7: 44801647-44801648 on Build GRCh38
rs920469185 Chr.7: 44802001-44802001 on Build GRCh38
rs960400112 Chr.7: 44801898-44801898 on Build GRCh38
rs1037609335 Chr.7: 44801802-44801802 on Build GRCh38
rs898988309 Chr.7: 44801773-44801773 on Build GRCh38
rs17860089 Chr.7: 44801973-44801973 on Build GRCh38
rs1011101257 Chr.7: 44801903-44801903 on Build GRCh38

View Full Product Details