Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 50443884-50444382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIGNL1
Entrez Gene ID
63979
Gene Name
fidgetin like 1
Gene Aliases
-
UniGene
Hs.137516
Gene Chromosome Location
Chr.7: 50437327-50450390 on Build GRCh38
Transcripts
NM_001042762.2 NM_001287492.1 NM_001287493.1 NM_001287494.1 NM_001287495.1 NM_001287496.1 NM_022116.4 XM_011515470.2 XM_017012501.1 XM_017012502.1

SNP Detail

SNP ID Chromosome Location
rs889216033 Chr.7: 50444013-50444013 on Build GRCh38
rs774849993 Chr.7: 50444296-50444296 on Build GRCh38
rs903670728 Chr.7: 50443939-50443939 on Build GRCh38
rs960264362 Chr.7: 50444180-50444180 on Build GRCh38
rs202070241 Chr.7: 50443945-50443945 on Build GRCh38
rs758293717 Chr.7: 50444205-50444205 on Build GRCh38
rs541490477 Chr.7: 50443974-50443974 on Build GRCh38
rs114989501 Chr.7: 50444276-50444276 on Build GRCh38
rs111262901 Chr.7: 50444241-50444241 on Build GRCh38
rs1045195851 Chr.7: 50444221-50444221 on Build GRCh38
rs182481621 Chr.7: 50444264-50444264 on Build GRCh38
rs949228041 Chr.7: 50444316-50444316 on Build GRCh38
rs965558562 Chr.7: 50444261-50444261 on Build GRCh38
rs200159545 Chr.7: 50444295-50444295 on Build GRCh38
rs187171531 Chr.7: 50444299-50444299 on Build GRCh38
rs979509068 Chr.7: 50444278-50444278 on Build GRCh38
rs3178610 Chr.7: 50444142-50444142 on Build GRCh38
rs947807167 Chr.7: 50443940-50443940 on Build GRCh38
rs756908145 Chr.7: 50444267-50444267 on Build GRCh38
rs899292411 Chr.7: 50444112-50444112 on Build GRCh38
rs1048617147 Chr.7: 50444232-50444232 on Build GRCh38
rs57651203 Chr.7: 50444218-50444218 on Build GRCh38
rs3173667 Chr.7: 50444355-50444355 on Build GRCh38
rs546895549 Chr.7: 50444223-50444223 on Build GRCh38
rs936590633 Chr.7: 50443957-50443957 on Build GRCh38
rs1016851918 Chr.7: 50444095-50444095 on Build GRCh38
rs979343489 Chr.7: 50444179-50444179 on Build GRCh38
rs925208598 Chr.7: 50444184-50444184 on Build GRCh38
rs905231058 Chr.7: 50444004-50444004 on Build GRCh38
rs769117434 Chr.7: 50444294-50444294 on Build GRCh38
rs766419363 Chr.7: 50443965-50443965 on Build GRCh38
rs921602447 Chr.7: 50444082-50444082 on Build GRCh38
rs1027679204 Chr.7: 50444029-50444029 on Build GRCh38
rs935138034 Chr.7: 50444189-50444189 on Build GRCh38
rs995753941 Chr.7: 50444015-50444015 on Build GRCh38
rs13228445 Chr.7: 50444247-50444247 on Build GRCh38
rs185748601 Chr.7: 50444132-50444132 on Build GRCh38
rs931911243 Chr.7: 50443937-50443937 on Build GRCh38
rs951089205 Chr.7: 50443927-50443927 on Build GRCh38
rs1023632246 Chr.7: 50443995-50443995 on Build GRCh38
rs565900684 Chr.7: 50444230-50444230 on Build GRCh38
rs1007610964 Chr.7: 50444028-50444028 on Build GRCh38
rs896571995 Chr.7: 50443975-50443975 on Build GRCh38
rs1026557367 Chr.7: 50444146-50444146 on Build GRCh38
rs13310009 Chr.7: 50444175-50444175 on Build GRCh38
rs969552387 Chr.7: 50444250-50444250 on Build GRCh38
rs550350215 Chr.7: 50444045-50444045 on Build GRCh38
rs577396126 Chr.7: 50444298-50444298 on Build GRCh38
rs925109008 Chr.7: 50443953-50443953 on Build GRCh38
rs77803747 Chr.7: 50444270-50444270 on Build GRCh38
rs148525056 Chr.7: 50443938-50443938 on Build GRCh38
rs1045146225 Chr.7: 50444325-50444325 on Build GRCh38
rs748616060 Chr.7: 50444317-50444317 on Build GRCh38
rs754942137 Chr.7: 50444055-50444055 on Build GRCh38
rs78898856 Chr.7: 50443979-50443979 on Build GRCh38
rs765274119 Chr.7: 50444068-50444068 on Build GRCh38
rs1004305862 Chr.7: 50444248-50444248 on Build GRCh38
rs935173038 Chr.7: 50443941-50443941 on Build GRCh38
rs1014587731 Chr.7: 50444065-50444065 on Build GRCh38
rs975841207 Chr.7: 50444070-50444070 on Build GRCh38
rs530564246 Chr.7: 50444039-50444039 on Build GRCh38
rs917737208 Chr.7: 50444314-50444314 on Build GRCh38
rs17152012 Chr.7: 50444254-50444254 on Build GRCh38
rs564340445 Chr.7: 50444210-50444210 on Build GRCh38
rs1052273273 Chr.7: 50444198-50444198 on Build GRCh38
rs568566917 Chr.7: 50444265-50444265 on Build GRCh38

View Full Product Details