Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66795421-66795856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABGEF1
Entrez Gene ID
27342
Gene Name
RAB guanine nucleotide exchange factor 1
Gene Aliases
RABEX5, RAP1, rabex-5
UniGene
Hs.530053
Gene Chromosome Location
Chr.7: 66682091-66811464 on Build GRCh38
Transcripts
NM_001287060.1 NM_001287061.1 NM_001287062.1 NM_014504.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5062546 c.571G>A p.E191K Chr.7: 66795526-66795526 on Build GRCh38
461561 c.538G>C p.E180Q Chr.7: 66795535-66795535 on Build GRCh38
6178372 c.538G>T p.E180* Chr.7: 66795535-66795535 on Build GRCh38
5816428 c.568G>C p.E190Q Chr.7: 66795523-66795523 on Build GRCh38
5537961 c.586C>T p.R196C Chr.7: 66795583-66795583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113807169 Chr.7: 66795677-66795677 on Build GRCh38
rs781399666 Chr.7: 66795577-66795577 on Build GRCh38
rs747186146 Chr.7: 66795573-66795573 on Build GRCh38
rs541014660 Chr.7: 66795567-66795567 on Build GRCh38
rs753448538 Chr.7: 66795625-66795625 on Build GRCh38
rs765187274 Chr.7: 66795504-66795504 on Build GRCh38
rs976204600 Chr.7: 66795452-66795452 on Build GRCh38
rs572729291 Chr.7: 66795492-66795492 on Build GRCh38
rs773784585 Chr.7: 66795595-66795595 on Build GRCh38
rs750323189 Chr.7: 66795634-66795634 on Build GRCh38
rs757288814 Chr.7: 66795538-66795538 on Build GRCh38
rs752228202 Chr.7: 66795617-66795617 on Build GRCh38
rs779438863 Chr.7: 66795617-66795618 on Build GRCh38
rs768742841 Chr.7: 66795462-66795462 on Build GRCh38
rs780286779 Chr.7: 66795570-66795570 on Build GRCh38
rs763713060 Chr.7: 66795514-66795514 on Build GRCh38
rs755705009 Chr.7: 66795512-66795512 on Build GRCh38
rs748160894 Chr.7: 66795584-66795584 on Build GRCh38
rs529067463 Chr.7: 66795769-66795769 on Build GRCh38
rs774830869 Chr.7: 66795619-66795619 on Build GRCh38
rs768728920 Chr.7: 66795574-66795574 on Build GRCh38
rs773769828 Chr.7: 66795485-66795485 on Build GRCh38
rs563408414 Chr.7: 66795761-66795761 on Build GRCh38
rs564037503 Chr.7: 66795622-66795622 on Build GRCh38
rs765656729 Chr.7: 66795484-66795484 on Build GRCh38
rs1056227211 Chr.7: 66795674-66795674 on Build GRCh38
rs1045292958 Chr.7: 66795751-66795751 on Build GRCh38
rs138355023 Chr.7: 66795473-66795473 on Build GRCh38
rs143374711 Chr.7: 66795566-66795566 on Build GRCh38
rs778706191 Chr.7: 66795544-66795544 on Build GRCh38
rs897837285 Chr.7: 66795784-66795784 on Build GRCh38
rs1014765262 Chr.7: 66795768-66795768 on Build GRCh38
rs762421989 Chr.7: 66795476-66795476 on Build GRCh38
rs776550837 Chr.7: 66795472-66795472 on Build GRCh38
rs758211772 Chr.7: 66795637-66795637 on Build GRCh38
rs1030453193 Chr.7: 66795826-66795826 on Build GRCh38
rs372795506 Chr.7: 66795601-66795601 on Build GRCh38
rs896185462 Chr.7: 66795709-66795709 on Build GRCh38
rs758125540 Chr.7: 66795568-66795568 on Build GRCh38
rs375319188 Chr.7: 66795769-66795770 on Build GRCh38
rs750480712 Chr.7: 66795533-66795533 on Build GRCh38
rs767280467 Chr.7: 66795503-66795503 on Build GRCh38
rs753282089 Chr.7: 66795737-66795737 on Build GRCh38
rs760419160 Chr.7: 66795620-66795620 on Build GRCh38
rs189892912 Chr.7: 66795820-66795820 on Build GRCh38
rs934957103 Chr.7: 66795664-66795664 on Build GRCh38
rs549206762 Chr.7: 66795788-66795788 on Build GRCh38
rs765226604 Chr.7: 66795631-66795631 on Build GRCh38
rs142831425 Chr.7: 66795758-66795758 on Build GRCh38
rs753260239 Chr.7: 66795516-66795516 on Build GRCh38
rs770346275 Chr.7: 66795586-66795586 on Build GRCh38
rs781748659 Chr.7: 66795600-66795600 on Build GRCh38
rs779863337 Chr.7: 66795640-66795640 on Build GRCh38
rs923622013 Chr.7: 66795649-66795649 on Build GRCh38
rs532980473 Chr.7: 66795713-66795713 on Build GRCh38
rs914860866 Chr.7: 66795605-66795605 on Build GRCh38
rs997885850 Chr.7: 66795786-66795786 on Build GRCh38

View Full Product Details