Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73331684-73332193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP6
Entrez Gene ID
8468
Gene Name
FK506 binding protein 6
Gene Aliases
FKBP36
UniGene
Hs.661266
Gene Chromosome Location
Chr.7: 73328152-73358637 on Build GRCh38
Transcripts
NM_001135211.2 NM_001281304.1 NM_003602.4 XM_006716153.2 XM_017012741.1 XM_017012742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6664286 c.504G>A p.T168T Chr.7: 73331707-73331707 on Build GRCh38
485532 c.556C>T p.R186C Chr.7: 73331744-73331744 on Build GRCh38
302244 c.548G>A p.R183H Chr.7: 73331736-73331736 on Build GRCh38
6664293 c.546C>T p.F182F Chr.7: 73331734-73331734 on Build GRCh38
6053705 c.554C>T p.A185V Chr.7: 73331757-73331757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202120437 Chr.7: 73331724-73331724 on Build GRCh38
rs143664466 Chr.7: 73331707-73331707 on Build GRCh38
rs782441232 Chr.7: 73331821-73331821 on Build GRCh38
rs376031664 Chr.7: 73331736-73331736 on Build GRCh38
rs981858730 Chr.7: 73332025-73332025 on Build GRCh38
rs782013821 Chr.7: 73331750-73331750 on Build GRCh38
rs530088388 Chr.7: 73332005-73332005 on Build GRCh38
rs781975885 Chr.7: 73331894-73331894 on Build GRCh38
rs928807182 Chr.7: 73332041-73332041 on Build GRCh38
rs782693683 Chr.7: 73331758-73331758 on Build GRCh38
rs975776720 Chr.7: 73331711-73331711 on Build GRCh38
rs782101184 Chr.7: 73331739-73331739 on Build GRCh38
rs781852574 Chr.7: 73331772-73331772 on Build GRCh38
rs201612134 Chr.7: 73331718-73331718 on Build GRCh38
rs564233247 Chr.7: 73331788-73331788 on Build GRCh38
rs1028675834 Chr.7: 73331936-73331936 on Build GRCh38
rs991733320 Chr.7: 73332099-73332099 on Build GRCh38
rs535072735 Chr.7: 73332155-73332155 on Build GRCh38
rs782495498 Chr.7: 73331710-73331710 on Build GRCh38
rs376304654 Chr.7: 73331794-73331794 on Build GRCh38
rs140580205 Chr.7: 73331835-73331835 on Build GRCh38
rs1056480846 Chr.7: 73331824-73331824 on Build GRCh38
rs369087706 Chr.7: 73331728-73331728 on Build GRCh38
rs782299797 Chr.7: 73331799-73331799 on Build GRCh38
rs781799671 Chr.7: 73331760-73331760 on Build GRCh38
rs1043134450 Chr.7: 73331869-73331869 on Build GRCh38
rs782438676 Chr.7: 73331801-73331802 on Build GRCh38
rs1015257743 Chr.7: 73331965-73331965 on Build GRCh38
rs782223577 Chr.7: 73331825-73331825 on Build GRCh38
rs996757015 Chr.7: 73331928-73331928 on Build GRCh38
rs546867226 Chr.7: 73331913-73331913 on Build GRCh38
rs568302116 Chr.7: 73331937-73331937 on Build GRCh38
rs782469675 Chr.7: 73331766-73331766 on Build GRCh38
rs781811349 Chr.7: 73331778-73331779 on Build GRCh38
rs1005183271 Chr.7: 73331964-73331964 on Build GRCh38
rs1043083125 Chr.7: 73332159-73332159 on Build GRCh38
rs368312496 Chr.7: 73331944-73331944 on Build GRCh38
rs376672400 Chr.7: 73331716-73331716 on Build GRCh38
rs961108980 Chr.7: 73332009-73332009 on Build GRCh38
rs191397941 Chr.7: 73332065-73332065 on Build GRCh38
rs782723182 Chr.7: 73331707-73331707 on Build GRCh38
rs916162278 Chr.7: 73332121-73332121 on Build GRCh38
rs570049656 Chr.7: 73332088-73332088 on Build GRCh38
rs782584992 Chr.7: 73331822-73331822 on Build GRCh38
rs199580691 Chr.7: 73331712-73331712 on Build GRCh38
rs372751157 Chr.7: 73331902-73331902 on Build GRCh38
rs952382721 Chr.7: 73331962-73331962 on Build GRCh38
rs61394056 Chr.7: 73331968-73331968 on Build GRCh38
rs782084742 Chr.7: 73331903-73331903 on Build GRCh38
rs998652752 Chr.7: 73331895-73331895 on Build GRCh38
rs782737367 Chr.7: 73331771-73331771 on Build GRCh38
rs894458295 Chr.7: 73331826-73331826 on Build GRCh38
rs202156840 Chr.7: 73331745-73331745 on Build GRCh38
rs373152212 Chr.7: 73331751-73331751 on Build GRCh38
rs36080191 Chr.7: 73331733-73331733 on Build GRCh38
rs372290758 Chr.7: 73332045-73332045 on Build GRCh38

View Full Product Details