Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99453878-99454382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5J2-PTCD1
Entrez Gene ID
100526740
Gene Name
ATP5J2-PTCD1 readthrough
Gene Aliases
-
UniGene
Hs.632313
Gene Chromosome Location
Chr.7: 99416739-99466201 on Build GRCh38
Transcripts
NM_001198879.1 NM_001081559.2 NM_001318160.1 NM_001318161.1 NM_001318162.1 NM_006693.3 XM_011515755.2 XM_011515756.2 XM_011515757.2 XM_017011700.1 XM_017011701.1 XM_017011702.1 XM_017011703.1

Gene Symbol
CPSF4
Entrez Gene ID
10898
Gene Name
cleavage and polyadenylation specific factor 4
Gene Aliases
CPSF30, NAR, NEB-1, NEB1
UniGene
Hs.489287
Gene Chromosome Location
Chr.7: 99438920-99457377 on Build GRCh38
Transcripts
NM_001198879.1 NM_001081559.2 NM_001318160.1 NM_001318161.1 NM_001318162.1 NM_006693.3 XM_011515755.2 XM_011515756.2 XM_011515757.2 XM_017011700.1 XM_017011701.1 XM_017011702.1 XM_017011703.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634888 c.584C>T p.P195L Chr.7: 99453979-99453979 on Build GRCh38
5498974 c.656C>T p.P219L Chr.7: 99454051-99454051 on Build GRCh38
6634887 c.582T>C p.N194N Chr.7: 99453977-99453977 on Build GRCh38
6634889 c.657G>A p.P219P Chr.7: 99454052-99454052 on Build GRCh38
2867174 c.652A>G p.T218A Chr.7: 99454047-99454047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769586215 Chr.7: 99453952-99453952 on Build GRCh38
rs575051794 Chr.7: 99454253-99454253 on Build GRCh38
rs756069354 Chr.7: 99454016-99454016 on Build GRCh38
rs753862977 Chr.7: 99454009-99454009 on Build GRCh38
rs759444775 Chr.7: 99454110-99454110 on Build GRCh38
rs746204081 Chr.7: 99453932-99453932 on Build GRCh38
rs753357614 Chr.7: 99454002-99454002 on Build GRCh38
rs777720194 Chr.7: 99453919-99453919 on Build GRCh38
rs757693651 Chr.7: 99453924-99453924 on Build GRCh38
rs780407410 Chr.7: 99454043-99454043 on Build GRCh38
rs756179405 Chr.7: 99453918-99453918 on Build GRCh38
rs1030787771 Chr.7: 99454240-99454240 on Build GRCh38
rs779544824 Chr.7: 99453925-99453925 on Build GRCh38
rs755419410 Chr.7: 99454162-99454162 on Build GRCh38
rs878987421 Chr.7: 99453908-99453908 on Build GRCh38
rs368294752 Chr.7: 99454144-99454144 on Build GRCh38
rs752991967 Chr.7: 99453916-99453916 on Build GRCh38
rs540872785 Chr.7: 99454042-99454042 on Build GRCh38
rs547256967 Chr.7: 99454281-99454281 on Build GRCh38
rs747462840 Chr.7: 99454049-99454049 on Build GRCh38
rs956092169 Chr.7: 99454153-99454153 on Build GRCh38
rs1056180524 Chr.7: 99454127-99454127 on Build GRCh38
rs1026821132 Chr.7: 99454360-99454360 on Build GRCh38
rs146864927 Chr.7: 99453980-99453980 on Build GRCh38
rs374595432 Chr.7: 99454089-99454089 on Build GRCh38
rs766344289 Chr.7: 99453953-99453954 on Build GRCh38
rs1007919714 Chr.7: 99454310-99454310 on Build GRCh38
rs374374315 Chr.7: 99453936-99453936 on Build GRCh38
rs77770698 Chr.7: 99454185-99454185 on Build GRCh38
rs374544029 Chr.7: 99454180-99454180 on Build GRCh38
rs538010930 Chr.7: 99454102-99454102 on Build GRCh38
rs750869911 Chr.7: 99454151-99454151 on Build GRCh38
rs576966048 Chr.7: 99453994-99453994 on Build GRCh38
rs372398619 Chr.7: 99454148-99454148 on Build GRCh38
rs568570598 Chr.7: 99454306-99454306 on Build GRCh38
rs376215260 Chr.7: 99454205-99454205 on Build GRCh38
rs147308727 Chr.7: 99454317-99454317 on Build GRCh38
rs748972240 Chr.7: 99454282-99454282 on Build GRCh38
rs903353306 Chr.7: 99454232-99454232 on Build GRCh38
rs769370096 Chr.7: 99454088-99454088 on Build GRCh38
rs372992949 Chr.7: 99454188-99454188 on Build GRCh38
rs554391996 Chr.7: 99454355-99454355 on Build GRCh38
rs149850144 Chr.7: 99453979-99453979 on Build GRCh38
rs886679605 Chr.7: 99454249-99454249 on Build GRCh38
rs767832491 Chr.7: 99453915-99453915 on Build GRCh38
rs541958834 Chr.7: 99454150-99454150 on Build GRCh38
rs866394207 Chr.7: 99453987-99453987 on Build GRCh38
rs764195891 Chr.7: 99454112-99454112 on Build GRCh38
rs1005107457 Chr.7: 99454290-99454290 on Build GRCh38
rs750955832 Chr.7: 99454032-99454032 on Build GRCh38
rs775989764 Chr.7: 99453946-99453946 on Build GRCh38
rs45558438 Chr.7: 99454233-99454233 on Build GRCh38
rs370277726 Chr.7: 99454179-99454179 on Build GRCh38
rs528792174 Chr.7: 99454236-99454236 on Build GRCh38
rs186458191 Chr.7: 99453995-99453995 on Build GRCh38
rs563773674 Chr.7: 99454152-99454152 on Build GRCh38
rs139528752 Chr.7: 99454094-99454094 on Build GRCh38
rs778460888 Chr.7: 99454219-99454219 on Build GRCh38
rs1019979082 Chr.7: 99454346-99454346 on Build GRCh38
rs1044422101 Chr.7: 99454223-99454223 on Build GRCh38
rs556930324 Chr.7: 99454304-99454304 on Build GRCh38
rs371087605 Chr.7: 99453975-99453975 on Build GRCh38
rs779340033 Chr.7: 99454025-99454025 on Build GRCh38
rs770792634 Chr.7: 99454091-99454091 on Build GRCh38
rs752901925 Chr.7: 99453983-99453983 on Build GRCh38
rs751838986 Chr.7: 99454160-99454160 on Build GRCh38
rs890873800 Chr.7: 99454308-99454308 on Build GRCh38
rs757401363 Chr.7: 99454015-99454015 on Build GRCh38
rs370251051 Chr.7: 99453909-99453909 on Build GRCh38
rs780938148 Chr.7: 99454052-99454052 on Build GRCh38
rs200000721 Chr.7: 99453945-99453945 on Build GRCh38
rs1007876935 Chr.7: 99454344-99454344 on Build GRCh38
rs754315479 Chr.7: 99453922-99453922 on Build GRCh38
rs772444169 Chr.7: 99453934-99453934 on Build GRCh38
rs889490215 Chr.7: 99454328-99454328 on Build GRCh38
rs938730691 Chr.7: 99454124-99454124 on Build GRCh38
rs747588365 Chr.7: 99454061-99454061 on Build GRCh38
rs377447208 Chr.7: 99453950-99453950 on Build GRCh38
rs375692493 Chr.7: 99454158-99454158 on Build GRCh38
rs1003541872 Chr.7: 99453935-99453935 on Build GRCh38
rs963719479 Chr.7: 99454337-99454337 on Build GRCh38
rs1037891738 Chr.7: 99454292-99454292 on Build GRCh38
rs749243303 Chr.7: 99454090-99454090 on Build GRCh38

View Full Product Details