Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99504562-99505061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZKSCAN5
Entrez Gene ID
23660
Gene Name
zinc finger with KRAB and SCAN domains 5
Gene Aliases
ZFP-95, ZFP95, ZNF914, ZSCAN37
UniGene
Hs.110839
Gene Chromosome Location
Chr.7: 99501296-99534106 on Build GRCh38
Transcripts
NM_001318082.1 NM_001318083.1 NM_014569.3 NM_145102.3 XM_017011918.1 XM_017011919.1 XM_017011921.1 XM_017011922.1 XM_017011923.1 XM_017011924.1 XM_017011926.1 XM_017011927.1

SNP Detail

SNP ID Chromosome Location
rs577210557 Chr.7: 99504715-99504715 on Build GRCh38
rs181872478 Chr.7: 99504961-99504961 on Build GRCh38
rs912549888 Chr.7: 99504688-99504688 on Build GRCh38
rs990938721 Chr.7: 99505040-99505040 on Build GRCh38
rs1046234706 Chr.7: 99504716-99504716 on Build GRCh38
rs914674716 Chr.7: 99504696-99504696 on Build GRCh38
rs561600817 Chr.7: 99504922-99504922 on Build GRCh38
rs1017298935 Chr.7: 99504864-99504864 on Build GRCh38
rs926427590 Chr.7: 99505028-99505028 on Build GRCh38
rs893842849 Chr.7: 99504754-99504754 on Build GRCh38
rs536863505 Chr.7: 99504630-99504630 on Build GRCh38
rs560019633 Chr.7: 99504789-99504789 on Build GRCh38
rs927803878 Chr.7: 99504976-99504976 on Build GRCh38
rs867617336 Chr.7: 99504891-99504891 on Build GRCh38
rs536475963 Chr.7: 99504625-99504625 on Build GRCh38
rs927757451 Chr.7: 99504733-99504733 on Build GRCh38
rs376296814 Chr.7: 99504600-99504600 on Build GRCh38
rs956011303 Chr.7: 99504958-99504958 on Build GRCh38
rs567437315 Chr.7: 99504695-99504695 on Build GRCh38
rs938094512 Chr.7: 99504743-99504743 on Build GRCh38
rs963053566 Chr.7: 99504878-99504878 on Build GRCh38
rs1043073907 Chr.7: 99504792-99504792 on Build GRCh38
rs914601191 Chr.7: 99504987-99504987 on Build GRCh38
rs963178105 Chr.7: 99504655-99504655 on Build GRCh38
rs562180625 Chr.7: 99504691-99504691 on Build GRCh38
rs549178432 Chr.7: 99504858-99504858 on Build GRCh38
rs1029844962 Chr.7: 99504830-99504830 on Build GRCh38
rs954469143 Chr.7: 99504651-99504651 on Build GRCh38
rs10265587 Chr.7: 99504862-99504862 on Build GRCh38
rs954241060 Chr.7: 99504920-99504920 on Build GRCh38
rs1021195884 Chr.7: 99504588-99504588 on Build GRCh38
rs544949296 Chr.7: 99504752-99504752 on Build GRCh38
rs946694104 Chr.7: 99504760-99504760 on Build GRCh38
rs985815186 Chr.7: 99504652-99504652 on Build GRCh38
rs902767756 Chr.7: 99504804-99504804 on Build GRCh38
rs368242985 Chr.7: 99504631-99504631 on Build GRCh38
rs1028484193 Chr.7: 99504903-99504903 on Build GRCh38
rs896132211 Chr.7: 99504856-99504856 on Build GRCh38
rs1043569124 Chr.7: 99504785-99504785 on Build GRCh38
rs937816425 Chr.7: 99504983-99504983 on Build GRCh38
rs963001430 Chr.7: 99504677-99504677 on Build GRCh38
rs995704073 Chr.7: 99504880-99504880 on Build GRCh38
rs751467177 Chr.7: 99504693-99504693 on Build GRCh38
rs950674302 Chr.7: 99504940-99504940 on Build GRCh38
rs947489236 Chr.7: 99504992-99504992 on Build GRCh38
rs919177800 Chr.7: 99504681-99504681 on Build GRCh38
rs1002111283 Chr.7: 99504825-99504825 on Build GRCh38
rs767784318 Chr.7: 99504700-99504700 on Build GRCh38
rs1015726742 Chr.7: 99504871-99504871 on Build GRCh38
rs962496705 Chr.7: 99504877-99504877 on Build GRCh38
rs150225249 Chr.7: 99504670-99504670 on Build GRCh38
rs190220387 Chr.7: 99504679-99504679 on Build GRCh38
rs998810365 Chr.7: 99504828-99504828 on Build GRCh38
rs974277769 Chr.7: 99504669-99504669 on Build GRCh38
rs1026363843 Chr.7: 99504932-99504932 on Build GRCh38
rs554830734 Chr.7: 99504641-99504641 on Build GRCh38
rs1044907836 Chr.7: 99505014-99505015 on Build GRCh38
rs145737499 Chr.7: 99504838-99504838 on Build GRCh38
rs550287117 Chr.7: 99504924-99504924 on Build GRCh38
rs566802436 Chr.7: 99504599-99504600 on Build GRCh38
rs1053589355 Chr.7: 99504695-99504695 on Build GRCh38
rs548492623 Chr.7: 99504930-99504930 on Build GRCh38

View Full Product Details