Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100034824-100035318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZKSCAN1
Entrez Gene ID
7586
Gene Name
zinc finger with KRAB and SCAN domains 1
Gene Aliases
9130423L19Rik, KOX18, PHZ-37, ZNF139, ZNF36, ZSCAN33
UniGene
Hs.615360
Gene Chromosome Location
Chr.7: 100015572-100045523 on Build GRCh38
Transcripts
NM_001287054.1 NM_001287055.1 NM_003439.2 XM_005250565.4 XM_011516559.2

SNP Detail

SNP ID Chromosome Location
rs967025846 Chr.7: 100035048-100035049 on Build GRCh38
rs769332942 Chr.7: 100034928-100034928 on Build GRCh38
rs567992063 Chr.7: 100035243-100035243 on Build GRCh38
rs941860069 Chr.7: 100035220-100035220 on Build GRCh38
rs1034199357 Chr.7: 100035007-100035007 on Build GRCh38
rs898000763 Chr.7: 100035164-100035164 on Build GRCh38
rs900144494 Chr.7: 100034964-100034964 on Build GRCh38
rs10274894 Chr.7: 100035209-100035209 on Build GRCh38
rs562966926 Chr.7: 100035061-100035061 on Build GRCh38
rs866178428 Chr.7: 100034993-100034993 on Build GRCh38
rs910325059 Chr.7: 100035138-100035138 on Build GRCh38
rs898115392 Chr.7: 100034918-100034918 on Build GRCh38
rs929427124 Chr.7: 100035165-100035165 on Build GRCh38
rs906535985 Chr.7: 100034945-100034945 on Build GRCh38
rs1050604964 Chr.7: 100034885-100034885 on Build GRCh38
rs929090071 Chr.7: 100034893-100034895 on Build GRCh38
rs977251943 Chr.7: 100035286-100035286 on Build GRCh38
rs1006411118 Chr.7: 100034896-100034896 on Build GRCh38
rs189925036 Chr.7: 100035289-100035289 on Build GRCh38
rs772411836 Chr.7: 100035288-100035288 on Build GRCh38
rs1032541362 Chr.7: 100035076-100035076 on Build GRCh38
rs958096194 Chr.7: 100035014-100035014 on Build GRCh38
rs888840465 Chr.7: 100034889-100034889 on Build GRCh38
rs1021405463 Chr.7: 100035038-100035038 on Build GRCh38
rs990841599 Chr.7: 100035137-100035137 on Build GRCh38
rs932960562 Chr.7: 100034857-100034857 on Build GRCh38
rs185137337 Chr.7: 100035150-100035150 on Build GRCh38
rs977742508 Chr.7: 100035082-100035082 on Build GRCh38
rs900260511 Chr.7: 100035285-100035285 on Build GRCh38
rs887514403 Chr.7: 100034905-100034905 on Build GRCh38
rs967305198 Chr.7: 100035057-100035057 on Build GRCh38
rs923177713 Chr.7: 100035085-100035085 on Build GRCh38
rs932991403 Chr.7: 100035090-100035090 on Build GRCh38
rs982082856 Chr.7: 100035159-100035159 on Build GRCh38
rs769059701 Chr.7: 100035153-100035153 on Build GRCh38
rs893926699 Chr.7: 100035239-100035239 on Build GRCh38
rs1037863950 Chr.7: 100034897-100034897 on Build GRCh38
rs528408364 Chr.7: 100035162-100035162 on Build GRCh38
rs906761962 Chr.7: 100035189-100035189 on Build GRCh38
rs779656729 Chr.7: 100034958-100034958 on Build GRCh38
rs1006001855 Chr.7: 100034927-100034927 on Build GRCh38
rs530640356 Chr.7: 100035113-100035113 on Build GRCh38
rs1002639598 Chr.7: 100035190-100035190 on Build GRCh38
rs929271112 Chr.7: 100035169-100035169 on Build GRCh38
rs967190568 Chr.7: 100035253-100035253 on Build GRCh38
rs1000094701 Chr.7: 100035060-100035060 on Build GRCh38
rs147402573 Chr.7: 100035035-100035035 on Build GRCh38
rs774513536 Chr.7: 100035166-100035166 on Build GRCh38
rs139814922 Chr.7: 100035133-100035133 on Build GRCh38
rs1010942436 Chr.7: 100035240-100035240 on Build GRCh38
rs1025187053 Chr.7: 100034932-100034932 on Build GRCh38

View Full Product Details