Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100104946-100105445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP4M1
Entrez Gene ID
9179
Gene Name
adaptor related protein complex 4 mu 1 subunit
Gene Aliases
CPSQ3, MU-4, MU-ARP2, SPG50
UniGene
Hs.632317
Gene Chromosome Location
Chr.7: 100101413-100107180 on Build GRCh38
Transcripts
NM_004722.3 XM_005250689.4 XM_005250690.4 XM_006716175.3 XM_011516685.2 XM_017012790.1 XM_017012791.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6565794 c.826C>T p.Q276* Chr.7: 100105338-100105338 on Build GRCh38
3715865 c.799C>G p.H267D Chr.7: 100105311-100105311 on Build GRCh38
297846 c.733_736delGGGC p.G245fs*14 Chr.7: 100105245-100105248 on Build GRCh38
3762938 c.812G>A p.R271H Chr.7: 100105324-100105324 on Build GRCh38
1093966 c.832G>A p.E278K Chr.7: 100105344-100105344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772536501 Chr.7: 100105031-100105031 on Build GRCh38
rs369201294 Chr.7: 100105050-100105050 on Build GRCh38
rs759324120 Chr.7: 100105223-100105223 on Build GRCh38
rs878997437 Chr.7: 100104977-100104977 on Build GRCh38
rs895548740 Chr.7: 100105321-100105321 on Build GRCh38
rs369306233 Chr.7: 100105384-100105384 on Build GRCh38
rs377555463 Chr.7: 100104984-100104984 on Build GRCh38
rs763873727 Chr.7: 100105039-100105039 on Build GRCh38
rs376551171 Chr.7: 100105228-100105228 on Build GRCh38
rs374130321 Chr.7: 100105131-100105131 on Build GRCh38
rs370797954 Chr.7: 100105033-100105033 on Build GRCh38
rs1016995917 Chr.7: 100105116-100105116 on Build GRCh38
rs553061919 Chr.7: 100105273-100105273 on Build GRCh38
rs149820983 Chr.7: 100105075-100105075 on Build GRCh38
rs370260221 Chr.7: 100105239-100105239 on Build GRCh38
rs779863105 Chr.7: 100104980-100104980 on Build GRCh38
rs371712994 Chr.7: 100105051-100105051 on Build GRCh38
rs1013897342 Chr.7: 100105158-100105158 on Build GRCh38
rs774947350 Chr.7: 100105018-100105018 on Build GRCh38
rs561103710 Chr.7: 100105375-100105375 on Build GRCh38
rs545459309 Chr.7: 100105283-100105283 on Build GRCh38
rs927923217 Chr.7: 100105175-100105175 on Build GRCh38
rs937805871 Chr.7: 100105176-100105176 on Build GRCh38
rs780030221 Chr.7: 100105314-100105314 on Build GRCh38
rs565167866 Chr.7: 100105403-100105403 on Build GRCh38
rs768107678 Chr.7: 100105088-100105094 on Build GRCh38
rs769502788 Chr.7: 100105263-100105263 on Build GRCh38
rs758148261 Chr.7: 100104978-100104978 on Build GRCh38
rs768917277 Chr.7: 100105129-100105129 on Build GRCh38
rs369874184 Chr.7: 100105406-100105406 on Build GRCh38
rs777352543 Chr.7: 100105343-100105343 on Build GRCh38
rs759428856 Chr.7: 100105278-100105278 on Build GRCh38
rs748639289 Chr.7: 100105218-100105219 on Build GRCh38
rs113641113 Chr.7: 100105146-100105146 on Build GRCh38
rs781616312 Chr.7: 100105323-100105323 on Build GRCh38
rs565296810 Chr.7: 100105315-100105315 on Build GRCh38
rs373696169 Chr.7: 100105254-100105254 on Build GRCh38
rs750601796 Chr.7: 100105246-100105246 on Build GRCh38
rs773785064 Chr.7: 100105354-100105354 on Build GRCh38
rs745310821 Chr.7: 100105412-100105412 on Build GRCh38
rs745807118 Chr.7: 100105355-100105355 on Build GRCh38
rs774897281 Chr.7: 100105219-100105220 on Build GRCh38
rs1042860628 Chr.7: 100105112-100105112 on Build GRCh38
rs766796124 Chr.7: 100105024-100105024 on Build GRCh38
rs753222064 Chr.7: 100105393-100105393 on Build GRCh38
rs748826096 Chr.7: 100105030-100105031 on Build GRCh38
rs375327296 Chr.7: 100105063-100105063 on Build GRCh38
rs747332440 Chr.7: 100105127-100105127 on Build GRCh38
rs767953033 Chr.7: 100105392-100105392 on Build GRCh38
rs767244572 Chr.7: 100105224-100105224 on Build GRCh38
rs759950294 Chr.7: 100105177-100105177 on Build GRCh38
rs897456893 Chr.7: 100105332-100105332 on Build GRCh38
rs761038629 Chr.7: 100104994-100104994 on Build GRCh38
rs144253083 Chr.7: 100105289-100105289 on Build GRCh38
rs562389602 Chr.7: 100104986-100104986 on Build GRCh38
rs765352573 Chr.7: 100105178-100105178 on Build GRCh38
rs147670505 Chr.7: 100105325-100105325 on Build GRCh38
rs761491801 Chr.7: 100105132-100105132 on Build GRCh38
rs771629792 Chr.7: 100105418-100105418 on Build GRCh38
rs757605059 Chr.7: 100105232-100105232 on Build GRCh38
rs375680568 Chr.7: 100104981-100104981 on Build GRCh38
rs368830741 Chr.7: 100105258-100105258 on Build GRCh38
rs746411626 Chr.7: 100105020-100105020 on Build GRCh38
rs971782025 Chr.7: 100105299-100105299 on Build GRCh38
rs1042146797 Chr.7: 100105226-100105226 on Build GRCh38
rs895567354 Chr.7: 100105138-100105138 on Build GRCh38
rs777989411 Chr.7: 100105077-100105077 on Build GRCh38
rs200726009 Chr.7: 100105267-100105267 on Build GRCh38
rs756416380 Chr.7: 100104982-100104982 on Build GRCh38
rs200900793 Chr.7: 100105405-100105405 on Build GRCh38
rs766148541 Chr.7: 100105065-100105065 on Build GRCh38
rs769269742 Chr.7: 100105142-100105142 on Build GRCh38
rs377672447 Chr.7: 100105207-100105207 on Build GRCh38
rs772522035 Chr.7: 100105219-100105224 on Build GRCh38
rs377426269 Chr.7: 100105121-100105121 on Build GRCh38
rs775577262 Chr.7: 100105367-100105367 on Build GRCh38
rs372641895 Chr.7: 100105068-100105068 on Build GRCh38
rs534691439 Chr.7: 100105162-100105162 on Build GRCh38
rs953631200 Chr.7: 100105362-100105362 on Build GRCh38
rs772767645 Chr.7: 100105122-100105122 on Build GRCh38
rs370110611 Chr.7: 100105124-100105124 on Build GRCh38
rs201255015 Chr.7: 100105100-100105100 on Build GRCh38
rs775520329 Chr.7: 100105421-100105421 on Build GRCh38
rs781303911 Chr.7: 100105257-100105257 on Build GRCh38
rs986194668 Chr.7: 100104996-100104996 on Build GRCh38
rs902199140 Chr.7: 100105053-100105053 on Build GRCh38
rs368267562 Chr.7: 100105352-100105352 on Build GRCh38
rs374785430 Chr.7: 100105035-100105035 on Build GRCh38
rs1027881207 Chr.7: 100105351-100105351 on Build GRCh38
rs771974744 Chr.7: 100105360-100105360 on Build GRCh38
rs1051745199 Chr.7: 100105253-100105253 on Build GRCh38
rs752745814 Chr.7: 100105262-100105262 on Build GRCh38
rs145887263 Chr.7: 100105252-100105252 on Build GRCh38
rs766269157 Chr.7: 100105208-100105208 on Build GRCh38
rs748967782 Chr.7: 100105344-100105344 on Build GRCh38
rs750687666 Chr.7: 100105298-100105298 on Build GRCh38
rs574214816 Chr.7: 100105010-100105010 on Build GRCh38
rs982409621 Chr.7: 100105172-100105172 on Build GRCh38
rs371795483 Chr.7: 100105394-100105394 on Build GRCh38
rs755494077 Chr.7: 100105319-100105319 on Build GRCh38
rs1007065682 Chr.7: 100105096-100105096 on Build GRCh38
rs750376210 Chr.7: 100104976-100104976 on Build GRCh38
rs376000948 Chr.7: 100105378-100105378 on Build GRCh38
rs375129284 Chr.7: 100105216-100105216 on Build GRCh38
rs374565045 Chr.7: 100105422-100105422 on Build GRCh38
rs762387733 Chr.7: 100105266-100105266 on Build GRCh38
rs962896326 Chr.7: 100105265-100105265 on Build GRCh38
rs371973205 Chr.7: 100105376-100105376 on Build GRCh38
rs902147115 Chr.7: 100105230-100105230 on Build GRCh38
rs772766548 Chr.7: 100105202-100105202 on Build GRCh38
rs758378130 Chr.7: 100105064-100105064 on Build GRCh38
rs41280968 Chr.7: 100105324-100105324 on Build GRCh38
rs766467478 Chr.7: 100105380-100105380 on Build GRCh38
rs971452596 Chr.7: 100105171-100105171 on Build GRCh38
rs946448880 Chr.7: 100105218-100105218 on Build GRCh38
rs994513548 Chr.7: 100105338-100105338 on Build GRCh38
rs576875998 Chr.7: 100105040-100105040 on Build GRCh38
rs202088924 Chr.7: 100105090-100105090 on Build GRCh38
rs773729417 Chr.7: 100105219-100105222 on Build GRCh38
rs749492321 Chr.7: 100105084-100105084 on Build GRCh38
rs774302896 Chr.7: 100105274-100105274 on Build GRCh38
rs779177357 Chr.7: 100105106-100105106 on Build GRCh38
rs1010643109 Chr.7: 100105026-100105026 on Build GRCh38
rs760489210 Chr.7: 100105371-100105371 on Build GRCh38
rs117820338 Chr.7: 100105390-100105390 on Build GRCh38

View Full Product Details