Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100177834-100178335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAG3
Entrez Gene ID
10734
Gene Name
stromal antigen 3
Gene Aliases
-
UniGene
Hs.592283
Gene Chromosome Location
Chr.7: 100177563-100219334 on Build GRCh38
Transcripts
NM_001282716.1 NM_001282717.1 NM_001282718.1 NM_012447.3 XM_011515742.1 XM_017011683.1 XM_017011684.1 XM_017011685.1 XM_017011686.1 XM_017011687.1

SNP Detail

SNP ID Chromosome Location
rs982553620 Chr.7: 100178016-100178016 on Build GRCh38
rs1035600971 Chr.7: 100178024-100178024 on Build GRCh38
rs530183008 Chr.7: 100178239-100178239 on Build GRCh38
rs915718000 Chr.7: 100178045-100178045 on Build GRCh38
rs142005563 Chr.7: 100178059-100178060 on Build GRCh38
rs1050875703 Chr.7: 100178140-100178141 on Build GRCh38
rs543793938 Chr.7: 100178013-100178013 on Build GRCh38
rs886552841 Chr.7: 100178218-100178218 on Build GRCh38
rs934329754 Chr.7: 100178133-100178133 on Build GRCh38
rs78894444 Chr.7: 100178310-100178310 on Build GRCh38
rs899342773 Chr.7: 100177960-100177960 on Build GRCh38
rs890144318 Chr.7: 100178142-100178142 on Build GRCh38
rs990779007 Chr.7: 100177954-100177954 on Build GRCh38
rs998521292 Chr.7: 100178213-100178213 on Build GRCh38
rs78325712 Chr.7: 100178028-100178028 on Build GRCh38
rs190818574 Chr.7: 100178150-100178150 on Build GRCh38
rs1017760218 Chr.7: 100177938-100177938 on Build GRCh38
rs756879563 Chr.7: 100178017-100178017 on Build GRCh38
rs73397519 Chr.7: 100177992-100177992 on Build GRCh38
rs1007711958 Chr.7: 100177935-100177935 on Build GRCh38
rs902816969 Chr.7: 100177878-100177878 on Build GRCh38
rs916152047 Chr.7: 100177956-100177956 on Build GRCh38
rs947232861 Chr.7: 100178046-100178046 on Build GRCh38
rs890729663 Chr.7: 100177925-100177925 on Build GRCh38
rs752314026 Chr.7: 100178031-100178031 on Build GRCh38
rs929343412 Chr.7: 100178055-100178055 on Build GRCh38
rs937270819 Chr.7: 100177979-100177979 on Build GRCh38
rs566187464 Chr.7: 100178224-100178224 on Build GRCh38
rs1022840512 Chr.7: 100178259-100178259 on Build GRCh38
rs112326232 Chr.7: 100178246-100178246 on Build GRCh38
rs899291978 Chr.7: 100178187-100178187 on Build GRCh38
rs899728808 Chr.7: 100178114-100178114 on Build GRCh38
rs1026390736 Chr.7: 100178199-100178199 on Build GRCh38
rs978443125 Chr.7: 100178053-100178053 on Build GRCh38
rs949914372 Chr.7: 100177963-100177963 on Build GRCh38
rs1051889415 Chr.7: 100178134-100178134 on Build GRCh38
rs979038879 Chr.7: 100178295-100178295 on Build GRCh38
rs1038718983 Chr.7: 100178096-100178096 on Build GRCh38
rs1039311104 Chr.7: 100178186-100178186 on Build GRCh38
rs957646932 Chr.7: 100177942-100177942 on Build GRCh38
rs1003527727 Chr.7: 100178235-100178235 on Build GRCh38
rs114147543 Chr.7: 100178247-100178247 on Build GRCh38
rs71276827 Chr.7: 100178062-100178063 on Build GRCh38
rs1045604046 Chr.7: 100177970-100177970 on Build GRCh38
rs60823426 Chr.7: 100178136-100178137 on Build GRCh38
rs998931390 Chr.7: 100177891-100177891 on Build GRCh38
rs567378534 Chr.7: 100178251-100178251 on Build GRCh38
rs1052021636 Chr.7: 100177895-100177895 on Build GRCh38
rs1026818878 Chr.7: 100177985-100177985 on Build GRCh38
rs970328582 Chr.7: 100178277-100178277 on Build GRCh38
rs35270185 Chr.7: 100178061-100178062 on Build GRCh38
rs1031316874 Chr.7: 100178237-100178237 on Build GRCh38
rs977090692 Chr.7: 100177911-100177911 on Build GRCh38
rs3835191 Chr.7: 100178060-100178061 on Build GRCh38
rs924397270 Chr.7: 100178097-100178097 on Build GRCh38
rs905758628 Chr.7: 100177974-100177974 on Build GRCh38
rs995043074 Chr.7: 100178190-100178190 on Build GRCh38
rs1031263000 Chr.7: 100177942-100177942 on Build GRCh38
rs968522986 Chr.7: 100178294-100178294 on Build GRCh38
rs568385032 Chr.7: 100178091-100178092 on Build GRCh38
rs547766431 Chr.7: 100178222-100178222 on Build GRCh38
rs887756316 Chr.7: 100178075-100178075 on Build GRCh38

View Full Product Details