Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100319594-100320092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPDYE3
Entrez Gene ID
441272
Gene Name
speedy/RINGO cell cycle regulator family member E3
Gene Aliases
-
UniGene
Hs.703626
Gene Chromosome Location
Chr.7: 100306799-100322196 on Build GRCh38
Transcripts
NM_001004351.4 XM_011516234.2 XM_011516235.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
748216 c.1542C>T p.F514F Chr.7: 100319760-100319760 on Build GRCh38
6235787 c.1395G>C p.K465N Chr.7: 100319613-100319613 on Build GRCh38
748214 c.1597G>A p.A533T Chr.7: 100319937-100319937 on Build GRCh38
3698651 c.1408C>A p.L470M Chr.7: 100319626-100319626 on Build GRCh38
6279078 c.1587G>A p.E529E Chr.7: 100319805-100319805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765856582 Chr.7: 100319919-100319919 on Build GRCh38
rs367569338 Chr.7: 100319771-100319771 on Build GRCh38
rs201228230 Chr.7: 100319693-100319693 on Build GRCh38
rs923089148 Chr.7: 100319922-100319922 on Build GRCh38
rs754797039 Chr.7: 100319773-100319773 on Build GRCh38
rs895394818 Chr.7: 100320052-100320052 on Build GRCh38
rs763696295 Chr.7: 100319745-100319745 on Build GRCh38
rs949594790 Chr.7: 100320062-100320062 on Build GRCh38
rs763807304 Chr.7: 100320005-100320005 on Build GRCh38
rs762243256 Chr.7: 100319914-100319914 on Build GRCh38
rs201844139 Chr.7: 100319782-100319782 on Build GRCh38
rs762608726 Chr.7: 100319917-100319917 on Build GRCh38
rs777054893 Chr.7: 100319651-100319651 on Build GRCh38
rs374508596 Chr.7: 100319817-100319817 on Build GRCh38
rs536053477 Chr.7: 100319973-100319973 on Build GRCh38
rs747495576 Chr.7: 100319902-100319902 on Build GRCh38
rs777001966 Chr.7: 100319913-100319913 on Build GRCh38
rs780118835 Chr.7: 100319966-100319966 on Build GRCh38
rs189896330 Chr.7: 100319827-100319827 on Build GRCh38
rs779951592 Chr.7: 100319702-100319702 on Build GRCh38
rs764858992 Chr.7: 100319797-100319797 on Build GRCh38
rs768580650 Chr.7: 100319820-100319821 on Build GRCh38
rs756800095 Chr.7: 100319694-100319694 on Build GRCh38
rs758425540 Chr.7: 100319965-100319965 on Build GRCh38
rs750413655 Chr.7: 100319762-100319762 on Build GRCh38
rs771003503 Chr.7: 100319777-100319777 on Build GRCh38
rs767238582 Chr.7: 100319952-100319952 on Build GRCh38
rs371409173 Chr.7: 100319789-100319789 on Build GRCh38
rs375412654 Chr.7: 100319733-100319733 on Build GRCh38
rs767373090 Chr.7: 100319889-100319890 on Build GRCh38
rs558482666 Chr.7: 100319657-100319657 on Build GRCh38
rs760527204 Chr.7: 100320004-100320004 on Build GRCh38
rs1017516175 Chr.7: 100319870-100319870 on Build GRCh38
rs770267336 Chr.7: 100319974-100319974 on Build GRCh38
rs575066768 Chr.7: 100319712-100319712 on Build GRCh38
rs370742497 Chr.7: 100319747-100319747 on Build GRCh38
rs577036288 Chr.7: 100319660-100319660 on Build GRCh38
rs774835240 Chr.7: 100319662-100319662 on Build GRCh38
rs372544800 Chr.7: 100319633-100319633 on Build GRCh38
rs759477480 Chr.7: 100319946-100319946 on Build GRCh38
rs1006184847 Chr.7: 100319760-100319760 on Build GRCh38
rs565566030 Chr.7: 100319967-100319967 on Build GRCh38
rs766483210 Chr.7: 100320023-100320023 on Build GRCh38
rs752539303 Chr.7: 100319955-100319955 on Build GRCh38
rs764785874 Chr.7: 100319748-100319748 on Build GRCh38
rs778924634 Chr.7: 100319780-100319780 on Build GRCh38
rs1027408903 Chr.7: 100319828-100319828 on Build GRCh38
rs376903923 Chr.7: 100319852-100319852 on Build GRCh38
rs780486335 Chr.7: 100319901-100319901 on Build GRCh38
rs747287616 Chr.7: 100319723-100319726 on Build GRCh38
rs776705721 Chr.7: 100319771-100319772 on Build GRCh38
rs755873868 Chr.7: 100319957-100319957 on Build GRCh38
rs770378602 Chr.7: 100319976-100319976 on Build GRCh38
rs111727167 Chr.7: 100320040-100320040 on Build GRCh38
rs772689053 Chr.7: 100319954-100319981 on Build GRCh38
rs200369353 Chr.7: 100319812-100319812 on Build GRCh38
rs751713672 Chr.7: 100320027-100320027 on Build GRCh38
rs770868873 Chr.7: 100319760-100319770 on Build GRCh38
rs777914689 Chr.7: 100319775-100319775 on Build GRCh38
rs750633525 Chr.7: 100319964-100319964 on Build GRCh38
rs769055476 Chr.7: 100319792-100319792 on Build GRCh38
rs761081183 Chr.7: 100319744-100319744 on Build GRCh38
rs370768793 Chr.7: 100319625-100319625 on Build GRCh38
rs772190255 Chr.7: 100319894-100319894 on Build GRCh38
rs751500593 Chr.7: 100319832-100319832 on Build GRCh38
rs766292528 Chr.7: 100319823-100319823 on Build GRCh38
rs748903725 Chr.7: 100319975-100319975 on Build GRCh38
rs759952351 Chr.7: 100319665-100319665 on Build GRCh38
rs775669111 Chr.7: 100319743-100319743 on Build GRCh38
rs541094130 Chr.7: 100319687-100319687 on Build GRCh38
rs757095118 Chr.7: 100319632-100319632 on Build GRCh38
rs748421476 Chr.7: 100319704-100319704 on Build GRCh38
rs754091737 Chr.7: 100319887-100319887 on Build GRCh38
rs749280611 Chr.7: 100319631-100319631 on Build GRCh38
rs757422380 Chr.7: 100319888-100319888 on Build GRCh38
rs200892093 Chr.7: 100319614-100319614 on Build GRCh38
rs773450291 Chr.7: 100319715-100319715 on Build GRCh38
rs112622797 Chr.7: 100319793-100319793 on Build GRCh38
rs1065453 Chr.7: 100319613-100319613 on Build GRCh38
rs945125599 Chr.7: 100319929-100319929 on Build GRCh38
rs745701181 Chr.7: 100319640-100319640 on Build GRCh38
rs528578494 Chr.7: 100319890-100319890 on Build GRCh38
rs761014553 Chr.7: 100319791-100319791 on Build GRCh38
rs781753979 Chr.7: 100319970-100319970 on Build GRCh38
rs750653459 Chr.7: 100320007-100320007 on Build GRCh38
rs755553640 Chr.7: 100319968-100319968 on Build GRCh38
rs745989229 Chr.7: 100319893-100319893 on Build GRCh38
rs546672565 Chr.7: 100319958-100319958 on Build GRCh38
rs774333230 Chr.7: 100319839-100319839 on Build GRCh38
rs755684175 Chr.7: 100319686-100319686 on Build GRCh38
rs758947901 Chr.7: 100319980-100319980 on Build GRCh38
rs779064773 Chr.7: 100319699-100319699 on Build GRCh38
rs767813103 Chr.7: 100319679-100319679 on Build GRCh38
rs749998865 Chr.7: 100319806-100319806 on Build GRCh38
rs759969100 Chr.7: 100319739-100319739 on Build GRCh38
rs763064256 Chr.7: 100319659-100319659 on Build GRCh38
rs774851916 Chr.7: 100319734-100319734 on Build GRCh38
rs748248459 Chr.7: 100319656-100319656 on Build GRCh38
rs376445936 Chr.7: 100319746-100319746 on Build GRCh38
rs370261673 Chr.7: 100319725-100319725 on Build GRCh38
rs752400008 Chr.7: 100319683-100319683 on Build GRCh38
rs758269161 Chr.7: 100319700-100319700 on Build GRCh38
rs374034824 Chr.7: 100319770-100319770 on Build GRCh38
rs201988177 Chr.7: 100320003-100320003 on Build GRCh38
rs566270077 Chr.7: 100319850-100319850 on Build GRCh38
rs767532268 Chr.7: 100319992-100319992 on Build GRCh38
rs777828553 Chr.7: 100319682-100319684 on Build GRCh38
rs749454523 Chr.7: 100319776-100319776 on Build GRCh38
rs758592521 Chr.7: 100320022-100320022 on Build GRCh38
rs749160923 Chr.7: 100319801-100319809 on Build GRCh38
rs761492932 Chr.7: 100319850-100319850 on Build GRCh38
rs749394083 Chr.7: 100319726-100319726 on Build GRCh38
rs768910978 Chr.7: 100319908-100319908 on Build GRCh38
rs773863013 Chr.7: 100319978-100319978 on Build GRCh38
rs774160710 Chr.7: 100319943-100319943 on Build GRCh38
rs771992167 Chr.7: 100319641-100319641 on Build GRCh38
rs772992715 Chr.7: 100319915-100319915 on Build GRCh38
rs866504552 Chr.7: 100319869-100319869 on Build GRCh38

View Full Product Details