Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100554003-100554505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGFG2
Entrez Gene ID
3268
Gene Name
ArfGAP with FG repeats 2
Gene Aliases
HRBL, RABR
UniGene
Hs.521083
Gene Chromosome Location
Chr.7: 100539211-100568220 on Build GRCh38
Transcripts
NM_006076.4 XM_005250306.2 XM_017012112.1 XM_017012113.1 XM_017012114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3630780 c.642C>T p.P214P Chr.7: 100554149-100554149 on Build GRCh38
451972 c.694G>A p.D232N Chr.7: 100554201-100554201 on Build GRCh38
598029 c.699C>T p.P233P Chr.7: 100554206-100554206 on Build GRCh38
7130830 c.591C>T p.V197V Chr.7: 100554098-100554098 on Build GRCh38
7245449 c.631C>G p.Q211E Chr.7: 100554138-100554138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008046860 Chr.7: 100554425-100554425 on Build GRCh38
rs943724850 Chr.7: 100554185-100554185 on Build GRCh38
rs763384626 Chr.7: 100554167-100554168 on Build GRCh38
rs776784932 Chr.7: 100554283-100554283 on Build GRCh38
rs775661541 Chr.7: 100554115-100554115 on Build GRCh38
rs148018854 Chr.7: 100554194-100554194 on Build GRCh38
rs770692803 Chr.7: 100554221-100554221 on Build GRCh38
rs762386901 Chr.7: 100554145-100554145 on Build GRCh38
rs1028167470 Chr.7: 100554402-100554402 on Build GRCh38
rs765664721 Chr.7: 100554066-100554066 on Build GRCh38
rs765605174 Chr.7: 100554151-100554151 on Build GRCh38
rs773144781 Chr.7: 100554419-100554419 on Build GRCh38
rs1048589222 Chr.7: 100554338-100554338 on Build GRCh38
rs760794616 Chr.7: 100554129-100554129 on Build GRCh38
rs747557845 Chr.7: 100554203-100554203 on Build GRCh38
rs374005428 Chr.7: 100554231-100554231 on Build GRCh38
rs772348346 Chr.7: 100554114-100554114 on Build GRCh38
rs61742738 Chr.7: 100554096-100554096 on Build GRCh38
rs764627681 Chr.7: 100554043-100554043 on Build GRCh38
rs995350902 Chr.7: 100554030-100554030 on Build GRCh38
rs754504227 Chr.7: 100554095-100554095 on Build GRCh38
rs777568708 Chr.7: 100554217-100554217 on Build GRCh38
rs756138442 Chr.7: 100554213-100554213 on Build GRCh38
rs750685361 Chr.7: 100554067-100554067 on Build GRCh38
rs143496851 Chr.7: 100554111-100554111 on Build GRCh38
rs898260314 Chr.7: 100554363-100554363 on Build GRCh38
rs748209980 Chr.7: 100554118-100554118 on Build GRCh38
rs776017832 Chr.7: 100554155-100554156 on Build GRCh38
rs1028136438 Chr.7: 100554072-100554072 on Build GRCh38
rs757654573 Chr.7: 100554065-100554065 on Build GRCh38
rs766172524 Chr.7: 100554172-100554172 on Build GRCh38
rs939729967 Chr.7: 100554291-100554291 on Build GRCh38
rs770395317 Chr.7: 100554300-100554300 on Build GRCh38
rs779835238 Chr.7: 100554088-100554088 on Build GRCh38
rs931570116 Chr.7: 100554303-100554303 on Build GRCh38
rs546170476 Chr.7: 100554384-100554384 on Build GRCh38
rs774696544 Chr.7: 100554304-100554306 on Build GRCh38
rs749263005 Chr.7: 100554228-100554228 on Build GRCh38
rs758134300 Chr.7: 100554078-100554078 on Build GRCh38
rs748151391 Chr.7: 100554100-100554100 on Build GRCh38
rs78834980 Chr.7: 100554153-100554153 on Build GRCh38
rs754346092 Chr.7: 100554051-100554051 on Build GRCh38
rs887315348 Chr.7: 100554343-100554343 on Build GRCh38
rs200779668 Chr.7: 100554168-100554168 on Build GRCh38
rs140340569 Chr.7: 100554436-100554436 on Build GRCh38
rs1013395067 Chr.7: 100554468-100554468 on Build GRCh38
rs758827052 Chr.7: 100554167-100554167 on Build GRCh38
rs776532327 Chr.7: 100554130-100554130 on Build GRCh38
rs528795430 Chr.7: 100554362-100554362 on Build GRCh38
rs764378309 Chr.7: 100554169-100554169 on Build GRCh38
rs749122018 Chr.7: 100554218-100554218 on Build GRCh38
rs780760584 Chr.7: 100554195-100554195 on Build GRCh38
rs761782755 Chr.7: 100554294-100554294 on Build GRCh38
rs773821277 Chr.7: 100554309-100554309 on Build GRCh38
rs200992157 Chr.7: 100554110-100554110 on Build GRCh38
rs746581473 Chr.7: 100554094-100554094 on Build GRCh38
rs1004330075 Chr.7: 100554376-100554376 on Build GRCh38
rs749070028 Chr.7: 100554112-100554112 on Build GRCh38
rs577955527 Chr.7: 100554256-100554256 on Build GRCh38
rs772235085 Chr.7: 100554286-100554286 on Build GRCh38
rs751231873 Chr.7: 100554192-100554192 on Build GRCh38
rs368144368 Chr.7: 100554255-100554255 on Build GRCh38
rs3115583 Chr.7: 100554125-100554125 on Build GRCh38

View Full Product Details